Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Progesterone Receptor Antibodies

          Invitrogen

          Progesterone Receptor Monoclonal Antibody (Alpha PR6)

          Advanced Verification
          19 Published Figures
          51 References
          View all (117) Progesterone Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Progesterone Receptor Monoclonal Antibody (Alpha PR6)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (19)
          • Advanced Verification (2)
          Progesterone Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Progesterone Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 25

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Progesterone Receptor Antibody (MA1-411) in ICC/IF

          Immunofluorescent analysis of Progesterone Receptor (green) in T47D cells untreated (left) or stimulated with 100nm promegestone, R5020, for 1 hour (right). The cells were fixed with formalin for 15 minutes, permeabilized with 0.1% Triton X-100 in TBS for 10 minutes, and blocked with 3% Blocker BSA (Product # 37525) for 15 minutes at room temperature. Cells were stained with Progesterone Receptor mouse mono... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Progesterone Receptor Antibody in Immunocytochemistry (ICC/IF)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Western Blot (WB)
          Progesterone Receptor Antibody in Western Blot (WB)
          Progesterone Receptor Antibody in Western Blot (WB)
          Progesterone Receptor Antibody in Immunocytochemistry (ICC/IF)
          Progesterone Receptor Antibody in Immunocytochemistry (ICC/IF)
          Progesterone Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Immunohistochemistry (IHC)
          Progesterone Receptor Antibody in Western Blot (WB)
          Progesterone Receptor Antibody in Western Blot (WB)
          Progesterone Receptor Antibody in Western Blot (WB)
          Progesterone Receptor Antibody
          Progesterone Receptor Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Progesterone Receptor Monoclonal Antibody (Alpha PR6)

          Product Details

          MA1-411

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 36 publications 36 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          5 µg/mL
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          20 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Guinea pig, Human, Mouse, Sheep, Rabbit, Rat

          Published species

          Camelid, Chicken, Human, Mouse, Non-human primate, Rat, Sheep

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          Alpha PR6

          Immunogen

          Progesterone receptor purified from chick oviduct cytosol.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2283820

          Product Specific Information

          MA1-411 reacts with high affinity to the B form of the progesterone receptor from human, mouse, rat, rabbit, bovine, ovine, chicken, and guinea pig tissues. This antibody does not cross-react with estrogen receptor or glucocorticoid receptor.

          MA1-411 has been successfully used in Western blot, immunocytochemical, immunohistochemical and immunoprecipitation procedures. By Western blot, this antibody detects a 110 kDa protein representing the B form of the PR in chickens."

          The MA1-411 immunogen is progesterone receptor purified from chick oviduct cytosol.

          Reconstitute with PBS.

          Target Information

          The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintenance of the endometrium, maintenance of pregnancy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: nuclear receptor subfamily 3 group C member 3; pot. alt. progesterone receptor; putative; PR; progesterone receptor; progesterone receptor A; progesterone-receptor; steroid hormone receptor; unnamed protein product

          View more View less

          Gene Aliases: 9930019P03Rik; NR3C3; PR; PR-A; PR-B

          View more View less

          UniProt ID: (Rabbit) P06186, (Chicken) P07812, (Human) P06401, (Mouse) Q00175, (Rat) Q63449

          View more View less

          Entrez Gene ID: (Sheep) 443444, (Rabbit) 100009094, (Bovine) 100848466, (Chicken) 396198, (Human) 5241, (Guinea pig) 100724945, (Mouse) 18667, (Rat) 25154

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding transcription factor activity, sequence-specific DNA binding steroid hormone receptor activity RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding zinc ion binding enzyme binding Hsp70 protein binding chromatin DNA binding sequence-specific DNA binding Hsp90 protein binding steroid hormone binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription coactivator binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding receptor binding steroid binding protein binding estrogen response element binding identical protein binding ATPase binding sequence-specific double-stranded DNA binding transcription regulatory region sequence-specific DNA binding hormone binding
          Process(es)
          regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter intracellular steroid hormone receptor signaling pathway response to progesterone steroid hormone mediated signaling pathway response to estrogen signal transduction cell-cell signaling positive regulation of gene expression negative regulation of gene expression negative regulation of phosphorylation positive regulation of transcription from RNA polymerase II promoter progesterone receptor signaling pathway maintenance of protein location in nucleus ovulation from ovarian follicle epithelial cell maturation positive regulation of cell proliferation female gonad development positive regulation of smooth muscle cell migration ovulation cycle process mammary gland development paracrine signaling response to cocaine negative regulation of apoptotic process positive regulation of transcription, DNA-templated lung alveolus development regulation of epithelial cell proliferation female mating behavior tertiary branching involved in mammary gland duct morphogenesis negative regulation of granulosa cell apoptotic process luteinization diterpenoid metabolic process estrous cycle negative regulation of transcription, DNA-templated smooth muscle relaxation of the bladder outlet cellular response to gonadotropin stimulus cellular response to follicle-stimulating hormone stimulus cellular response to luteinizing hormone stimulus cellular response to estrogen stimulus response to ketone cellular response to wortmannin
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-sf8lr:80/100.66.74.151:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline