Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Prolactin Receptor Antibodies

          Invitrogen

          Prolactin Receptor Monoclonal Antibody (U5)

          Advanced Verification
          7 Published Figures
          36 References
          View all (38) Prolactin Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Prolactin Receptor Monoclonal Antibody (U5)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (7)
          • Advanced Verification (1)
          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Prolactin Receptor Antibody (MA1-610) in IHC

          Mammary gland tissue from lactating light horse mare (equine) was incubated in MA1-610 anti-prolactin receptor monoclonal mouse antibody at 1:100 for 1 hour at 23°C and for 24 hours at 4°C. Second antibody was biotinylated goat anti-mouse IgG at 23°C for 1 hour. Development using avidin-biotin complex 23°C for 1 hour plus 3, 3-diaminobenzadine for 15 min and counterstained with haematoxylin. Notes: Antigen ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Prolactin Receptor Antibody in Western Blot (WB)
          Prolactin Receptor Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Prolactin Receptor Antibody in Immunohistochemistry (IHC)
          Prolactin Receptor Antibody in Western Blot (WB)
          Prolactin Receptor Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Prolactin Receptor Monoclonal Antibody (U5)

          Product Details

          MA1-610

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 15 publications 15 publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 2 publications 2 publications

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Horse, Human, Mouse, Pig, Rabbit, Rat

          Published species

          Baboon, Bovine, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          U5

          Immunogen

          Purified rat liver PRL receptor.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          1 mg/mL once reconstituted

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2171291

          Product Specific Information

          MA1-610 detects prolactin (PRL) receptor from rat, human, rabbit, horse, and pig tissues.

          MA1-610 has been successfully used in Western blot, immunohistochemistry, immunocytochemistry, immunoprecipitation, immunofluorescence, and FACS procedures. By Western blot, this antibody detects an ~40 kDa protein and an ~100 kDa protein representing both the short and long forms of the PRL receptor, respectively, in various tissues. Immunohistochemical staining of PRL receptor in rat Islets of Langerhans with MA1-610 results in a staining pattern consistent with cytoplasmic vesicle staining. MA1-610, after biotinylation, has been used to detect human lymphocyte PRL receptor by FACS. MA1-610 does not inhibit the interaction of prolactin with the receptor.

          The MA1-610 immunogen is purified PRL receptor from rat liver. This antibody has been shown to react with the extracellular portion of the receptor, but not in the ligand binding domain. MA1-610 does not inhibit the interaction of prolactin with the receptor.

          Reconstitute with 100 µL PBS.

          Target Information

          Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor. Immunoreactive PRL receptor (PRLR), a member of the cytokine receptor family, varies in size (short and long forms) with tissue source and species, from ~40 kDa to 100 kDa. PRLR consists of at least three separate domains: an extracellular region with 5 cysteines which contains the prolactin binding site, a single transmembrane domain and a cytoplasmic region, the length of which appears to influence ligand binding and regulate cellular function.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cytokine receptor; cytokine receptor; PRLRSFA; cytokine receptor; PRLRSFB; delta-S1 PRLr; hPRL receptor; lactogen receptor; lactogen receptor precursor; Prl R; PRL-R; prolactin receptor; prolactin receptor 2 precursor; prolactin receptor precursor; prolactin receptor related sequence 1; prolactin receptor short isoform; pseudo-prolactin receptor precursor; secreted prolactin binding protein; unnamed protein product

          View more View less

          Gene Aliases: HPRL; hPRLrI; MFAB; pPRLR-SF; Pr-1; Pr-3; Prlr-rs1; RATPRLR; RI-PRLR

          View more View less

          UniProt ID: (Rabbit) P14787, (Pig) Q6JTA8, (Human) P16471, (Rat) P05710, (Mouse) Q08501

          View more View less

          Entrez Gene ID: (Rabbit) 100009046, (Pig) 414916, (Horse) 100053793, (Human) 5618, (Rat) 24684, (Mouse) 19116

          View more View less

          Function(s)
          metal ion binding cytokine receptor activity prolactin receptor activity protein binding lipid binding peptide hormone binding protein kinase binding cytokine binding transmembrane signal receptor
          Process(es)
          prolactin signaling pathway oncostatin-M-mediated signaling pathway leukemia inhibitory factor signaling pathway steroid biosynthetic process activation of transmembrane receptor protein tyrosine kinase activity embryo implantation lactation positive regulation of cell proliferation cytokine-mediated signaling pathway positive regulation of B cell proliferation activation of Janus kinase activity negative regulation of apoptotic process cellular response to granulocyte macrophage colony-stimulating factor stimulus positive regulation of cold-induced thermogenesis JAK-STAT cascade response to bacterium regulation of cell adhesion regulation of epithelial cell differentiation positive regulation of protein autophosphorylation mammary gland epithelial cell differentiation prostate gland growth mammary gland alveolus development mammary gland epithelium development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch