Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Actin Antibodies

          Invitrogen

          Actin Monoclonal Antibody (mAbGEa)

          Advanced Verification
          72 Published Figures
          154 References
          View all (22) Actin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Actin Monoclonal Antibody (mAbGEa)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          • Published Figures (72)
          • Advanced Verification (2)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 82

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Actin Antibody (MA1-744) in ICC/IF

          Immunofluorescence analysis of Actin, cytoplasmic 1 was performed using 70% confluent log phase DU 145 cells. The cells were fixed with 4% paraformaldehyde for 5 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Actin Monoclonal Antibody (mAbGEa) (Product # MA1-744) at 1:100 dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor Pl... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Actin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Actin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Immunohistochemistry (IHC)
          Actin Antibody in Immunohistochemistry (IHC)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Western Blot (WB)
          Actin Antibody in Immunoprecipitation (IP)
          Actin Antibody
          Actin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Actin Monoclonal Antibody (mAbGEa)

          Product Details

          MA1-744

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:10,000
          View 138 publications 138 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Bovine, Fruit fly, Human, Mouse, Sheep, Plant, Rat, Xenopus, Yeast, Zebrafish

          Published species

          Bacteria, Bovine, Fruit fly, Goat, Human, Insect, Mouse, Pig, Plant, Rat, Xenopus, Yeast

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          mAbGEa

          Immunogen

          Purified Arabidopsis actin protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          ascites

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2223496

          Product Specific Information

          MA1-744 detects actin protein in human, mouse, rat, bovine, sheep, drosophila, zebra fish (Danio rerio), yeast, arabidopsis and xenopus samples. This antibody has shown cross-reactivity with actin 1, 2, 3, 4, 7, 8, 11 and 12.

          MA1-744 has successfully been used in Western blot and ELISA procedures. By Western blot, this antibody detects a 45 kDa protein representing actin.

          The MA1-744 immunogen is purified actin protein from Arabidopsis.

          Target Information

          Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 5C actin; A-X actin; A-X actin-like protein; Act5C-PA; Act5C-PB; Act5C-PC; Act5C-PD; Act5C-PE; ACTA1; ACTA2; ACTB; ACTC; ACTG2; actin; actin 5 C; actin 5c; actin A1; actin alpha 1; actin alpha cardiac; Actin is a protein with a high degree of evolutionary conservation that undergoes polymerization to generate filaments, which subsequently assemble into interconnected networks within the cytoplasm of cells.; actin, alpha 2, smooth muscle, aorta; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; actin, beta, cytoplasmic; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin-like protein; actin5C; alfa sma; alpha 1 skeletal muscle actin; alpha actin 2; alpha smooth muscle actin; alpha-actin; alpha-actin cardiac; alpha-actin-1; alpha-actin-2; alpha-actin-3; alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; anon-fast-evolving-2D2; aSMA; aSmooth Muscle Actin; Atcb; beta cytoskeletal actin; beta-actin; beta-actin FE-3; cell growth-inhibiting gene 46 protein; cellular cytoskeletal beta-actin; CG4027-PA; CG4027-PB; CG4027-PC; CG4027-PD; CG4027-PE; cytoplasmic beta-actin; cytoplasmic protein; cytoskeletal gamma-actin; Dm-actin5C; epididymis luminal protein 176; F-actin; G-actin; gamma-2-actin; gamma-actin; gamma-enteric smooth muscle actin; GIG46; I(2)-actin; melanoma X-actin; nemaline myopathy type 3; pan actin; PS1TP5-binding protein 1; RE02927p; skeletal muscle alpha-actin; sma; smooth muscle alpha-actin; smooth muscle gamma-actin; unnamed protein product

          View more View less

          Gene Aliases: 0610041G09Rik; A; a-SMA; A4V404_DROME; Ac5C; ACT; act 5C; Act-4; Act-5C; ACT1_DROME; Act4; Act5; ACT5C; ACTA; Acta-2; acta1; ACTA3; actb; ACTC; Actc-1; ACTE; ACTG; ACTGE; Actin; Actin/BAP47; Actin5C; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; Actx; alphaSMA; anon-EST:fe2D2; ArpA; ASD5; ASMA; BAP47; beta-actin; beta-actin/Bap47; BKRNS; BNS; BRWS1; CFTD; CFTD1; CFTDM; CG4027; chrX:5748184..5748304; CMD1R; CMH11; CMYO2A; CMYO2B; CMYO2C; CMYP2A; CMYP2B; CMYP2C; CSMH; cyt5C; DDS1; DFNA20; DFNA26; Dmel\CG4027; Dmel_CG4027; E430023M04Rik; E51; F-actin; G-actin; HEL-176; l(1)G0009; l(1)G0010; l(1)G0025; l(1)G0079; l(1)G0117; l(1)G0177; l(1)G0245; l(1)G0330; l(1)G0420; l(1)G0486; LVNC4; M32055; MMIHS5; MPFD; NEM1; NEM2; NEM3; PS1TP5BP1; SHPM; SMAalpha; SMalphaA; SMDYS; SMGA; T11; THC8; VSCM; VSCM1; wu:fb63d03; XELAEV_18029136mg; XELAEV_18045052mg

          View more View less

          UniProt ID: (Human) P68133, (Bovine) P68138, (Bovine) P62739, (Human) P62736, (Bovine) P60712, (Human) P60709, (Sheep) P60713, (Bovine) Q3ZC07, (Human) P68032, (Human) P63261, (Bovine) P63258, (Bovine) Q5E9B5, (Human) P63267, (Fruit fly) P10987, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737, (Rat) P60711, (Mouse) P60710, (Rat) P68035, (Mouse) P68033, (Mouse) P63260, (Rat) P63259, (Rat) P63269, (Mouse) P63268, (Xenopus) O93400

          View more View less

          Entrez Gene ID: (Human) 58, (Bovine) 281592, (Sheep) 101107662, (Bovine) 515610, (Sheep) 101121051, (Human) 59, (Bovine) 280979, (Human) 60, (Sheep) 443052, (Bovine) 533219, (Sheep) 101111554, (Human) 70, (Human) 71, (Bovine) 404122, (Sheep) 443340, (Sheep) 101114256, (Bovine) 281595, (Human) 72, (Fruit fly) 31521, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Rat) 81822, (Mouse) 11461, (Rat) 29275, (Mouse) 11464, (Mouse) 11465, (Rat) 287876, (Rat) 25365, (Mouse) 11468, (Xenopus) 432314, (Zebrafish) 322509, (Xenopus) 398459

          View more View less

          Function(s)
          structural constituent of cytoskeleton protein binding ATP binding ATPase activity myosin binding ADP binding protein kinase binding kinesin binding Tat protein binding identical protein binding nitric-oxide synthase binding nitric-oxide synthase regulator activity nucleosomal DNA binding tau protein binding structural constituent of postsynaptic actin cytoskeleton transporter regulator activity microfilament motor activity profilin binding ubiquitin protein ligase binding nucleotide binding actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          muscle contraction positive regulation of gene expression skeletal muscle thin filament assembly skeletal muscle fiber development mesenchyme migration regulation of blood pressure response to virus vascular smooth muscle contraction glomerular mesangial cell development cytoskeleton organization positive regulation of hepatic stellate cell migration positive regulation of hepatic stellate cell contraction positive regulation of ERK1 and ERK2 cascade cellular response to transforming growth factor beta stimulus juxtaglomerular apparatus development positive regulation of hepatic stellate cell activation morphogenesis of a polarized epithelium establishment or maintenance of cell polarity regulation of transmembrane transporter activity negative regulation of protein binding adherens junction assembly apical protein localization synaptic vesicle endocytosis regulation of norepinephrine uptake protein localization to adherens junction cellular response to cytochalasin B postsynaptic actin cytoskeleton organization regulation of protein localization to plasma membrane chromatin remodeling regulation of transcription from RNA polymerase II promoter microtubule nucleation axonogenesis positive regulation of cell proliferation substantia nigra development regulation of mitotic metaphase/anaphase transition maintenance of blood-brain barrier regulation of apoptotic process positive regulation of T cell differentiation negative regulation of cell differentiation positive regulation of cell differentiation positive regulation of myoblast differentiation positive regulation of transcription, DNA-templated cell motility positive regulation of norepinephrine uptake regulation of cell cycle regulation of G0 to G1 transition platelet aggregation regulation of transepithelial transport regulation of synaptic vesicle endocytosis positive regulation of stem cell population maintenance positive regulation of double-strand break repair via homologous recombination regulation of G1/S transition of mitotic cell cycle regulation of double-strand break repair positive regulation of double-strand break repair regulation of nucleotide-excision repair actin filament organization actin filament-based movement actomyosin structure organization actin-myosin filament sliding cardiac myofibril assembly cardiac muscle tissue morphogenesis heart contraction angiogenesis positive regulation of cell migration regulation of stress fiber assembly regulation of focal adhesion assembly positive regulation of wound healing tight junction assembly protein localization to bicellular tight junction response to ethanol cellular response to interleukin-6 cellular response to acetaldehyde mitotic cytokinesis sperm individualization maintenance of protein location in cell tube formation response to mechanical stimulus cellular response to potassium ion skeletal muscle fiber adaptation response to steroid hormone circadian rhythm dynein-driven meiotic oscillatory nuclear movement response to immobilization stress retina development in camera-type eye cellular response to electrical stimulus response to xenobiotic stimulus negative regulation of apoptotic process cardiac muscle contraction cytoplasmic actin-based contraction involved in cell motility sarcomere organization cellular response to interferon-gamma response to calcium ion blood circulation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch