Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Presenilin 1 Antibodies

          Invitrogen

          Presenilin 1 Monoclonal Antibody (APS 11)

          1 Reference
          View all (31) Presenilin 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Presenilin 1 Monoclonal Antibody (APS 11)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          Presenilin 1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Presenilin 1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Presenilin 1 Antibody (MA1-751) in ICC/IF

          Immunocytochemical staining of PS1 in mouse fibroblasts using Product # MA1-751. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Presenilin 1 Antibody in Immunocytochemistry (ICC/IF)
          Presenilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Presenilin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Presenilin 1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Presenilin 1 Monoclonal Antibody (APS 11)

          Product Details

          MA1-751

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:100-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          0.75 µg/mL
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          4.5 µg/mL
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          APS 11

          Immunogen

          Synthetic peptide corresponding to residues C H(21) L S N T V R S Q N D N R E(34) of human PS1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          200 µg

          Purification

          Protein G

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325074

          Product Specific Information

          MA1-751 detects presenilin 1 protein (PS1) from mouse, rat, human, and nonhuman primate samples. No cross-reactivity is seen with presenilin 2.

          MA1-751 has successfully been used immunofluorescence, immunocytochemistry, immunohistochemistry, Western blot , and ELISA procedures. By Western blot, this antibody detects an ~28 kDa protein representing PS1 N-terminus cleavage product from ST15 cell lysate transfected with human PS1. In formalin-fixed, paraffin embedded sections of human brain, this antibody showed strong staining of both the plaque core and dystrophic neurites.

          The MA1-751 immunogen is a synthetic peptide corresponding to residues C H(21) L S N T V R S Q N D N R E(34) of human PS1.

          Target Information

          Presenilin1 was initially identified a marker of susceptibility to early-onset Alzheimer's disease. In addition to PEN2, nicastrin and APH-1, Presenilin1 forms the -gamma-secretase protein complex, a membrane-bound aspartyl protease that can cleave certain proteins at peptide bonds buried within the hydrophobic environment of the lipid bilayer. This cleavage is responsible for a key step in signaling from several cell-surface receptors and is thought to be required for the generation of the neurotoxic amyloid peptides that are central to the pathogenesis of Alzheimer's disease. Like the tumor necrosis factor-alpha-converting enzyme (TACE) and the beta-site cleavage enzyme (BACE) protease families, -gamma-secretase will cleave the amyloid precursor protein (APP), but within the intramembrane region of APP, resulting in either the non-toxic p3 (from the alpha and -gamma cleavage site) or the toxic Abeta amyloid peptide (from the beta and -gamma cleavage site). It is thought that accumulation of the Abeta peptide is the precursor to Alzheimer's disease. Multiple isoforms of presenilin1 are known to exist.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Alzheimer Disease 3; familial Alzheimer Disease; Minilin; presenilin1; PS-1; S182; S182 protein; Senilin 1; unnamed protein product

          View more View less

          Gene Aliases: ACNINV3; AD3; Ad3h; CMD1U; FAD; PS-1; PS1; PSNL1; S182

          View more View less

          UniProt ID: (Human) P49768, (Rat) P97887, (Mouse) P49769

          View more View less

          Entrez Gene ID: (Human) 5663, (Rat) 29192, (Mouse) 19164

          View more View less

          Function(s)
          endopeptidase activity aspartic-type endopeptidase activity calcium channel activity protein binding beta-catenin binding peptidase activity PDZ domain binding aspartic endopeptidase activity, intramembrane cleaving cadherin binding ATPase binding binding, bridging growth factor receptor binding
          Process(es)
          autophagosome assembly negative regulation of transcription from RNA polymerase II promoter blood vessel development cell fate specification somitogenesis neuron migration positive regulation of receptor recycling negative regulation of protein phosphorylation positive regulation of protein phosphorylation heart looping positive regulation of L-glutamate transport hematopoietic progenitor cell differentiation astrocyte activation involved in immune response T cell activation involved in immune response myeloid leukocyte differentiation neural retina development negative regulation of protein kinase activity membrane protein ectodomain proteolysis mitochondrial transport cellular calcium ion homeostasis autophagy cellular response to DNA damage stimulus response to oxidative stress Notch signaling pathway Notch receptor processing brain development heart development learning or memory memory glycoprotein biosynthetic process post-embryonic development gene expression regulation of gene expression positive regulation of gene expression negative regulation of gene expression regulation of neuron projection development protein transport choline transport protein processing cerebellum development cerebral cortex cell migration Cajal-Retzius cell differentiation dorsal/ventral neural tube patterning cerebral cortex development neurogenesis neuron differentiation embryonic limb morphogenesis forebrain development membrane protein intracellular domain proteolysis positive regulation of proteasomal ubiquitin-dependent protein catabolic process endoplasmic reticulum calcium ion homeostasis positive regulation of tumor necrosis factor production beta-amyloid formation segmentation intracellular signal transduction locomotion negative regulation of epidermal growth factor receptor signaling pathway eye photoreceptor cell development amyloid precursor protein metabolic process amyloid precursor protein catabolic process myeloid dendritic cell differentiation positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of MAP kinase activity negative regulation of neuron apoptotic process skin morphogenesis positive regulation of glycolytic process positive regulation of protein kinase activity positive regulation of transcription, DNA-templated astrocyte activation regulation of synaptic plasticity thymus development neuron development skeletal system morphogenesis brain morphogenesis beta-amyloid metabolic process epithelial cell proliferation negative regulation of axonogenesis synapse organization positive regulation of coagulation T cell receptor signaling pathway neuron apoptotic process smooth endoplasmic reticulum calcium ion homeostasis protein maturation regulation of synaptic transmission, glutamatergic calcium ion homeostasis regulation of resting membrane potential regulation of canonical Wnt signaling pathway positive regulation of dendritic spine development calcium ion transmembrane transport cell-cell adhesion regulation of synaptic vesicle cycle L-glutamate import across plasma membrane regulation of postsynapse organization protein catabolic process at postsynapse cellular response to beta-amyloid positive regulation of amyloid fibril formation neuron projection maintenance negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process negative regulation of apoptotic signaling pathway positive regulation of protein import into nucleus synaptic vesicle transport long-term synaptic potentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-v8ns5:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0