Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PPAR alpha Antibodies

          Invitrogen

          PPAR alpha Monoclonal Antibody (3B6/PPAR)

          Advanced Verification
          14 Published Figures
          27 References
          View all (31) PPAR alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PPAR alpha Monoclonal Antibody (3B6/PPAR)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (14)
          • Advanced Verification (1)
          PPAR alpha Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PPAR alpha Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPAR alpha Antibody (MA1-822) in WB

          Western blot analysis of PPAR alpha was performed by loading 25 µg of Hela (Lane 1), Jurkat (Lane 2), and NIH-3T3 cell lysates (Lane 3) and a molecular weight protein ladder onto an SDS polyacrylamide gel. Proteins were transferred to a PVDF membrane and blocked with a blocking buffer at 4°C overnight. The membrane was probed with a PPAR alpha monoclonal antibody (Product # MA1-822) at a dilution of 1:1000 (Hela and Jurkat) and 1:500 (NIH-3T3) overnight at 4°C, washed in TBST, and probed with an HRP-conjugated secondary antibody for 1 hr a... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Flow Cytometry (Flow)
          PPAR alpha Antibody in Flow Cytometry (Flow)
          PPAR alpha Antibody in Flow Cytometry (Flow)
          PPAR alpha Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PPAR alpha Antibody in Immunohistochemistry (IHC)
          PPAR alpha Antibody in Immunohistochemistry (IHC)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in Western Blot (WB)
          PPAR alpha Antibody in ChIP Assay (ChIP)
          PPAR alpha Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA1-822

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2-5 µg/mL
          View 18 publications 18 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          1 µg / 10^6 cells
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 2 publications 2 publications

          ChIP assay (ChIP)

          Assay-dependent
          View 1 publication 1 publication

          Gel Shift (EMSA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Bovine, Human, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3B6/PPAR

          Immunogen

          Purified recombinant PPAR alpha protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2165745

          Product Specific Information

          MA1-822 detects peroxisome proliferator activated receptors (PPAR) alpha from human, mouse, and rat tissues.

          MA1-822 has been successfully used in Western blot, Gel Shift and immunoprecipitation procedures. By Western blot, this antibody detects a ~52 kDa protein which corresponds to PPAR alpha from mouse adipose tissue extract. This antibody detects some non-specific bands on NIH-3T3 cell lysates.

          The MA1-822 immunogen is purified recombinant PPAR alpha protein.

          Target Information

          Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed peroxisome proliferator activated receptors (PPAR's). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPAR's are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY-14,643, as well as by some fatty acids. It has also been shown that PPAR's can induce transcription of acyl coenzyme A oxidase & cytochrome P450 (CYP450) A6 through interaction with specific response elements. PPAR, like several other nuclear hormone receptors, heterodimerizes with retinoid X receptor (RXR) alpha.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: MGC2237; MGC2452; nuclear receptor subfamily 1 group C member 1; peroxisome proliferative activated receptor, alpha; peroxisome proliferator; peroxisome proliferator activated receptor; ppar alpha; PPAR-alpha; PPARA; PPARalpha; unnamed protein product

          View more View less

          Gene Aliases: 4933429D07Rik; hPPAR; NR1C1; PPAR; PPAR-alpha; PPARalpha

          View more View less

          UniProt ID: (Human) Q07869, (Mouse) P23204, (Rat) P37230

          View more View less

          Entrez Gene ID: (Human) 5465, (Mouse) 19013, (Rat) 25747

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding bacterial-type RNA polymerase transcriptional activator activity, sequence-specific DNA binding transcription coactivator binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding steroid hormone receptor activity RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding zinc ion binding lipid binding phosphatase binding protein domain specific binding mitogen-activated protein kinase kinase kinase binding ubiquitin conjugating enzyme binding signaling receptor activity sequence-specific DNA binding macromolecular complex binding NFAT protein binding RNA polymerase II sequence-specific DNA binding transcription factor binding MDM2/MDM4 family protein binding DNA-binding transcription factor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter response to hypoxia regulation of transcription, DNA-templated fatty acid metabolic process heart development response to nutrient lactation cellular response to starvation hormone-mediated signaling pathway regulation of cellular ketone metabolic process negative regulation of macrophage derived foam cell differentiation negative regulation of cholesterol storage regulation of fatty acid metabolic process cell differentiation negative regulation of transforming growth factor beta receptor signaling pathway intracellular steroid hormone receptor signaling pathway intracellular receptor signaling pathway positive regulation of fatty acid beta-oxidation negative regulation of appetite response to insulin circadian regulation of gene expression behavioral response to nicotine peroxisome proliferator activated receptor signaling pathway lipoprotein metabolic process regulation of circadian rhythm response to ethanol negative regulation of blood pressure negative regulation of glycolytic process positive regulation of transcription, DNA-templated positive regulation of fatty acid metabolic process positive regulation of transcription from RNA polymerase II promoter nitric oxide metabolic process positive regulation of fatty acid oxidation positive regulation of lipid biosynthetic process negative regulation of inflammatory response defense response to virus negative regulation of protein kinase B signaling negative regulation of cell growth involved in cardiac muscle cell development cellular response to fructose stimulus negative regulation of cytokine production involved in inflammatory response negative regulation of pri-miRNA transcription from RNA polymerase II promoter negative regulation of leukocyte cell-cell adhesion negative regulation of reactive oxygen species biosynthetic process negative regulation of hepatocyte apoptotic process regulation of fatty acid transport positive regulation of ATP biosynthetic process glucose metabolic process lipid metabolic process fatty acid beta-oxidation response to stress signal transduction epidermis development response to glucose gene expression regulation of gene expression protein ubiquitination wound healing positive regulation of gluconeogenesis enamel mineralization response to fatty acid positive regulation of transformation of host cell by virus negative regulation of cardiac muscle hypertrophy negative regulation of cell growth
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch