Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Calcium Sensing Receptor Antibodies

          Invitrogen

          Calcium Sensing Receptor Monoclonal Antibody (5C10, ADD)

          Advanced Verification
          53 Published Figures
          70 References
          View all (27) Calcium Sensing Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Calcium Sensing Receptor Monoclonal Antibody (5C10, ADD)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Published Figures (53)
          • Advanced Verification (1)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Calcium Sensing Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 59

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calcium Sensing Receptor Antibody (MA1-934) in IHC (P)

          Immunohistochemistry was performed on normal biopsies of deparaffinized Human kidney tissue. To expose target proteins, heat induced antigen retrieval was performed using 10mM sodium citrate (pH6.0) buffer, microwaved for 8-15 minutes. Following antigen retrieval tissues were blocked in 3% BSA-PBS for 30 minutes at room temperature. Tissues were then probed at a dilution of 1:100 with a mouse monoclonal ant... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calcium Sensing Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calcium Sensing Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calcium Sensing Receptor Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunocytochemistry (ICC/IF)
          Calcium Sensing Receptor Antibody in Immunocytochemistry (ICC/IF)
          Calcium Sensing Receptor Antibody in Immunocytochemistry (ICC/IF)
          Calcium Sensing Receptor Antibody in Immunocytochemistry (ICC/IF)
          Calcium Sensing Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Calcium Sensing Receptor Antibody in Immunocytochemistry (ICC/IF)
          Calcium Sensing Receptor Antibody in Immunocytochemistry (ICC/IF)
          Calcium Sensing Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Calcium Sensing Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Calcium Sensing Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Calcium Sensing Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Calcium Sensing Receptor Antibody in Western Blot, Immunohistochemistry (Paraffin) (WB, IHC (P))
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (IHC)
          Calcium Sensing Receptor Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Calcium Sensing Receptor Antibody in Flow Cytometry (Flow)
          Calcium Sensing Receptor Antibody in Flow Cytometry (Flow)
          Calcium Sensing Receptor Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Calcium Sensing Receptor Monoclonal Antibody (5C10, ADD)

          Product Details

          MA1-934

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 32 publications 32 publications

          Immunohistochemistry (IHC)

          -
          View 17 publications 17 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100
          View 5 publications 5 publications

          Immunohistochemistry (Frozen) (IHC (F))

          2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          -
          View 12 publications 12 publications

          Flow Cytometry (Flow)

          -
          View 3 publications 3 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Rat

          Published species

          Human, Mouse, Pig, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          5C10, ADD

          Immunogen

          Synthetic peptide corresponding to residues (214) A D D D Y G R P G I E K F RE E A E E R D I (235) of human calcium sensing receptor.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325374

          Product Specific Information

          MA1-934 detects human, bovine, mouse, and rat calcium sensing receptor.

          MA1-934 has been successfully used in Western blot, immunohistochemistry, and ELISA procedures. By Western blot, this antibody detects an ~ 150 and 130 kDa proteins from human HEK-293 cells.This antibody has also been used in immunohistochemical on frozen bovine parathyroid gland and rat thyroid gland.

          The MA1-934 immunogen is a synthetic peptide corresponding to residues (214) A D D D Y G R P G I E K F RE E A E E R D I (235) of human calcium sensing receptor.

          Target Information

          Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ca sensing receptor; calcium receptor|CaR; Calcium-sensing receptor (hypocalciuric hypercalcemia 1, severe neonatal hyperparathyroidism); CaSR antibody; cation sensing receptor; extracellular domain; G protein-coupled membrane receptor; G protein-coupled receptor, family C, group 2, member A; G-protein coupled receptor; parathyroid Ca(2+)-sensing receptor 1; parathyroid Cell calcium-sensing receptor; parathyroid cell calcium-sensing receptor 1; PCaR1; unnamed protein product

          View more View less

          Gene Aliases: BoPCaR1; CAR; EIG8; FHH; FIH; GPRC2A; hCasR; HHC; HHC1; HYPOC1; NSHPT; PCAR1; RaKCaR

          View more View less

          UniProt ID: (Bovine) P35384, (Human) P41180, (Mouse) Q9QY96, (Rat) P48442

          View more View less

          Entrez Gene ID: (Bovine) 281038, (Human) 846, (Mouse) 12374, (Rat) 24247

          View more View less

          Function(s)
          G-protein coupled receptor activity calcium ion binding protein binding amino acid binding protein homodimerization activity integrin binding protein kinase binding identical protein binding ion channel binding G-protein coupled receptor
          Process(es)
          ossification detection of calcium ion calcium ion transport cellular calcium ion homeostasis G-protein coupled receptor signaling pathway JNK cascade positive regulation of cell proliferation positive regulation of gene expression positive regulation of ATPase activity vasodilation calcium ion import response to ischemia adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway chemosensory behavior anatomical structure morphogenesis response to metal ion positive regulation of insulin secretion bile acid secretion cellular response to hepatocyte growth factor stimulus positive regulation of vasoconstriction branching morphogenesis of an epithelial tube positive regulation of positive chemotaxis response to calcium ion regulation of calcium ion transport fat pad development positive regulation of ERK1 and ERK2 cascade cellular response to vitamin D cellular response to glucose stimulus cellular response to low-density lipoprotein particle stimulus cellular response to hypoxia response to fibroblast growth factor positive regulation of calcium ion import regulation of presynaptic membrane potential positive regulation of NLRP3 inflammasome complex assembly cellular response to peptide chloride transmembrane transport positive regulation of intracellular signal transduction adenylate cyclase-activating G protein-coupled cAMP receptor signaling pathway detection of chemical stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch