Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • p23 Antibodies

          Invitrogen

          p23 Monoclonal Antibody (JJ3)

          Advanced Verification
          15 Published Figures
          35 References
          View all (43) p23 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite p23 Monoclonal Antibody (JJ3)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (15)
          • Published Figures (15)
          • Advanced Verification (2)
          p23 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          p23 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 32

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          p23 Antibody (MA3-414) in ICC/IF

          Immunofluorescent analysis of p23 using Anti-p23 Monoclonal Antibody (JJ5) (Product # MA3-414) shows staining in Hela Cells. p23 staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with or an antibody recognizing p23 (Product # MA3-414) at a dilution of 1:500 over night at 4°C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 35503, Goat ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          p23 Antibody in Immunocytochemistry (ICC/IF)
          p23 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Flow Cytometry (Flow)
          p23 Antibody in Immunocytochemistry (ICC/IF)
          p23 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          p23 Antibody in Flow Cytometry (Flow)
          p23 Antibody in Immunocytochemistry (ICC/IF)
          p23 Antibody in Immunocytochemistry (ICC/IF)
          p23 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Flow Cytometry (Flow)
          p23 Antibody in Immunoprecipitation (IP)
          p23 Antibody in RNA Immunoprecipitation (RIP)
          p23 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          p23 Antibody in Immunocytochemistry (ICC/IF)
          p23 Antibody in Immunocytochemistry (ICC/IF)
          p23 Antibody in Immunocytochemistry (ICC/IF)
          p23 Antibody in Immunohistochemistry (IHC)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Western Blot (WB)
          p23 Antibody in Immunoprecipitation (IP)
          p23 Antibody in Proximity Ligation Assay (PLA) (PLA)
          p23 Antibody
          p23 Antibody

          Product Details

          MA3-414

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 20 publications 20 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:200-1:2,000
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          1/100
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 6 publications 6 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Chicken, Guinea pig, Human, Mouse, Rabbit, Rat

          Published species

          Human, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          JJ3

          Immunogen

          Recombinant human p23 protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          ascites

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2175197

          Product Specific Information

          MA3-414 detects p23 from chicken, guinea pig, human, rat, mouse and rabbit samples.

          MA3-414 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects a 19 or 23 kDa protein representing p23 from rabbit reticulocyte lysate. MA3-414 can be used to immunoprecipitate both free and complexed p23.

          The MA3-414 antigen is recombinant human p23.

          Target Information

          Steroid receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Prior to activation, steroid receptors associate with a number of different proteins in both a stable and transient fashion. Steroid receptor complex proteins include heat shock proteins (HSP70 and HSP90), immunosuppressant binding proteins called immunophilins (the FK506 binding proteins, FKBP52 & FKBP54 and the cyclosporin binding protein, CyP-40) and at least three other proteins termed p23, p60 and p48. p23 along with HSP70, HSP90 and p60, combine with progesterone receptor (PR) as members of a transient intermediate complex. Cloned human p23 encodes a protein of 160 amino acids that is highly conserved between species and shows no homology to previously identified proteins. p23 is a highly acidic phosphoprotein with an aspartic acid-rich C-terminal domain and multiple potential phosphorylation sites. In vitro studies have suggested that p23 binds to HSP90 and is necessary for the binding of HSP90 and CyP-40 to PR. While neither its exact function nor mechanism of action have been identified, p23 appears to be an important factor in PR function.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Co chaperone p23; cPGES; cytosolic prostaglandin E synthase; Cytosolic prostaglandin E2 synthase; Hsp90 co chaperone; Hsp90 co-chaperone; p23; p23 cochaperone; Progesterone receptor complex p23; Prostaglandin E synthase 3; prostaglandin E synthase 3 (cytosolic); prostaglandin-E synthase 3; ptges3 {ECO:0000250|UniProtKB:Q15185}; Sid 3177; telomerase binding protein, p23; Telomerase-binding protein p23; unactive progesterone receptor, 23 kD; unnamed protein product

          View more View less

          Gene Aliases: 5730442A20Rik; CPGES; P23; Ptges; PTGES3; RGD1561913; Sid3177; TEBP

          View more View less

          UniProt ID: (Chicken) Q90955, (Human) Q15185, (Rat) P83868, (Mouse) Q9R0Q7

          View more View less

          Entrez Gene ID: (Rabbit) 100351480, (Chicken) 100859133, (Human) 10728, (Rabbit) 100345490, (Rat) 362809, (Guinea pig) 100718334, (Mouse) 56351

          View more View less

          Function(s)
          telomerase activity progesterone receptor binding prostaglandin-E synthase activity unfolded protein binding chaperone binding Hsp90 protein binding protein binding isomerase activity DNA polymerase binding p53 binding enzyme binding chaperone
          Process(es)
          prostaglandin biosynthetic process protein folding telomere maintenance via telomerase chaperone-mediated protein complex assembly telomerase holoenzyme complex assembly telomere maintenance lipid metabolic process fatty acid metabolic process fatty acid biosynthetic process prostaglandin metabolic process signal transduction positive regulation of telomere maintenance via telomerase prostanoid biosynthetic process protein stabilization glycogen biosynthetic process RNA-dependent DNA biosynthetic process cell proliferation positive regulation of gene expression sensory perception of pain glucocorticoid receptor signaling pathway skin development lung saccule development negative regulation of cell death chaperone cofactor-dependent protein refolding DNA biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0