Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • VCP Antibodies
          Validated with Transparency. Independently tested through the YCharOS Open Science Initiative.

          Invitrogen

          VCP Monoclonal Antibody (5)

          Advanced Verification
          55 Published Figures
          59 References
          View all (42) VCP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite VCP Monoclonal Antibody (5)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (10)
          • Published Figures (55)
          • Advanced Verification (7)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          VCP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 72

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VCP Antibody (MA3-004) in ICC/IF

          Immunofluorescence analysis of VCP was performed using HeLa cells. The cells were fixed and permeabilized with ice cold 100% Acetone for 5 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with VCP Monoclonal Antibody (5) (Product # MA3-004) at 5 microgram/mL concentration in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Goat anti-Mouse IgG (H+L) Superclonal™ Recombinant Secondary Antibody, Alexa Fluor® 488 (Product # A28175) for 45 minutes at room temperature (Panel a: green)... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VCP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Immunoprecipitation (IP)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunocytochemistry (ICC/IF)
          VCP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VCP Antibody in Immunohistochemistry (IHC)
          VCP Antibody in Immunohistochemistry (IHC)
          VCP Antibody in Immunohistochemistry (IHC)
          VCP Antibody in Immunohistochemistry (IHC)
          VCP Antibody in Immunohistochemistry (IHC)
          VCP Antibody in Immunohistochemistry (IHC)
          VCP Antibody in Immunohistochemistry (IHC)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot, Immunoprecipitation (WB, IP)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot, Immunoprecipitation (WB, IP)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot, Immunoprecipitation (WB, IP)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Western Blot (WB)
          VCP Antibody in Immunoprecipitation (IP)
          VCP Antibody in Immunoprecipitation (IP)
          VCP Antibody in Immunoprecipitation (IP)
          VCP Antibody in Immunoprecipitation (IP)
          VCP Antibody
          VCP Antibody
          VCP Antibody
          VCP Antibody
          VCP Antibody
          VCP Antibody
          VCP Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          VCP Monoclonal Antibody (5)

          Product Details

          MA3-004

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 31 publications 31 publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:500
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:500
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 10 publications 10 publications

          Flow Cytometry (Flow)

          1/400
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Yeast

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          5

          Immunogen

          Synthetic peptide corresponding to residues C G(792) G S V Y T E D N D D D L Y G(806) of mouse VCP.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.0 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.4, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2214638

          Product Specific Information

          MA3-004 detects VCP protein in human, mouse, and rat samples.

          MA3-004 has successfully been used in Western blot immunoprecipitation, and immunohistochemical procedures. By Western blot, this antibody detects an ~97 kDa protein representing VCP from total lysate of cultured human B cells.

          The MA3-004 immunogen is a synthetic peptide corresponding to residues C G(792) G S V Y T E D N D D D L Y G(806) of mouse VCP. This peptide (Cat. # PEP-239) is available for use in neutralization and control experiments.

          Target Information

          The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 15S Mg(2+)-ATPase p97 subunit; AAA ATPase p97; homolog of yeast cdc48; Inv protein; or HEL-S-70; p97 atpase; TER ATPase; unnamed protein product

          View more View less

          Gene Aliases: 3110001E05; CDC48; FTDALS6; p97; p97/VCP; TERA

          View more View less

          UniProt ID: (Human) P55072, (Mouse) Q01853, (Rat) P46462

          View more View less

          Entrez Gene ID: (Human) 7415, (Mouse) 269523, (Rat) 116643

          View more View less

          Function(s)
          RNA binding protein binding ATP binding hydrolase activity ATPase activity protein phosphatase binding protein domain specific binding polyubiquitin binding ubiquitin protein ligase binding deubiquitinase activator activity K48-linked polyubiquitin binding MHC class I protein binding identical protein binding ADP binding ubiquitin-like protein ligase binding macromolecular complex binding ubiquitin-dependent protein binding BAT3 complex binding ubiquitin-specific protease binding
          Process(es)
          mitophagy DNA repair double-strand break repair ubiquitin-dependent protein catabolic process ER to Golgi vesicle-mediated transport autophagy cellular response to DNA damage stimulus proteasomal protein catabolic process positive regulation of mitochondrial membrane potential macroautophagy protein ubiquitination peptidyl-lysine trimethylation viral genome replication NAD metabolic process translesion synthesis endoplasmic reticulum unfolded protein response retrograde protein transport, ER to cytosol positive regulation of protein complex assembly positive regulation of proteasomal ubiquitin-dependent protein catabolic process endosome to lysosome transport via multivesicular body sorting pathway cellular response to heat negative regulation of hippo signaling stress granule disassembly interstrand cross-link repair ERAD pathway regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process protein unfolding establishment of protein localization positive regulation of protein catabolic process negative regulation of smoothened signaling pathway ATP metabolic process regulation of synapse organization mitotic spindle disassembly endoplasmic reticulum stress-induced pre-emptive quality control cellular response to misfolded protein rescue of stalled ribosome flavin adenine dinucleotide catabolic process positive regulation of canonical Wnt signaling pathway autophagosome maturation protein-DNA covalent cross-linking repair negative regulation of protein localization to chromatin cytoplasm protein quality control positive regulation of NIK/NF-kappaB signaling positive regulation of protein K63-linked deubiquitination regulation of aerobic respiration cellular response to arsenite ion positive regulation of oxidative phosphorylation regulation of protein localization to chromatin ribosome-associated ubiquitin-dependent protein catabolic process positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process positive regulation of ATP biosynthetic process I-kappaB kinase/NF-kappaB signaling aggresome assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch