Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HSC70 Antibodies

          Invitrogen

          HSC70 Monoclonal Antibody (13D3)

          9 Published Figures
          29 References
          View all (44) HSC70 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite HSC70 Monoclonal Antibody (13D3)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (11)
          • Published Figures (9)
          HSC70 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HSC70 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HSC70 Antibody (MA3-014) in ICC/IF

          Immunofluorescent analysis of Heat Shock Protein 70 (HSC70) (green) in HeLa and NIH3T3 cells. Formalin fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 10 minutes at room temperature and blocked with 1% Blocker BSA (Product # 37525) for 15 minutes at room temperature. Cells were probed with a HSC70 monoclonal antibody (Product # MA3-014) at a dilution of 1:50 for at least 1 hour at room temperature, washed with PBS, and incubated with DyLight 488 goat anti-mouse IgG secondary antibody (Product # 35502) at a dilution of 1:40... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HSC70 Antibody in Immunocytochemistry (ICC/IF)
          HSC70 Antibody in Immunocytochemistry (ICC/IF)
          HSC70 Antibody in Immunocytochemistry (ICC/IF)
          HSC70 Antibody in Immunocytochemistry (ICC/IF)
          HSC70 Antibody in Immunocytochemistry (ICC/IF)
          HSC70 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSC70 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSC70 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSC70 Antibody in Western Blot (WB)
          HSC70 Antibody in Western Blot (WB)
          HSC70 Antibody in Immunoprecipitation (IP)
          HSC70 Antibody in Immunocytochemistry (ICC/IF)
          HSC70 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          HSC70 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          HSC70 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          HSC70 Antibody in Immunohistochemistry (IHC)
          HSC70 Antibody in Western Blot (WB)
          HSC70 Antibody in Western Blot (WB)
          HSC70 Antibody in Western Blot (WB)
          HSC70 Antibody in Neutralization (Neu)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          HSC70 Monoclonal Antibody (13D3)

          Product Details

          MA3-014

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 15 publications 15 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:250
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          -
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          2 µL
          -

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Competition Assay (Competition)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Cat, Human, Mouse, Non-human primate, Rat

          Published species

          Bovine, Cat, Fish, Human, Monkey, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgM

          Class

          Monoclonal

          Type

          Antibody

          Clone

          13D3

          Immunogen

          Mouse spermatogenic cell protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography - MBP

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325462

          Product Specific Information

          MA3-014 detects heat shock cognate protein 70 kDa (HSC70) from human, feline, primate, rat, and mouse tissues. This antibody displays slight cross-reactivity to HSP70.

          MA3-014 has been successfully used in Western blot, immunohistochemistry, immunofluorescence, and immunoprecipitation procedures By Western blot, this antibody detects a 70 kDa protein representing HSC70. By 2D gel electrophoresis, MA3-014 was found to bind strongly to HSC70 and faintly to HSP70 both before and after heat shock. Immunohistochemical staining of HSC70 in mouse spermatids/spermatozoa with MA3-014 results in staining restricted to the post acrosomal region in condensing spermatids and to the midpiece in spermatozoa.

          The MA3-014 antigen is mouse spermatogenic cell protein.

          Target Information

          The HSP70 family is a set of highly conserved proteins that are induced by a variety of biological stresses, including heat stress, in every organism in which the proteins have been examined. The human HSP70 family members include: HSP70, a protein which is strongly inducible in all organisms but which is also constitutively expressed in primate cells; HSP72, a 72 kDa protein that is induced exclusively under stress conditions; HSC70, or cognate protein, is a 72 kDa, constitutively expressed, protein which is involved in the uncoating of clathrin coated vesicles; GRP78, or BiP, is a glucose regulated 78 kDa protein localized in the endoplasmic reticulum; and p75, or HSP75, a 75 kDa protein that is found within the mitochondria. HSC70 (also known as HSC71, HSC73, HSP73, p72, prp73) is expressed constitutively and is slightly heat-inducible. HSC70 binds to the exposed loop of clathrin light chains to promote uncoating and can also bind the cytoskeleton which may facilitate cytoskeletal rearrangements. HSC70 has been shown to stimulate lysosomal degradation of intracellular proteins and to retard both aggregation and folding of mitochondrial precursor proteins in vitro.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 70 kDa heat-shock-like protein; AAG2; constitutive heat shock protein 70; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 72p; FAM10A1; heat shock 70 kDa protein 8; heat shock 70kd protein 10; heat shock 70kD protein 8; heat shock 70kDa protein 8; heat shock cognate hsc73; heat shock cognate protein 54; Heat shock cognate protein 70; heat shock protein 70 cognate; heat shock protein 8; heat shock protein A8; heat shock protein cognate 70; heat shock protein family A member 8; HIP; LAP; lipopolysaccharide-associated protein 1; LPS-associated protein 1; MGC131511; MGC29929; N-myristoyltransferase inhibitor protein 71; Protein FAM10A1; SNC6; unnamed protein product

          View more View less

          Gene Aliases: 2410008N15Rik; HEL-33; HEL-S-72p; HSC54; HSC70; HSC71; Hsc73; HSP71; HSP73; HSPA10; LAP-1; LAP1; NIP71

          View more View less

          UniProt ID: (Human) P11142, (Mouse) P63017, (Rat) P63018

          View more View less

          Entrez Gene ID: (Human) 3312, (Mouse) 15481, (Rat) 24468

          View more View less

          Function(s)
          G-protein coupled receptor binding RNA binding protein binding ATP binding ATPase activity enzyme binding MHC class II protein complex binding protein binding, bridging heat shock protein binding ubiquitin protein ligase binding protein binding involved in protein folding cadherin binding receptor agonist activity chaperone binding C3HC4-type RING finger domain binding ATP-dependent protein disaggregase activity ATP-dependent protein folding chaperone phosphatidylserine binding receptor binding lipid binding A1 adenosine receptor binding AP-3 adaptor complex binding peptide binding lncRNA binding prostaglandin binding clathrin-uncoating ATPase activity
          Process(es)
          G1/S transition of mitotic cell cycle mRNA splicing, via spliceosome kidney development positive regulation of T cell mediated cytotoxicity protein folding protein import into nucleus autophagy signal transduction skeletal muscle tissue development axo-dendritic transport response to xenobiotic stimulus response to nickel cation positive regulation of gene expression negative regulation of cardiac muscle cell apoptotic process response to activity cerebellum development protein catabolic process positive regulation of cell migration forebrain development regulation of protein stability response to estradiol response to progesterone protein-containing complex disassembly cellular response to heat protein refolding response to starvation positive regulation of catalytic activity intracellular protein transmembrane import modulation by host of viral process positive regulation by host of viral genome replication estrous cycle response to ethanol positive regulation of proteolysis negative regulation of transcription, DNA-templated ATP metabolic process protein autophosphorylation positive regulation of mRNA splicing, via spliceosome positive regulation of phagocytosis regulation of cell cycle regulation of protein complex stability chaperone-mediated autophagy late endosomal microautophagy protein targeting to lysosome involved in chaperone-mediated autophagy cellular response to hydrogen peroxide cellular response to cadmium ion clathrin coat disassembly positive regulation of lysosomal membrane permeability maintenance of postsynaptic specialization structure regulation of postsynapse organization positive regulation of ferroptosis negative regulation of NLRP3 inflammasome complex assembly negative regulation of fibril organization positive regulation of protein refolding chaperone-mediated autophagy translocation complex disassembly slow axonal transport response to odorant response to stress response to unfolded protein cellular response to starvation negative regulation of signal transduction regulation of cytoplasmic pattern recognition receptor signaling pathway regulation of programmed cell death regulation of protein complex assembly negative regulation of cellular component organization membrane organization cellular response to steroid hormone stimulus regulation of fibril organization regulation of protein import
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch