Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NFATC1 Antibodies

          Invitrogen

          NFATC1 Monoclonal Antibody (7A6)

          Advanced Verification
          52 Published Figures
          93 References
          View all (23) NFATC1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NFATC1 Monoclonal Antibody (7A6)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (52)
          • Advanced Verification (6)
          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 65

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NFATC1 Antibody (MA3-024) in ICC/IF

          Immunofluorescent analysis of NFATc1 using NFATc1 Monoclonal Antibody (7A6) (Product # MA3-024) shows staining in MCF-7 Cells. NFATc1 (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with an antibody recognizing NFATc1 (Product # MA3-024) at a dilution of 1:20 over night at 4 °C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 35552 for GAR, P... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          NFATC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NFATC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NFATC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot, Immunohistochemistry, Immunocytochemistry (WB, IHC, ICC/IF)
          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          NFATC1 Antibody in Immunocytochemistry (ICC/IF)
          NFATC1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          NFATC1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          NFATC1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          NFATC1 Antibody in Western Blot, Immunohistochemistry (Paraffin) (WB, IHC (P))
          NFATC1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          NFATC1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Western Blot (WB)
          NFATC1 Antibody in Flow Cytometry (Flow)
          NFATC1 Antibody in Immunoprecipitation (IP)
          NFATC1 Antibody in ChIP Assay (ChIP)
          NFATC1 Antibody in ChIP Assay (ChIP)
          NFATC1 Antibody in ChIP Assay (ChIP)
          NFATC1 Antibody in ChIP Assay (ChIP)
          NFATC1 Antibody in ChIP Assay (ChIP)
          NFATC1 Antibody in ChIP Assay (ChIP)
          NFATC1 Antibody in Gel Shift (EMSA)
          NFATC1 Antibody in Gel Shift (EMSA)
          NFATC1 Antibody
          NFATC1 Antibody
          NFATC1 Antibody
          NFATC1 Antibody
          NFATC1 Antibody
          NFATC1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NFATC1 Monoclonal Antibody (7A6)

          Product Details

          MA3-024

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:2,000
          View 46 publications 46 publications

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:250
          View 14 publications 14 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          View 3 publications 3 publications

          ChIP assay (ChIP)

          Assay-dependent
          View 9 publications 9 publications

          Gel Shift (EMSA)

          Assay-dependent
          View 12 publications 12 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Published species

          Hamster, Human, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          7A6

          Immunogen

          Bacterially expressed glutathione S-transferase (GST) fusion protein containing NF-ATc1 residues 1-654.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          ascites, PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2236037

          Product Specific Information

          MA3-024 detects NFATc1 (NFAT2) from transfected human, rat, mouse, and monkey cells. This antibody does not cross react with NFATc2 (NFAT1).

          MA3-024 has been successfully used in Western blot, immunoprecipitation, ChIP, immunofluorescence and gel shift procedures. By Western blot, this antibody detects an ~105 kDa band representing transfected NFAT2 from COS cell lysate.

          The MA3-024 immunogen is bacterially expressed glutathione S-transferase (GST) fusion protein containing NF-ATc1 residues 1-654.

          Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell-based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization, including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody, may have to be performed for your high content assay.

          Target Information

          The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. The product of this gene is an inducible nuclear component. It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for this gene. Different isoforms of this protein may regulate inducible expression of different cytokine genes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: MGC138448; NF-ATc1; NFAT tra; NFAT trans; NFAT transcription complex cytosolic component; nuclear factor of activated T cells; nuclear factor of activated T-cells 'c'; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1; transcription factor; transcription factor NF-ATc; unnamed protein product

          View more View less

          Gene Aliases: 2210017P03Rik; NF-ATC; NF-ATc1.2; NFAT2; NFATc; Nfatcb

          View more View less

          UniProt ID: (Human) O95644

          View more View less

          Entrez Gene ID: (Human) 4772, (Rat) 100361818, (Mouse) 18018

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding FK506 binding protein phosphatase 2B binding mitogen-activated protein kinase p38 binding RNA polymerase II sequence-specific DNA binding transcription factor binding sequence-specific double-stranded DNA binding transcription coactivator binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding chromatin binding sequence-specific DNA binding DNA-binding transcription factor binding RNA polymerase II regulatory region sequence-specific DNA binding
          Process(es)
          aortic valve morphogenesis pulmonary valve morphogenesis regulation of transcription, DNA-templated negative regulation of Wnt signaling pathway calcineurin-NFAT signaling cascade intracellular signal transduction positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of inflammatory response negative regulation of vascular smooth muscle cell differentiation G1/S transition of mitotic cell cycle negative regulation of transcription from RNA polymerase II promoter epithelial to mesenchymal transition lymphangiogenesis B-1a B cell differentiation heart valve development aortic valve development pulmonary valve development heart valve morphogenesis endocardial cushion development regulation of transcription from RNA polymerase II promoter calcium ion transport heart development gene expression positive regulation of gene expression response to muscle activity transition between fast and slow fiber osteoclast differentiation wound healing regulation of hair cycle skeletal muscle adaptation negative regulation of osteoblast differentiation ventricular septum morphogenesis patterning of lymph vessels heart trabecula morphogenesis cellular response to calcium ion cellular response to tumor necrosis factor cellular response to transforming growth factor beta stimulus mononuclear cell differentiation semi-lunar valve development positive regulation of DNA biosynthetic process negative regulation of stem cell proliferation cellular response to metal ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch