Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Calnexin Antibodies

          Invitrogen

          Calnexin Monoclonal Antibody (AF18)

          25 Published Figures
          82 References
          View all (54) Calnexin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Calnexin Monoclonal Antibody (AF18)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (10)
          • Published Figures (25)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 35

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calnexin Antibody (MA3-027) in ICC/IF

          Immunofluorescent analysis of calnexin (red) in U87 cells. Cell fixed with 4% paraformaldehyde were permeabilized with 0.05% Triton X-100 in PBS for 60 seconds at room temperature and blocked with 5% normal donkey serum for 30 minutes at room temperature. Cells were probed with a calnexin monoclonal antibody (Product # MA3-027) at a dilution of 1:100 for 2 hours at room temperature, washed with PBS, and incubated with a fluorescently-conjugated goat anti-mouse IgG secondary antibody at a dilution of 1:100 for 45 minutes at room temperature... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calnexin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Immunoprecipitation (IP)
          Calnexin Antibody in RNA Immunoprecipitation (RIP)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Immunocytochemistry (ICC/IF)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Calnexin Antibody in Western Blot (WB)
          Calnexin Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Calnexin Monoclonal Antibody (AF18)

          Product Details

          MA3-027

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:100-1:1,000
          View 34 publications 34 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20
          -

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 37 publications 37 publications

          Immunoprecipitation (IP)

          5 µg
          View 3 publications 3 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          -
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Dog, Hamster, Human, Mouse, Non-human primate, Rat, Yeast

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AF18

          Immunogen

          Focus human hepatoma cell lysate.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 555 
          • Alexa Fluor 647 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2069043

          Product Specific Information

          MA3-027 detects calnexin from human and mouse tissues.

          MA3-027 has been successfully used in Western blot, immunocytochemistry, immunofluorescence and immunoprecipitation protocols. By Western blot, this antibody detects calnexin in human liver, skin, adrenal gland, heart, colon, testes and ovary extracts. This antibody is not recommended for human kidney or mouse liver lysates in Western blot applications. The predicted molecular weight of this antibody is ~67kDa; however, it may run on SDS-PAGE as an ~90kDA protein. This is due mainly to negative charges at the C-terminus of the protein, which effects the SDS binding to the molecules and consequently its SDS-PAGE mobility. Immunocytochemical staining of calnexin in Huh-7 cells with MA3-027 yields a pattern consistent with specific endoplasmic reticulum staining.

          The MA3-027 antigen is Focus human hepatoma cell lysate.

          Target Information

          Calnexin, also referred to as IP90, p88 and p90, is an approximately 90 kDa integral membrane protein of the endoplasmic reticulum (ER). Many resident ER proteins act as molecular chaperones and participate in the proper folding of polypeptides and their assembly into multisubunit proteins. Studies indicate that calnexin associates with the major histocompatibility complex (MHC) class I heavy chains, partial complexes of the T cell receptor and B cell membrane immunoglobulin, but not with completed receptor complexes. It has been shown that calnexin is a chaperone that retains incompletely or improperly folded proteins in the ER. The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble ER resident proteins such as GRP78 and GRP94 and protein disulfide isomerase. Integral membrane ER resident proteins, like calnexin, often lack this KDEL sequence but contain positively charged cytosolic residues that ensure ER retention. Calnexin contains a large ER luminal domain (461 amino acids), a transmembrane segment (22 amino acids), and a cytoplasmic tail (89 amino acids).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ca(2+)-binding phosphoprotein p90; epididymis secretory sperm binding protein; FLJ26570; major histocompatibility complex class I antigen-binding protein p88; p88; unnamed protein product

          View more View less

          Gene Aliases: 1110069N15Rik; CNX; D11Ertd153e; IP90; P90

          View more View less

          UniProt ID: (Human) P27824, (Mouse) P35564

          View more View less

          Entrez Gene ID: (Human) 821, (Mouse) 12330

          View more View less

          Function(s)
          RNA binding calcium ion binding protein binding apolipoprotein binding ionotropic glutamate receptor binding
          Process(es)
          protein folding protein secretion viral protein processing protein folding in endoplasmic reticulum ERAD pathway synaptic vesicle endocytosis clathrin-mediated endocytosis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch