Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • mtHSP70 Antibodies

          Invitrogen

          mtHSP70 Monoclonal Antibody (JG1)

          Advanced Verification
          41 Published Figures
          148 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite mtHSP70 Monoclonal Antibody (JG1)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          • Published Figures (41)
          • Advanced Verification (1)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 49

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          mtHSP70 Antibody (MA3-028) in ICC/IF

          Immunofluorescent analysis of Heat Shock Protein 70 using anti-Heat Shock Protein 70 monoclonal antibody (Product # MA3-028) shows staining in NS-1 Cells. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          mtHSP70 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          mtHSP70 Antibody in Immunocytochemistry (ICC/IF)
          mtHSP70 Antibody in Immunohistochemistry (IHC)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody in Western Blot (WB)
          mtHSP70 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          mtHSP70 Monoclonal Antibody (JG1)

          Product Details

          MA3-028

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 100 publications 100 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1:50
          View 34 publications 34 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 2 publications 2 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 9 publications 9 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Non-human primate

          Published species

          Dog, Guinea pig, Hamster, Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgG3

          Class

          Monoclonal

          Type

          Antibody

          Clone

          JG1

          Immunogen

          Synthetic peptide corresponding to residues G(615) S G S S G T G E Q K E D Q K E E K Q(633) of mouse mtHSP70.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.07 mg/mL

          Amount

          107 µg

          Purification

          Protein A

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325474

          Product Specific Information

          MA3-028 detects mitochondrial heat shock protein 70 kDa (mtHSP70) from human, non-human primate, canine and mouse tissues.

          MA3-028 has been successfully used in Western blot, immunocytochemical, immunofluorescence, immunohistochemical (paraffin), and immunoprecipitation procedures. By Western blot, this antibody detects a single ~75 kDa band representing mtHSP70 from U2OS cell homogenate. Immunocytochemical staining of mtHSP70 in DAP.3 cells with MA3-028 results in a worm-like staining pattern, consistent with mitochondrial localization.

          Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay.

          Target Information

          The HSP70 family is a set of highly conserved proteins that are induced by a variety of biological stresses, including heat stress, in every organism in which the proteins have been examined. The human HSP70 family members include: HSP70, a 70 kDa protein which is strongly inducible in all organisms but which is also constitutively expressed in primate cells; HSP72, a 72 kDa protein which is induced exclusively under stress conditions; HSC70, or cognate protein, is a 72 kDa constitutively expressed protein which is involved in uncoating clathrin-coated vesicles; GRP78, or BiP, is a glucose-regulated 78 kDa protein localized to the endoplasmic reticulum; and mitochondrial HSP70 (mtHSP70, GRP75 or mortalin) a 75 kDa protein that is found within the mitochondria. mtHSP70 is a mitochondrial resident protein that is involved in protein translocation into the mitochondria. Preproteins cross the mitochondrial membranes in an extended conformation. This requires unfolding of preproteins before entering translocation pores in the mitochondrial outer membrane. Preprotein unfolding is thought to be mediated by mtHSP70 and other inner membrane proteins of the mitochondria.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 68 kDa heat shock protein; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; epididymis secretory sperm binding protein; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; heat shock 70 kDa protein 3; heat shock 70kD protein 1A; heat shock 70kDa protein 1A; heat shock protein 1A; heat shock protein family A member 1A; heat shock protein, 70 kDa 3; heat shock-induced protein; HSP70-1/HSP70-2; HSP70.1/HSP70.2; inducible heat shock protein 70; unnamed protein product

          View more View less

          Gene Aliases: HEL-S-103; hsp68; HSP70; HSP70-1; HSP70-1A; HSP70-2; Hsp70-3; HSP70.1; HSP70.2; Hsp70.3; hsp70A1; HSP70I; HSP72; HSPA1

          View more View less

          UniProt ID: (Human) P0DMV8, (Mouse) Q61696

          View more View less

          Entrez Gene ID: (Human) 3303, (Mouse) 193740

          View more View less

          Function(s)
          G-protein coupled receptor binding transcription corepressor activity RNA binding receptor binding protein binding ATP binding ATPase activity enzyme binding heat shock protein binding ubiquitin protein ligase binding histone deacetylase binding protein binding involved in protein folding cadherin binding receptor agonist activity misfolded protein binding C3HC4-type RING finger domain binding disordered domain specific binding transcription regulator inhibitor activity ATP-dependent protein disaggregase activity protease binding death receptor agonist activity NF-kappaB binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter mRNA catabolic process response to unfolded protein lysosomal transport negative regulation of cell proliferation positive regulation of gene expression negative regulation of cell growth negative regulation of transforming growth factor beta receptor signaling pathway endoplasmic reticulum unfolded protein response regulation of protein ubiquitination negative regulation of protein ubiquitination positive regulation of proteasomal ubiquitin-dependent protein catabolic process positive regulation of interleukin-8 production positive regulation of RNA splicing cellular response to oxidative stress cellular response to heat cellular response to unfolded protein protein refolding negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of erythrocyte differentiation ATP metabolic process protein stabilization chaperone-mediated protein complex assembly cellular heat acclimation positive regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway cellular response to steroid hormone stimulus positive regulation of microtubule nucleation negative regulation of inclusion body assembly negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway regulation of mitotic spindle assembly negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of tumor necrosis factor-mediated signaling pathway negative regulation of extrinsic apoptotic signaling pathway in absence of ligand telomere maintenance DNA repair protein folding response to heat
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-qh5rm:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0