Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Huntingtin Antibodies
          Validated with Transparency. Independently tested through the YCharOS Open Science Initiative.

          Invitrogen

          Huntingtin Monoclonal Antibody (1HU-4C8)

          Advanced Verification
          3 Published Figures
          2 References
          View all (37) Huntingtin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Huntingtin Monoclonal Antibody (1HU-4C8)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (3)
          • Advanced Verification (3)
          Huntingtin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Huntingtin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Huntingtin Antibody (MA3-040) in ICC/IF

          Immunofluorescence analysis of Huntingtin was performed using 70% confluent log phase SH-SY5Y cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Huntingtin Monoclonal Antibody (1HU-4C8) (Product # MA3-040) at 1:200 dilution in 0.1% BSA, incubated at ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Huntingtin Antibody in Immunocytochemistry (ICC/IF)
          Huntingtin Antibody in Western Blot (WB)
          Huntingtin Antibody in Western Blot (WB)
          Huntingtin Antibody in Western Blot (WB)
          Huntingtin Antibody in Western Blot (WB)
          Huntingtin Antibody in Immunoprecipitation (IP)
          Huntingtin Antibody in Western Blot (WB)
          Huntingtin Antibody in Western Blot (WB)
          Huntingtin Antibody in Western Blot (WB)
          Huntingtin Antibody
          Huntingtin Antibody
          Huntingtin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Huntingtin Monoclonal Antibody (1HU-4C8)

          Product Details

          MA3-040

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:2,000
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:200
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1HU-4C8

          Immunogen

          Huntingtin fragment from aa 181 to 810
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          ascites

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2608783

          Product Specific Information

          MA3-040 has been used successfully in Western blotting to detect huntingtin in the brains lysates from mouse and rat.

          Target Information

          Huntingtin is a disease gene linked to Huntington's disease (HD), a neurodegenerative disorder characterized by loss of striatal neurons. This is thought to be caused by an expanded, unstable trinucleotide repeat in the huntingtin gene, which translates as a polyglutamine repeat in the protein product. HD is a mid-life onset autosomal dominant neurodegenerative disease that is characterized by psychiatric disorders, dementia, and involuntary movements (chorea), leading to death in 10-20 years.The huntingtin locus is large, spanning 180 kb and consisting of 67 exons. The huntingtin gene is widely expressed and is required for normal development. It is expressed as 2 alternatively polyadenylated forms displaying different relative abundance in various fetal and adult tissues. The larger transcript is approximately 13.7 kb and is expressed predominantly in adult and fetal brain whereas the smaller transcript of approximately 10.3 kb is more widely expressed. The genetic defect leading to Huntington's disease may not necessarily eliminate transcription, but may confer a new property on the mRNA or alter the function of the protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HD protein homolog; huntingtin (Huntington disease); Huntington disease gene homolog; huntington disease protein; huntington disease protein homolog; Huntington gene; unnamed protein product

          View more View less

          Gene Aliases: C430023I11Rik; HD; Hdh; IT15; LOMARS

          View more View less

          UniProt ID: (Human) P42858

          View more View less

          Entrez Gene ID: (Human) 3064, (Rat) 29424, (Mouse) 15194

          View more View less

          Function(s)
          p53 binding phosphoprotein phosphatase activity protein binding profilin binding kinase binding heat shock protein binding dynactin binding identical protein binding ion channel binding dynein intermediate chain binding beta-tubulin binding receptor binding diazepam binding
          Process(es)
          urea cycle establishment of mitotic spindle orientation mitochondrial transport ER to Golgi vesicle-mediated transport retrograde vesicle-mediated transport, Golgi to ER mitochondrion organization microtubule-based process microtubule depolymerization endoplasmic reticulum organization Golgi organization dopamine receptor signaling pathway spermatogenesis gastrulation central nervous system development brain development learning or memory learning grooming behavior locomotory behavior axo-dendritic transport associative learning determination of adult lifespan visual learning anatomical structure morphogenesis anterior/posterior pattern specification regulation of signal transduction gene expression regulation of gene expression endosomal transport lactate biosynthetic process from pyruvate quinolinate biosynthetic process striatum development olfactory lobe development neural plate formation neurogenesis peptide hormone secretion insulin secretion protein destabilization protein localization to nucleus social behavior vocal learning hormone metabolic process positive regulation of apoptotic process negative regulation of apoptotic process regulation of neuron apoptotic process positive regulation of cilium assembly vesicle transport along microtubule regulation of synaptic plasticity paraxial mesoderm formation synaptic vesicle transport neuron development positive regulation of calcium-mediated signaling mRNA transport response to calcium ion multicellular organismal iron ion homeostasis protein localization to centrosome cellular senescence postsynapse to nucleus signaling pathway regulation of protein localization to nucleus positive regulation of mitophagy positive regulation of nonmotile primary cilium assembly positive regulation of lipophagy regulation of intracellular mRNA localization regulation of CAMKK-AMPK signaling cascade positive regulation of CAMKK-AMPK signaling cascade positive regulation of aggrephagy positive regulation of motile cilium assembly negative regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-sf8lr:80/100.66.74.151:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline