Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Calsequestrin Antibodies

          Invitrogen

          Calsequestrin Monoclonal Antibody (VIIID12)

          Advanced Verification
          37 Published Figures
          80 References
          View all (12) Calsequestrin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Calsequestrin Monoclonal Antibody (VIIID12)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (37)
          • Advanced Verification (2)
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 46

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calsequestrin Antibody (MA3-913) in ICC/IF

          Immunofluorescent analysis of Calsequestrin using Anti-Calsequestrin Monoclonal Antibody (VIIID12) (Product # MA3-913) shows staining in MCF-7 Cells. Calsequestrin staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with or an antibody recognizin... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Calsequestrin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calsequestrin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Calsequestrin Antibody in Immunohistochemistry (IHC)
          Calsequestrin Antibody in Immunohistochemistry (IHC)
          Calsequestrin Antibody in Immunohistochemistry (IHC)
          Calsequestrin Antibody in Immunohistochemistry (IHC)
          Calsequestrin Antibody in Immunohistochemistry (IHC)
          Calsequestrin Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Calsequestrin Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody
          Calsequestrin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Calsequestrin Monoclonal Antibody (VIIID12)

          Product Details

          MA3-913

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 66 publications 66 publications

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 4 publications 4 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Chicken, Human, Mouse, Pig, Rabbit, Rat

          Published species

          Chicken, Dog, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Rodent

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          VIIID12

          Immunogen

          Purified rabbit skeletal muscle sarcoplasmic reticulum.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          PBS, ascites

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325496

          Product Specific Information

          MA3-913 detects calsequestrin from human, mouse, rat, canine, porcine, chicken, and rabbit skeletal muscle tissues. This antibody recognizes calsequestrin in both type I (slow) and type II (fast) skeletal muscle tissues.

          MA3-913 has been successfully used in Western blot procedures. By Western blot, this antibody detects a 63 kDa protein representing calsequestrin from canine skeletal muscle extracts. Higher molecular weight proteins are seen on the Western blot and are believed to be calsequestrin-like proteins found in the sarcoplasmic reticulum. The staining pattern yields double rows of fluorescent dots corresponding to triad pairs on either side of the Z-line.

          The MA3-913 antigen is purified rabbit skeletal muscle sarcoplasmic reticulum.

          Target Information

          The sarcoplasmic reticulum (SR) is, in part, responsible for maintaining the level of intracellular calcium in cardiac and skeletal muscle by storing and releasing calcium. Several intralumenal SR calcium binding proteins have been identified, the most prominent of these is calsequestrin. Calsequstrin is a calcium binding protein known to sequester calcium accumulated in the sarcoplasmic reticulum of muscle cells during relaxation and is found discretely localized to the junctional and corbular (terminal cisternae) SR. Calsequestrin functions to localize calcium near the junctional face of the terminal cisternae from which calcium can be released into the cytosol via the ryanodine receptor. This protein is highly acidic and has a large capacity and moderate to low affinity for calcium.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Aspartactin; calmitin; calmitine; calsequestrin; calsequestrin 1 (fast-twitch, skeletal muscle); Calsequestrin-1; calsquestrin 1; Laminin-binding protein; mitochondrial calcium-binding protein; skeletal muscle calsequestrin 1; unnamed protein product

          View more View less

          Gene Aliases: CASQ; CSQ; CSQ-1; CSQ1; PDIB1; sCSQ; VMCQA

          View more View less

          UniProt ID: (Rabbit) P07221, (Human) P31415, (Rat) P19633, (Mouse) O09165

          View more View less

          Entrez Gene ID: (Rabbit) 100009095, (Pig) 100156448, (Dog) 608401, (Human) 844, (Rat) 686019, (Mouse) 12372

          View more View less

          Function(s)
          calcium ion binding identical protein binding protein binding calcium-binding protein
          Process(es)
          positive regulation of release of sequestered calcium ion into cytosol positive regulation of store-operated calcium channel activity regulation of store-operated calcium entry skeletal muscle tissue development regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion response to muscle inactivity response to denervation involved in regulation of muscle adaptation sarcomere organization protein polymerization regulation of release of sequestered calcium ion into cytosol endoplasmic reticulum organization response to heat regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum regulation of muscle system process regulation of muscle contraction regulation of striated muscle contraction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch