Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SERCA2 ATPase Antibodies

          Invitrogen

          SERCA2 ATPase Monoclonal Antibody (2A7-A1)

          Advanced Verification
          77 Published Figures
          252 References
          View all (24) SERCA2 ATPase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SERCA2 ATPase Monoclonal Antibody (2A7-A1)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (9)
          • Published Figures (77)
          • Advanced Verification (2)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 88

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SERCA2 ATPase Antibody (MA3-919) in ICC/IF

          Immunofluorescence analysis of SERCA2 ATPase was performed using 70 confluent log phase A549 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with SERCA2 ATPase Monoclonal Antibody (2A7-A1) (Product # MA3-919) at 1:100 dilution in 0.1% BSA, incubated at... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SERCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SERCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          SERCA2 ATPase Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          SERCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA2 ATPase Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Western Blot (WB)
          SERCA2 ATPase Antibody in Immunoprecipitation (IP)
          SERCA2 ATPase Antibody
          SERCA2 ATPase Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SERCA2 ATPase Monoclonal Antibody (2A7-A1)

          Product Details

          MA3-919

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 220 publications 220 publications

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1 µg/mL
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1/100
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100-1:250
          View 13 publications 13 publications

          Flow Cytometry (Flow)

          1:20
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 6 publications 6 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat

          Published species

          Amphibian, Dog, Fruit fly, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Rodent, Sheep, Xenopus

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2A7-A1

          Immunogen

          Purified canine cardiac sarcoplasmic reticulum vesicles.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          ascites

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325502

          Product Specific Information

          MA3-919 detects sarcoplasmic or endoplasmic reticulum calcium (SERCA) 2 ATPase from canine, porcine, human, mouse, rabbit and rat tissues. MA3-919 recognizes the SERCA2a and SERCA2b isoforms identically. However, the antibody does not recognize the skeletal muscle (SERCA1) or housekeeping isoforms (SERCA3).

          MA3-919 has been successfully used in Western blot, immunocytochemistry and immunoprecipitation procedures. By Western blot, this antibody detects an ~110 kDa protein representing SERCA2 ATPase in rat cardiac tissue.

          The MA3-919 antigen is purified canine cardiac sarcoplasmic reticulum vesicles.

          Target Information

          This gene encodes one of the SERCA Ca(2+)-ATPases, which are intracellular pumps located in the sarcoplasmic or endoplasmic reticula of muscle cells. This enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol into the sarcoplasmic reticulum lumen, and is involved in regulation of the contraction/relaxation cycle. Mutations in this gene cause Darier-White disease, also known as keratosis follicularis, an autosomal dominant skin disorder characterized by loss of adhesion between epidermal cells and abnormal keratinization. Alternative splicing results in multiple transcript variants encoding different isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATPase Ca++ transporting cardiac muscle slow twitch 2; ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2; ATPase, Ca++ transporting, cardiac muscle, slow twitch 2; ATPase, Ca++ transporting, slow twitch 2; Ca(2+)-transport ATPase class 3; calcium ATPase; calcium pump 2; calcium-ATPase (EC 3.6.1.3); calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; cardiac Ca2+ ATPase; endoplasmic reticulum class 1/2 Ca(2+) ATPase; HK1; HK2; non-muscle ATPase; putative SERCA isoform; sarco(endo)plasmic reticulum Ca(2+)-dependent ATPase 2; sarco/endoplasmic reticulum Ca2+-ATPase 2; sarcoplasmic/endoplasmic-reticulum Ca(2+) pump gene 2; SERCA; SERCA ATPase; SERCA-2b class 2; SERCA2; SR Ca(2+)-ATPase 2; unnamed protein product

          View more View less

          Gene Aliases: 9530097L16Rik; ATP2; ATP2B; D5Wsu150e; DAR; DD; mKIAA4195; RHABDO2; SERCA2; SERCA2A; SERCA2B; SercaII

          View more View less

          UniProt ID: (Dog) O46674, (Human) P16615, (Rabbit) P20647, (Pig) P11607, (Rat) P11507, (Mouse) O55143

          View more View less

          Entrez Gene ID: (Dog) 403878, (Human) 488, (Rabbit) 100038308, (Pig) 396875, (Rat) 29693, (Guinea pig) 100174978, (Mouse) 11938

          View more View less

          Function(s)
          ATP binding metal ion binding nucleotide binding calcium channel regulator activity calcium-transporting ATPase activity calcium ion binding protein binding ATPase activity enzyme binding ion channel binding S100 protein binding calcium-transporting ATPase activity involved in regulation of cardiac muscle cell membrane potential lncRNA binding lutropin-choriogonadotropic hormone receptor binding primary active transporter
          Process(es)
          regulation of cardiac muscle cell membrane potential hydrogen ion transmembrane transport autophagosome assembly muscle system process calcium ion transport cellular calcium ion homeostasis regulation of muscle contraction cell adhesion epidermis development positive regulation of heart rate regulation of cardiac muscle contraction by calcium ion signaling autophagosome membrane docking endoplasmic reticulum calcium ion homeostasis positive regulation of endoplasmic reticulum calcium ion concentration ion transmembrane transport response to endoplasmic reticulum stress relaxation of cardiac muscle sarcoplasmic reticulum calcium ion transport calcium ion transmembrane transport regulation of cardiac muscle cell action potential involved in regulation of contraction organelle localization by membrane tethering calcium ion transport from cytosol to endoplasmic reticulum regulation of cardiac conduction calcium ion import into sarcoplasmic reticulum regulation of the force of heart contraction ER-nucleus signaling pathway organelle organization transition between fast and slow fiber cardiac muscle hypertrophy in response to stress T-tubule organization cellular response to oxidative stress negative regulation of heart contraction negative regulation of receptor binding regulation of calcium ion-dependent exocytosis of neurotransmitter regulation of heart contraction positive regulation of cardiac muscle cell apoptotic process response to lipopolysaccharide cellular response to heat relaxation of skeletal muscle response to peptide hormone
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch