Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ATP4B Antibodies

          Invitrogen

          ATP4B Monoclonal Antibody (2G11)

          17 Published Figures
          59 References
          View all (10) ATP4B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ATP4B Monoclonal Antibody (2G11)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Published Figures (17)
          ATP4B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ATP4B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ATP4B Antibody (MA3-923) in ICC/IF

          Immunofluorescent analysis of Hydrogen/Potassium ATPase beta using Anti-Hydrogen/Potassium ATPase beta Monoclonal Antibody (2G11) (Product # MA3-923) shows staining in A2058 Cells. Hydrogen/Potassium ATPase beta staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with or an antibody recognizing Hydrogen/Potassium ATPase beta (Product # MA3-923) at a dilution of 1:100 over night ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ATP4B Antibody in Immunocytochemistry (ICC/IF)
          ATP4B Antibody in Immunocytochemistry (ICC/IF)
          ATP4B Antibody in Immunocytochemistry (ICC/IF)
          ATP4B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP4B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP4B Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Immunohistochemistry (IHC)
          ATP4B Antibody in Western Blot (WB)
          ATP4B Antibody in Western Blot (WB)
          ATP4B Antibody in Flow Cytometry (Flow)
          ATP4B Antibody in Flow Cytometry (Flow)
          ATP4B Antibody in Flow Cytometry (Flow)
          ATP4B Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          ATP4B Monoclonal Antibody (2G11)

          Product Details

          MA3-923

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:4,000
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 37 publications 37 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:2,000
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:2,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 11 publications 11 publications

          Flow Cytometry (Flow)

          1/50
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication

          ChIP assay (ChIP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Human, Mustelid, Mouse, Pig, Rabbit, Rat

          Published species

          Bovine, Guinea pig, Human, Mouse, Rabbit, Rat, Rodent

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2G11

          Immunogen

          Purified 34 kDa core peptide from deglycosylated hog gastric microsomes.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          PBS, ascites

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2227726

          Product Specific Information

          MA3-923 detects the beta-subunit of hydrogen/potassium ATPase from bovine, human, canine, porcine, rabbit, mouse, ferret, and rat tissues.

          MA3-923 has been successfully used in Western blot, immunohistochemistry, immunocytochemistry, and immunoprecipitation procedures (ChIP). By Western blot, this antibody detects various forms of the beta-subunit, including a 60 to 80 kDa glycosylated protein, a 52 kDa beta-subunit precursor, and the 34 kDa core peptide from rabbit stomach extracts. Immunohistochemical staining of hydrogen/potassium ATPase in rabbit gastric glands with MA3-923 results in specific staining of parietal cells.

          MA3-923 has been shown to inhibit the enzymatic activity of the hydrogen/potassium ATPase and to alter the affinity of the cytoplasmic potassium binding site.

          The MA3-923 antigen is purified 34 kDa core peptide from deglycosylated hog gastric microsomes. This antibody recognizes an epitope between amino acid residues 1-13 or 15-28 located on the cytoplasmic side of the beta-subunit.

          Target Information

          The hydrogen/potassium ATPase, or gastric proton pump, belongs to a family of P-type cation-transporting ATPases. This family of ATPases shares a number of functional and structural similarities including the common feature of consisting of an alpha and beta subunit. Like the ubiquitous sodium/potassium ATPase, the hydrogen/potassium ATPase consists of a large transmembrane catalytic subunit, termed the alpha- subunit which contains sites for ATP binding and phosphorylation, and an associated smaller glycoprotein, termed the beta-subunit which may play a role in maintaining the structural and functional integrity of the complex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATPase H+/K+ transporting beta subunit; ATPase H+K+-transporting beta / (gastric HK-ATPase beta subunit) defined by SSR; ATPase, H+,K+-transporting, beta / (gastric H,K-ATPase beta subunit), defined by SSR; ATPase, H+/K+ exchanging, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide, gastric specific; gastric H(+)/K(+) ATPase subunit beta; gastric H+/K+ ATPase beta subunit; gastric H+/K+-ATPase beta subunit; gastric hydrogen-potassium ATPase, beta; gp60-90; H+,K+-ATPase; H+/K+ ATPase beta; H+/K+ ATPase beta subunit; H+/K+-ATPase beta; H+/K+-ATPase beta subunit; H,K-ATPase-Beta; H.K-ATPase, beta subunit; potassium-transporting ATPase beta chain; proton pump beta chain; unnamed protein product

          View more View less

          Gene Aliases: ATP6B; S76401S1

          View more View less

          UniProt ID: (Pig) P18434, (Rabbit) P18597, (Human) P51164, (Dog) P33704, (Rat) P18598, (Mouse) P50992

          View more View less

          Entrez Gene ID: (Pig) 397131, (Rabbit) 100009125, (Human) 496, (Bovine) 614308, (Dog) 404020, (Rat) 24217, (Mouse) 11945

          View more View less

          Function(s)
          protein binding ATPase activator activity hydrogen:potassium-exchanging ATPase activity heterocyclic compound binding primary active transporter
          Process(es)
          cell adhesion establishment or maintenance of transmembrane electrochemical gradient positive regulation of ATPase activity potassium ion transport sodium ion transport cellular sodium ion homeostasis cellular potassium ion homeostasis response to lipopolysaccharide sodium ion export from cell pH reduction hydrogen ion transmembrane transport potassium ion import across plasma membrane potassium ion transmembrane transport glycoprotein biosynthetic process regulation of proton transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-x4rxd:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0