Hamburger Menu Button
Thermo Fisher Scientific Logo
로그인
회원이 아니신가요? 계정 생성하기
  • 모든 제품 주문
    • 항체
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR 분석
    • 피펫 및 피펫 팁
    • 실험실용 원심분리기
    • 초저온 냉동고
    • 분광학
    • 비이커
    • PCR 장비 및 용품
    • 제품 카테고리 전체보기
  • 응용 분야 및 기법
    • 세포 분석
    • 실험실 장비
    • Real-Time PCR
    • PCR
    • 크로마토그래피
    • 세포 배양 및 트랜스펙션
    • DNA/RNA 추출과 분석
    • 단백질 생물학
    • 유세포분석
    • 화학 제품
    • 응용 분야 및 기법 전체보기
  • 서비스
    • 맞춤형 서비스
    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 신제품
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo

Search

전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          • Primary Antibodies ›
          • RyR1 Antibodies

          Invitrogen

          RyR1 Monoclonal Antibody (34C)

          48 Published Figures
          194 References
          View all (7) RyR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RyR1 Monoclonal Antibody (34C)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (48)
          RyR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          RyR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 50

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RyR1 Antibody (MA3-925) in IHC (P)

          Immunohistochemistry was performed on biopsies of deparaffinized Human skeletal muscle tissue. To expose target proteins, heat induced antigen retrieval was performed using 10mM sodium citrate (pH6.0) buffer, microwaved for 8-15 minutes. Following antigen retrieval tissues were blocked in 3% BSA-PBS for 30 minutes at room temperature. Tissues were then probed at a dilution of 1:20 with a mouse monoclonal an... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RyR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Immunocytochemistry (ICC/IF)
          RyR1 Antibody in Immunocytochemistry (ICC/IF)
          RyR1 Antibody in Immunocytochemistry (ICC/IF)
          RyR1 Antibody in Immunocytochemistry (ICC/IF)
          RyR1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          RyR1 Antibody in Immunocytochemistry (ICC/IF)
          RyR1 Antibody in Immunocytochemistry (ICC/IF)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry (IHC)
          RyR1 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          RyR1 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          RyR1 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          RyR1 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          RyR1 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)
          RyR1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA3-925

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000
          View 122 publications 122 publications

          Immunohistochemistry (IHC)

          -
          View 26 publications 26 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          1:1,000
          View 2 publications 2 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 31 publications 31 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 9 publications 9 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Amphibian, Bovine, Dog, Chicken, Fish, Human, Mustelid, Mouse, Non-human primate, Sheep, Rabbit, Rat

          Published species

          Bovine, Cat, Chicken, Dog, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          34C

          Immunogen

          Partially purified chicken pectoral muscle ryanodine receptor.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2254138

          Product Specific Information

          MA3-925 detects ryanodine receptor (RyR)-1 and RyR-2 isoforms in amphibian, chicken, bovine, canine, fish, human, non-human primate, mouse, rabbit, mink, rat and sheep tissues. In chickens this antibody detects the alpha, beta and cardiac isoforms. This antibody also detects RyR-3 in mouse cells. In frog, MA3-925 detects the alpha and beta isoforms and in fish it detects the alpha isoform.

          MA3-925 has been successfully used in Western blot, immunohistochemistry, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects a 565 kDa protein representing RyR from rat skeletal muscle extracts. In non-mammalian vertebrates, a doublet is seen at 565 kDa representing the alpha and beta isoforms of the receptor. Immunohistochemical staining of RyR in chicken brain with MA3-925 results in intense staining of cerebellar Purkinje neurons.

          The MA3-925 antigen is partially purified chicken pectoral muscle ryanodine receptor.

          Target Information

          Ryanodine receptors have been shown to play critical roles in the intracellular Ca2+ signaling occurring during cell activation in muscle cells and non-muscled cells. Ryanodine receptor, ~500 kDa, can be isolated from a variety of cell types such as lymphocytes, macrophages, granulocytes, fibroblasts, epithelial, endothelial cells, skeletal muscles, cardiac muscle and brain tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: calcium release channel; central core disease of muscle; channel; ryr1; ion channel; protein phosphatase 1, regulatory subunit 137; Ryanodine receptor; ryanodine receptor 1 (skeletal); ryanodine receptor 1, skeletal muscle; ryanodine receptor 1-like, skeletal muscle; sarcoplasmic reticulum calcium release channel; skeletal muscle calcium release channel; skeletal muscle ryanodine receptor; skeletal muscle-type ryanodine receptor; type 1 ryanodine receptor; type 1-like ryanodine receptor; unnamed protein product

          View more View less

          Gene Aliases: CCO; CMYO1A; CMYO1B; CMYP1A; CMYP1B; KDS; MHS; MHS1; PPP1R137; RYDR; RYR; RYR-1; Ryr1l; SKRR

          View more View less

          UniProt ID: (Rabbit) P11716, (Human) P21817, (Mouse) E9PZQ0

          View more View less

          Entrez Gene ID: (Sheep) 101112591, (Rabbit) 100009540, (Bovine) 100139857, (Human) 6261, (Dog) 606491, (Rat) 114207, (Mouse) 20190

          View more View less

          Function(s)
          protease binding ion channel activity ryanodine-sensitive calcium-release channel activity voltage-gated calcium channel activity calcium channel activity calcium ion binding calmodulin binding ATP binding calcium-release channel activity enzyme binding calcium-induced calcium release activity protein binding ligand-gated ion channel
          Process(es)
          response to hypoxia outflow tract morphogenesis ion transport calcium ion transport cellular calcium ion homeostasis muscle contraction striated muscle contraction release of sequestered calcium ion into cytosol by sarcoplasmic reticulum calcium-mediated signaling response to caffeine skin development ossification involved in bone maturation skeletal muscle fiber development release of sequestered calcium ion into cytosol protein homotetramerization regulation of cytosolic calcium ion concentration transmembrane transport calcium ion transmembrane transport cellular response to calcium ion cellular response to caffeine skeletal muscle contraction response to muscle activity negative regulation of transforming growth factor beta receptor signaling pathway response to estradiol regulation of response to osmotic stress response to electrical stimulus response to caloric restriction sarcoplasmic reticulum calcium ion transport cellular response to ATP regulation of muscle contraction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          About Thermo Fisher Plus Icon Minus Icon
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)

           

          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)

           

          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-xmwlv:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline