Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Sodium/Calcium Exchanger Antibodies

          Invitrogen

          Sodium/Calcium Exchanger Monoclonal Antibody (C2C12)

          Advanced Verification
          31 Published Figures
          96 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Sodium/Calcium Exchanger Monoclonal Antibody (C2C12)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (31)
          • Advanced Verification (1)
          Sodium/Calcium Exchanger Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Sodium/Calcium Exchanger Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 38

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Sodium/Calcium Exchanger Antibody (MA3-926) in ICC/IF

          Immunofluorescence analysis of Sodium/Calcium Exchanger was performed using 70% confluent log phase SH-SY5Y cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Sodium/Calcium Exchanger Monoclonal Antibody (C2C12) (Product # MA3-926) at 1:200 dilution ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Sodium/Calcium Exchanger Antibody in Immunocytochemistry (ICC/IF)
          Sodium/Calcium Exchanger Antibody in Immunocytochemistry (ICC/IF)
          Sodium/Calcium Exchanger Antibody in Immunocytochemistry (ICC/IF)
          Sodium/Calcium Exchanger Antibody in Immunocytochemistry (ICC/IF)
          Sodium/Calcium Exchanger Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Sodium/Calcium Exchanger Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Sodium/Calcium Exchanger Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Sodium/Calcium Exchanger Antibody in Immunohistochemistry (IHC)
          Sodium/Calcium Exchanger Antibody in Immunohistochemistry (IHC)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody in Western Blot (WB)
          Sodium/Calcium Exchanger Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA3-926

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 72 publications 72 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1/100
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:200
          View 15 publications 15 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Guinea pig, Human, Mouse, Rabbit, Rat

          Published species

          Bovine, Dog, Guinea pig, Human, Marsupial, Mouse, Pig, Rabbit, Rat, Rodent

          Host/Isotype

          Mouse / IgM

          Class

          Monoclonal

          Type

          Antibody

          Clone

          C2C12

          Immunogen

          Purified canine cardiac sodium/calcium exchanger.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography - MBP

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2191111

          Product Specific Information

          MA3-926 detects sodium/calcium exchanger from human, canine, rabbit, mouse, guinea pig and rat kidney and cardiac tissues.

          MA3-926 has been successfully used in Western blot, immunofluorescence, immunohistochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects a 120 kDa protein representing the sodium/calcium exchanger from guinea pig cardiac extract. The bands seen at 70 kDa and 160 kDa represent a proteolytic fragment and non-reduced exchanger respectively. MA3-926 is not recommended for Western blot procedures of rat tissues. Immunohistochemical staining of the sodium/calcium exchanger in rat heart with MA3-926 results in staining of the plasma membrane and intense staining of cardiac T-tubular membrane.

          The MA3-926 antigen is purified canine cardiac sodium/calcium exchanger. This antibody recognizes an epitope between amino acids 371-525, which is on the intracellular side of the plasma membrane.

          Target Information

          Ca2+ plays a critical role in intracellular signaling and thus regulation of calcium balance is critical for the maintenance of this signaling function. The plasma membrane Na+/Ca2+ (NaCX) exchangers mediate Na+-dependent Ca2+ efflux in a wide variety of cell types. NaCX can move Ca2+ either into or out of cells, depending on the net Na+, Ca2+, and K+ gradient across the membrane. In mammals, at least 5 distinct genes code for the exchangers: Three NaCX (NaCX-1, NaCX-2, and NaCX-3), and two in the NaCKX family (NaCKX-1 and NaCKX-2). NaCX-1 is most prominently expressed in the heart where it plays a major role in excitation-contraction coupling (Matsuoka et al., 1993). NACX-1 may also be a target for protection of the heart and brain from ischemia and reperfusion injury (Hagihara et al., 2005; Bano et al., 2005; Choi, 2005).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cardiac sarcolemmal sodium-calcium exchanger; isoform NCX1.13; isoform NCX1.2; isoform NCX1.3; isoform NCX1.7; isoform NCX1.9; Na(+)/Ca(2+)-exchange protein 1; Na+/C2+ exchanger; Na+/Ca++ exchanger; Na+/Ca2+ exchanger; Na-Ca exchanger; Na/Ca exchanger; NCX 1.41; NCX1; NKX; sodium-calcium exchanger; sodium/calcium exchanger; sodium/calcium exchanger isoform NaCa13; sodium/calcium exchanger isoform NaCa3; sodium/calcium exchanger isoform NaCa9; solute carrier family 8 (sodium/calcium exchanger), member 1; solute carrier family 8 member 1; unnamed protein product

          View more View less

          Gene Aliases: D930008O12Rik; NACA; Ncx; NCX1

          View more View less

          UniProt ID: (Dog) P23685, (Human) P32418, (Rat) Q01728, (Mouse) P70414, (Guinea pig) P48766

          View more View less

          Entrez Gene ID: (Dog) 475738, (Rabbit) 100328570, (Human) 6546, (Rat) 29715, (Mouse) 20541, (Guinea pig) 100135620

          View more View less

          Function(s)
          calcium:sodium antiporter activity calcium ion binding calmodulin binding ankyrin binding calcium:cation antiporter activity involved in regulation of postsynaptic cytosolic calcium ion concentration protein binding cytoskeletal protein binding ion channel binding secondary carrier transporter
          Process(es)
          cellular sodium ion homeostasis cell communication positive regulation of bone mineralization sodium ion transmembrane transport response to muscle stretch calcium ion import calcium ion transmembrane transport sodium ion import across plasma membrane positive regulation of the force of heart contraction regulation of postsynaptic cytosolic calcium ion concentration regulation of the force of heart contraction regulation of heart rate ion transport calcium ion transport cellular calcium ion homeostasis muscle contraction regulation of gene expression regulation of cell communication by electrical coupling regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion regulation of cardiac muscle contraction by calcium ion signaling vascular smooth muscle contraction cellular response to reactive oxygen species sodium ion export from cell relaxation of smooth muscle negative regulation of cytosolic calcium ion concentration cardiac muscle cell development calcium ion homeostasis transmembrane transport relaxation of cardiac muscle cardiac muscle contraction calcium ion transport into cytosol cellular response to caffeine membrane depolarization during cardiac muscle cell action potential cell communication by electrical coupling involved in cardiac conduction calcium ion transmembrane import into cytosol calcium ion export negative regulation of intracellular signal transduction regulation of cardiac conduction calcium ion export from cell response to hypoxia embryonic placenta development regulation of sodium ion transport heart morphogenesis sodium ion transport positive regulation of cytosolic calcium ion concentration response to nutrient response to xenobiotic stimulus response to glucose post-embryonic development positive regulation of fibroblast migration telencephalon development response to ATP embryonic heart tube development response to immobilization stress response to hydrogen peroxide regulation of calcium ion transport muscle cell development cellular response to cAMP cellular response to hypoxia inorganic cation import across plasma membrane export across plasma membrane regulation of system process regulation of cardiac muscle cell membrane potential
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-nnlvt:80/100.66.74.151:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0