Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MUC5AC Antibodies

          Invitrogen

          MUC5AC Monoclonal Antibody (45M1)

          52 Published Figures
          299 References
          View all (67) MUC5AC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MUC5AC Monoclonal Antibody (45M1)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (52)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 56

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MUC5AC Antibody (MA5-12178) in ICC/IF

          Immunofluorescence analysis of Mucin 5AC was performed using 70% confluent log phase A549 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with Mucin 5AC Mouse Monoclonal Antibody (Product # MA5-12178) at 2 µg/mL in 0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product # A2... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MUC5AC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MUC5AC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          MUC5AC Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          MUC5AC Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          MUC5AC Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          MUC5AC Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunocytochemistry (ICC/IF)
          MUC5AC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MUC5AC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MUC5AC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MUC5AC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MUC5AC Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (IHC)
          MUC5AC Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          MUC5AC Antibody in Western Blot (WB)
          MUC5AC Antibody in Western Blot (WB)
          MUC5AC Antibody in Western Blot (WB)
          MUC5AC Antibody in Western Blot (WB)
          MUC5AC Antibody in Western Blot (WB)
          MUC5AC Antibody in Western Blot (WB)
          MUC5AC Antibody in Western Blot (WB)
          MUC5AC Antibody in ELISA (ELISA)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          MUC5AC Monoclonal Antibody (45M1)

          Product Details

          MA5-12178

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 34 publications 34 publications

          Immunohistochemistry (IHC)

          -
          View 129 publications 129 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1-2 µg/mL
          View 7 publications 7 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          2-3 µg/mL
          View 68 publications 68 publications

          Flow Cytometry (Flow)

          -
          View 4 publications 4 publications

          ELISA (ELISA)

          -
          View 52 publications 52 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Dot blot (DB)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Chicken, Cat, Human, Mouse, Non-human primate, Pig, Rabbit, Rodent, Rat

          Published species

          Avian, Guinea pig, Hamster, Human, Mouse, Non-human primate, Pig, Plant, Rabbit, Rat, Virus

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          45M1

          Immunogen

          M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4, with 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10978001

          Product Specific Information

          MA5-12178 targets Mucin 5AC in IHC (P) and ICC/IF applications and shows reactivity with Chicken, Feline, Human, mouse, Non-human primate, Porcine, Rabbit, Rodent, and Rat samples.

          The MA5-12178 immunogen is m1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient.

          Target Information

          The antibody 2-25LE reacts with Lewisb blood group antigen, a carbohydrate determinant carried on both glycolipids and glycoproteins, detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: gastric mucin; lewis B blood group antigen; major airway glycoprotein; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5ac; mucin 5AC, oligomeric mucus/gel-forming pseudogene; mucin-5 subtype AC, tracheobronchial; tracheobronchial mucin; unnamed protein product

          View more View less

          Gene Aliases: 2210005L13Rik; AABR07006032.1; leB; MGM; MUC5; mucin; TBM

          View more View less

          UniProt ID: (Human) P98088

          View more View less

          Entrez Gene ID: (Rabbit) 100356613, (Pig) 100170143, (Human) 4586, (Pig) 110259210, (Rat) 682837, (Mouse) 17833

          View more View less

          Function(s)
          extracellular matrix structural constituent protein binding extracellular matrix protein
          Process(es)
          response to ozone response to sulfur dioxide epithelial cell differentiation response to lipopolysaccharide response to vitamin A maintenance of lens transparency cellular response to retinoic acid cellular response to epidermal growth factor stimulus cellular response to glucocorticoid stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch