Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cardiac Troponin T Antibodies

          Invitrogen

          Cardiac Troponin T Monoclonal Antibody (13-11)

          Advanced Verification
          84 Published Figures
          343 References
          View all (51) Cardiac Troponin T antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cardiac Troponin T Monoclonal Antibody (13-11)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Published Figures (84)
          • Advanced Verification (2)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 91

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cardiac Troponin T Antibody (MA5-12960) in ICC/IF

          Immunofluorescent analysis of Troponin T Cardiac Isoform (green) in cultured primary cardiomyocytes. Primary cardiomyocytes were isolated and cultured using the Primary Cardiomyocyte Isolation Kit (Product # 88281). Cells were grown in 24-well plates (seeded at 5x10^5 cells/well) or in 35mm glass bottom plates (at a density of 2.5 x 106 cells/well). At day 1 and day 7, cells were fixed with 4% paraformaldeh... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cardiac Troponin T Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cardiac Troponin T Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Cardiac Troponin T Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Cardiac Troponin T Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunocytochemistry (IHC, IHC (PFA), ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Cardiac Troponin T Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry (ICC/IF)
          Cardiac Troponin T Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Cardiac Troponin T Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cardiac Troponin T Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cardiac Troponin T Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cardiac Troponin T Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cardiac Troponin T Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry, Immunohistochemistry (PFA fixed) (IHC, IHC (PFA))
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (IHC)
          Cardiac Troponin T Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Cardiac Troponin T Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Cardiac Troponin T Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Cardiac Troponin T Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody in Western Blot (WB)
          Cardiac Troponin T Antibody
          Cardiac Troponin T Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA5-12960

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 15 publications 15 publications

          Immunohistochemistry (IHC)

          -
          View 93 publications 93 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1-2 µg/mL
          View 7 publications 7 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 4 publications 4 publications

          Immunohistochemistry (Frozen) (IHC (F))

          5 µg/mL
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          View 141 publications 141 publications

          Flow Cytometry (Flow)

          -
          View 73 publications 73 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Avian, Dog, Chicken, Fish, Guinea pig, Human, Mouse, Pig, Rabbit, Rat

          Published species

          Dog, Fish, Hamster, Human, Mouse, Pig, Rat, Xenopus, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          13-11

          Immunogen

          Purified rabbit cardiac troponin T isoform (TnT4R)

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11000742

          Product Specific Information

          MA5-12960 targets Troponin T Cardiac Isoform in IF/ICC, IHC (P), and IM applications and shows reactivity with Avian, Canine, Chicken, Fish, Guinea Pig, Human, mouse, Porcine, Rabbit, and Rat samples.

          The MA5-12960 immunogen is purified rabbit cardiac troponin T isoform (TnT4R).

          Target Information

          Cardiac Troponin T is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Cardiac Troponin T is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. Cardiac Troponin T expression in the human heart is affected by heart failure. Mutations within the Cardiac Troponin T protein have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for the Cardiac Troponin T gene undergo alternative splicing that results in many tissue-specific isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alternatively spliced; cardiac muscle troponin T; cardiac TnT; cardiac troponin 2; cardiac troponin T; cardiac troponin T form I; cardiac troponin T isoform I; cardiac troponin T2; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, hypertrophic 2; cTnT; MGC3889; tnTC; troponin T; Troponin T cardiac; troponin T type 2 (cardiac); Troponin T, cardiac muscle; Troponin T2; unnamed protein product

          View more View less

          Gene Aliases: CMD1D; CMH2; CMPD2; cTnT; Ctt; CTTG; LVNC6; RATCTTG; RCM3; Tnnt3; Tnt; TnTC

          View more View less

          UniProt ID: (Human) P45379, (Chicken) P02642, (Rat) P50753, (Mouse) P50752

          View more View less

          Entrez Gene ID: (Human) 7139, (Rabbit) 100009428, (Pig) 100622450, (Dog) 403532, (Chicken) 396433, (Guinea pig) 100379561, (Rat) 24837, (Mouse) 21956

          View more View less

          Function(s)
          microfilament motor activity actin binding protein binding tropomyosin binding troponin C binding troponin I binding identical protein binding structural constituent of cytoskeleton protein binding, bridging macromolecular complex binding actin binding motor protein
          Process(es)
          regulation of muscle contraction striated muscle contraction regulation of heart contraction muscle filament sliding negative regulation of ATPase activity positive regulation of ATPase activity sarcomere organization response to calcium ion ventricular cardiac muscle tissue morphogenesis cardiac muscle contraction muscle contraction heart development response to bacterium regulation of muscle filament sliding speed tissue remodeling actin crosslink formation atrial cardiac muscle tissue morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch