Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Aurora A Antibodies

          Invitrogen

          Aurora A Monoclonal Antibody (1F8)

          1 Reference
          View all (48) Aurora A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Aurora A Monoclonal Antibody (1F8)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Aurora A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Aurora A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Aurora A Antibody (MA5-15803) in ICC/IF

          Immunofluorescence analysis of HeLa cells using AURKA monoclonal antibody (Product # MA5-15803) (Green). Blue: DRAQ5 fluorescent DNA dye. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Aurora A Antibody in Immunocytochemistry (ICC/IF)
          Aurora A Antibody in Western Blot (WB)
          Aurora A Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA5-15803

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:200-1:1,000
          -

          Flow Cytometry (Flow)

          1:200-1:400
          -

          ELISA (ELISA)

          1:10,000
          -
          Product Specifications

          Species Reactivity

          Human, Non-human primate, Rat

          Published species

          Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1F8

          Immunogen

          Purified recombinant fragment of human AURKA expressed in E. Coli.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          ascites

          Contains

          0.03% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11154080

          Product Specific Information

          MA5-15803 targets AURKA in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human, Non-human primate, and Rat samples.

          The MA5-15803 immunogen is purified recombinant fragment of human AURKA expressed in E. Coli.

          MA5-15803 detects AURKA which has a predicted molecular weight of approximately 48kDa.

          Target Information

          AURKA (Aurora A) is a serine/threonine kinase that regulates mitosis by association in vivo with centrosomes. Overexpression of AURKA is observed in many types of cancer. Aurora A plays a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal. Aurora A is viewed as a potential target for anticancer drug treatment. Tissue specificity: Highly expressed in testis and weakly in skeletal muscle, thymus and spleen.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: AURAK; aurora 2; Aurora A Kinase; aurora-A; aurora-related kinase 1; aurora/IP; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; AURORA2; breast tumor-amplified kinase; EC 2.7.11.1; hARK1; ipl1- and aurora-related kinase 1; IPL1-related kinase; MGC34538; OTTHUMP00000031341; OTTHUMP00000166072; protein kinase; protein phosphatase 1, regulatory subunit 47; ratAurA; RP5-1167H4.6; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine-protein kinase 15; serine/threonine-protein kinase 6; serine/threonine-protein kinase aurora-A; serine/threonine-protein kinase Ayk1; unnamed protein product

          View more View less

          Gene Aliases: AIK; ARK-1; ARK1; AURA; BTAK; PPP1R47; STK15; STK6; STK7

          View more View less

          UniProt ID: (Human) O14965

          View more View less

          Entrez Gene ID: (Human) 6790, (Rat) 261730

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity protein serine/threonine/tyrosine kinase activity protein binding ATP binding protein kinase binding protein heterodimerization activity protein serine kinase activity molecular function activator activity ubiquitin protein ligase binding histone kinase activity (H3-S10 specific)
          Process(es)
          meiotic spindle organization microtubule cytoskeleton organization mitotic cell cycle chromatin remodeling protein phosphorylation spindle organization mitotic spindle organization spindle assembly involved in female meiosis I centrosome cycle mitotic centrosome separation response to wounding anterior/posterior axis specification negative regulation of gene expression peptidyl-serine phosphorylation regulation of protein stability response to estradiol positive regulation of proteasomal ubiquitin-dependent protein catabolic process regulation of cytokinesis negative regulation of apoptotic process positive regulation of translation positive regulation of mitotic cell cycle meiotic cell cycle centrosome localization cilium disassembly protein localization to centrosome positive regulation of mitochondrial fission liver regeneration positive regulation of oocyte maturation positive regulation of meiotic cell cycle process involved in oocyte maturation neuron projection extension G2/M transition of mitotic cell cycle regulation of G2/M transition of mitotic cell cycle negative regulation of protein binding regulation of microtubule-based process cell cycle G2/M phase transition positive regulation of mitotic nuclear division regulation of centrosome cycle protein autophosphorylation positive regulation of cell cycle process regulation of signal transduction by p53 class mediator
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-xmwlv:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline