Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CRK Antibodies

          Invitrogen

          Crk Monoclonal Antibody (3G11E8)

          View all (28) CRK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Crk Monoclonal Antibody (3G11E8)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          Crk Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Crk Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Crk Antibody (MA5-15892) in ICC/IF

          Immunofluorescence analysis of 3T3-L1 cells using CRK monoclonal antibody (Product # MA5-15892) (Green). Blue: DRAQ5 fluorescent DNA dye. Red: actin filaments have been labeled with phalloidin. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Crk Antibody in Immunocytochemistry (ICC/IF)
          Crk Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Crk Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Crk Antibody in Western Blot (WB)
          Crk Antibody in Western Blot (WB)
          Crk Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA5-15892

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:200-1:1,000
          -

          Flow Cytometry (Flow)

          1:200-1:400
          -

          ELISA (ELISA)

          1:10,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3G11E8

          Immunogen

          Purified recombinant fragment of human CRK expressed in E. Coli.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          ascites

          Contains

          0.03% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11153760

          Product Specific Information

          MA5-15892 targets CRK in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples.

          The MA5-15892 immunogen is purified recombinant fragment of human CRK expressed in E. Coli. .

          MA5-15892 detects CRK which has a predicted molecular weight of approximately 42kDa.

          Target Information

          The Crk-I and Crk-II forms differ in their biological activities. Crk-II has less transforming activity than Crk-I. Crk- II mediates attachment-induced MAPK8 activation, membrane ruffling and cell motility in a Rac-dependent manner. Involved in phagocytosis of apoptotic cells and cell motility via its interaction with DOCK1 and DOCK4. May regulate the EFNA5-EPHA3 signaling.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: avian sarcoma virus CT10 (v-crk) oncogene homolog; Crk3; CSAID-binding protein; CT-10 related kinase 3; cytokine suppressive anti-inflammatory drug binding protein; encoded by transcript variant 4; p38 MAP kinase; encoded by transcript variant 5; p38 MAP kinase; MAP kinase 14; MAP kinase Mxi2; MAP kinase p38 alpha; MAPK 14; MAPK14; MAX-interacting protein 2; mitogen-activated protein kinase p38 alpha; negative regulator of Crk tyrosine-phosphorylation; p38 kinase; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38alpha Exip; proto-oncogene c-Crk; SAPK2a; CSAIDS binding protein 1; stress-activated protein kinase 2A; unnamed protein product; v-crk avian sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog (avian) isoform CRK_1; v-crk sarcoma virus CT10 oncogene homolog (avian) isoform CRK_2; v-crk sarcoma virus CT10 oncogene-like protein

          View more View less

          Gene Aliases: c-Crk; Crk-I; Crk-II; Crk-III; Crk1; Crk3; CRKII; CrkIII; Crko; CSBP; CSBP1; CSBP2; CSPB1; EXIP; Mxi2; p38; p38-alpha; p38a; p38ALPHA; p38MAPK; PRKM14; PRKM15; RK; SAPK2A

          View more View less

          UniProt ID: (Human) P46108, (Mouse) Q64010, (Human) Q16539, (Mouse) P47811

          View more View less

          Entrez Gene ID: (Human) 1398, (Mouse) 12928, (Human) 1432, (Mouse) 26416

          View more View less

          Function(s)
          phosphotyrosine binding insulin-like growth factor receptor binding protein binding cytoskeletal protein binding SH3 domain binding enzyme binding kinase binding protein domain specific binding receptor signaling complex scaffold activity protein binding, bridging receptor tyrosine kinase binding ubiquitin protein ligase binding signaling adaptor activity SH2 domain binding protein phosphorylated amino acid binding scaffold protein binding protein tyrosine kinase binding ephrin receptor binding protein kinase activity protein serine/threonine kinase activity MAP kinase activity MAP kinase kinase activity ATP binding protein phosphatase binding mitogen-activated protein kinase p38 binding NFAT protein binding protein serine kinase activity kinase activity
          Process(es)
          MAPK cascade negative regulation of inflammatory response to antigenic stimulus regulation of transcription from RNA polymerase II promoter chemotaxis signal transduction cell surface receptor signaling pathway positive regulation of gene expression positive regulation of myotube differentiation platelet activation osteoclast differentiation stress-activated protein kinase signaling cascade positive regulation of cyclase activity positive regulation of interleukin-12 production negative regulation of hippo signaling intracellular signal transduction cellular response to vascular endothelial growth factor stimulus p38MAPK cascade signal transduction in response to DNA damage positive regulation of blood vessel endothelial cell migration positive regulation of erythrocyte differentiation positive regulation of myoblast differentiation vascular endothelial growth factor receptor signaling pathway positive regulation of muscle cell differentiation stress-activated MAPK cascade 3'-UTR-mediated mRNA stabilization cellular response to lipopolysaccharide cellular response to lipoteichoic acid cellular response to ionizing radiation cellular response to UV-B cellular senescence stress-induced premature senescence cellular response to virus regulation of cytokine production involved in inflammatory response positive regulation of myoblast fusion positive regulation of reactive oxygen species metabolic process DNA damage checkpoint cell morphogenesis cartilage condensation angiogenesis osteoblast differentiation placenta development response to dietary excess chondrocyte differentiation glucose metabolic process regulation of transcription, DNA-templated protein phosphorylation cellular response to DNA damage stimulus transmembrane receptor protein serine/threonine kinase signaling pathway skeletal muscle tissue development myoblast differentiation involved in skeletal muscle regeneration peptidyl-serine phosphorylation fatty acid oxidation regulation of ossification lipopolysaccharide-mediated signaling pathway response to muramyl dipeptide response to lipopolysaccharide response to insulin tumor necrosis factor-mediated signaling pathway response to muscle stretch positive regulation of protein import into nucleus positive regulation of transcription from RNA polymerase II promoter positive regulation of glucose import protein autophosphorylation striated muscle cell differentiation protein maturation regulation of cardiac muscle cell proliferation positive regulation of cardiac muscle cell proliferation bone development pyroptosis cellular response to tumor necrosis factor negative regulation of canonical Wnt signaling pathway positive regulation of brown fat cell differentiation regulation of synaptic membrane adhesion NLRP1 inflammasome complex assembly neuron migration response to yeast regulation of leukocyte migration lipid metabolic process enzyme linked receptor protein signaling pathway synapse assembly cell proliferation regulation of cell shape regulation of signal transduction positive regulation of smooth muscle cell migration dendrite development cell migration hippocampus development cerebral cortex development establishment of cell polarity actin cytoskeleton organization positive regulation of cell growth regulation of actin cytoskeleton organization regulation of cell adhesion mediated by integrin regulation of Rac protein signal transduction positive regulation of Rac protein signal transduction helper T cell diapedesis response to hepatocyte growth factor reelin-mediated signaling pathway response to hydrogen peroxide negative regulation of natural killer cell mediated cytotoxicity positive regulation of JNK cascade ephrin receptor signaling pathway regulation of dendrite development cell chemotaxis negative regulation of wound healing response to cholecystokinin cellular response to transforming growth factor beta stimulus cellular response to nitric oxide protein localization to membrane activation of GTPase activity postsynaptic specialization assembly cerebellar neuron development positive regulation of substrate adhesion-dependent cell spreading response to peptide regulation of intracellular signal transduction cellular response to nerve growth factor stimulus cellular response to insulin-like growth factor stimulus cellular response to endothelin negative regulation of cell motility regulation of T cell migration cell adhesion regulation of wound healing regulation of cell motility
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-xbl4w:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0