Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HIF-2 alpha Antibodies

          Invitrogen

          HIF-2 alpha Monoclonal Antibody (ep190b), DyLight™ 550

          View all (40) HIF-2 alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite HIF-2 alpha Monoclonal Antibody (ep190b), DyLight™ 550

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          HIF-2 alpha Monoclonal Antibody (ep190b), DyLight™ 550

          Product Details

          MA5-16024

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:150-1:300
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          -

          Flow Cytometry (Flow)

          1 µg
          -

          ELISA (ELISA)

          Assay-Dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          ChIP assay (ChIP)

          Assay-dependent
          -

          Gel Shift (EMSA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Hamster, Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          ep190b

          Immunogen

          Region between amino acids 535-631.
          3D Epitope / Immunogen

          Conjugate

          DyLight™ 550 DyLight™ 550 DyLight™ 550
          • Unconjugated
          • Biotin 
          • DyLight 650
          • HRP
          • Request custom conjugation

          Excitation/Emission Max

          556/570 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.73 mg/mL

          Amount

          73 µg

          Purification

          Protein G

          Storage buffer

          50mM sodium borate

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11156407

          Target Information

          HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: basic-helix-loop-helix-PAS protein MOP2; class E basic helix-loop-helix protein 73; endothelial PAS domain protein 1; Endothelial PAS domain-containing protein 1; EPAS-1; Hif like protein; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF-2-alpha; HIF-related factor; HIF1 alpha-like factor; HIF1alpha-like factor; HIF2-alpha; HLF (HIF1alpha-like factor); HRF; hypoxia inducible factor 2, alpha subunit; hypoxia inducible factor 2a; hypoxia inducible transcription factor 2alpha; hypoxia-inducible factor 2 alpha; hypoxia-inducible factor 2-alpha; member of PAS protein 2; mHLF; PAS domain-containing protein 2; unnamed protein product

          View more View less

          Gene Aliases: bHLHe73; ECYT4; HIF-2 alpha; HIF-2alpha; HIF2 alpha; HIF2A; HLF; HRF; MOP2; PASD2

          View more View less

          UniProt ID: (Human) Q99814, (Rat) Q9JHS1, (Mouse) P97481

          View more View less

          Entrez Gene ID: (Bovine) 282711, (Human) 2034, (Rat) 29452, (Chinese hamster) 100771160, (Mouse) 13819

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding transcription coactivator binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding sequence-specific DNA binding protein heterodimerization activity protein dimerization activity RNA polymerase II sequence-specific DNA binding transcription factor binding cobalt ion binding basic helix-loop-helix transcription factor
          Process(es)
          angiogenesis response to hypoxia blood vessel remodeling regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter signal transduction erythrocyte differentiation lung development oxygen homeostasis positive regulation of transcription from RNA polymerase II promoter myoblast fate commitment cellular response to hypoxia positive regulation of cold-induced thermogenesis regulation of protein neddylation negative regulation of transcription from RNA polymerase II promoter embryonic placenta development regulation of heart rate response to oxidative stress mitochondrion organization heart development visual perception gene expression hemopoiesis cell differentiation norepinephrine metabolic process norepinephrine biosynthetic process surfactant homeostasis cell maturation positive regulation of multicellular organismal process multicellular organismal iron ion homeostasis cellular response to lipopolysaccharide cellular response to interleukin-1 circulatory system development positive regulation of dopamine biosynthetic process positive regulation of lung alveolus development tube development positive regulation of transcription, DNA-templated tissue remodeling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline