Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SIAH2 Antibodies

          Invitrogen

          SIAH2 Monoclonal Antibody (24E6H3)

          View all (9) SIAH2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SIAH2 Monoclonal Antibody (24E6H3)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          SIAH2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SIAH2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SIAH2 Antibody (MA5-24780) in ICC/IF

          Immunocytochemistry analysis of SIAH2 in ovarian carcinoma (OVCR3) cells. Samples were incubated with SIAH2 monoclonal antibody (Product # MA5-24780). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SIAH2 Antibody in Immunocytochemistry (ICC/IF)
          SIAH2 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SIAH2 Monoclonal Antibody (24E6H3)

          Product Details

          MA5-24780

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-Dependent
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:40-1:50
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:40-1:50
          -

          Immunocytochemistry (ICC/IF)

          1:10
          -

          Flow Cytometry (Flow)

          1:10
          -
          Product Specifications

          Species Reactivity

          Fruit fly, Human, Mouse, Pig

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          24E6H3

          Immunogen

          Synthetic peptide made to an N-terminal portion of the drosophila SINA protein (within residues 1-100).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          tissue culture supernatant

          Contains

          0.1% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2717253

          Product Specific Information

          This is specific for human SIAH2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells.

          Target Information

          SIAH-2 (seven in absentia homolog 2) is an E3 ligase that catalyzes ubiquitination and proteasome-mediated degradation of protein substrates. SIAH-2 encodes a 324 amino acid protein that shares 77% identity with human SIAH-1 and 68% identity with the Drosophila sina (seven in absentia) gene, on which development of the Drosophila R7 photoreceptor is dependent. SIAH-2 targets TRAF2 (which regulates cell responses to stress and cytokines through the regulation of key stress-signaling cascades) for degradation under stress conditions such as hypoxia. It targets HIF-1 alpha prolyl hydroxylase 3 (PHD3) for degradation upon exposure to hypoxic conditions, which coincides with an increase in SIAH-2 transcription. SIAH-2 can decrease TNF-alpha-dependent induction of JNK activity and transcriptional activation of NFkB. SIAH-2 null mice subjected to hypoxia display an impaired respiratory response and reduced levels of hemoglobin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: anon-fast-evolving-2H7; CG9949-PA; CG9949-PB; CG9949-PC; EC 6.3.2; HL08111p; mSiah2; RING-type E3 ubiquitin transferase SIAH2; seven in absenti; seven in absentia 2; seven in absentia homolog 2; seven-in-absentia; siah-2; sina-PA; sina-PB; sina-PC; unnamed protein product

          View more View less

          Gene Aliases: anon-EST:fe2H7; CG9949; Dmel\CG9949; Dmel_CG9949; hSiah2; sia; SINA; Sinh2

          View more View less

          UniProt ID: (Human) O43255, (Mouse) Q06986, (Fruit fly) P21461

          View more View less

          Entrez Gene ID: (Human) 6478, (Pig) 100622531, (Mouse) 20439, (Fruit fly) 39884

          View more View less

          Function(s)
          transcription corepressor activity ubiquitin-protein transferase activity protein binding zinc ion binding ubiquitin conjugating enzyme binding ubiquitin protein ligase activity ubiquitin-protein ligase protein modifying enzyme
          Process(es)
          ubiquitin-dependent protein catabolic process small GTPase mediated signal transduction protein ubiquitination regulation of protein ubiquitination regulation of circadian rhythm negative regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process negative regulation of transcription, DNA-templated canonical Wnt signaling pathway negative regulation of canonical Wnt signaling pathway amyloid fibril formation regulation of apoptotic signaling pathway negative regulation of extrinsic apoptotic signaling pathway protein catabolic process sensory organ development R7 cell fate commitment proteasomal protein catabolic process positive regulation of proteasomal ubiquitin-dependent protein catabolic process enteroendocrine cell differentiation sevenless signaling pathway regulation of R7 cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-sf8lr:80/100.66.74.151:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline