Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SIAH1/SIAH2 Antibodies

          Invitrogen

          SIAH1/SIAH2 Monoclonal Antibody (8G7H12)

          1 Published Figure
          1 Reference
          View all (3) SIAH1/SIAH2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SIAH1/SIAH2 Monoclonal Antibody (8G7H12)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          SIAH1/SIAH2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SIAH1/SIAH2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SIAH1/SIAH2 Antibody (MA5-24781) in WB

          Western blot analysis of SIAH1/SIAH2. Samples were incubated with SIAH1/SIAH2 monoclonal antibody (Product # MA5-24781). There are two SINA-like E3 ligases, SINA and SINAH, in Drosophila. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SIAH1/SIAH2 Antibody in Western Blot (WB)
          SIAH1/SIAH2 Antibody in RNA Immunoprecipitation (RIP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA5-24781

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          -

          Immunoprecipitation (IP)

          1:10-1:500
          -

          RNA Immunoprecipitation (RIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Fruit fly, Human, Mouse, Pig, Rat, Zebrafish

          Published species

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          8G7H12

          Immunogen

          Synthetic peptide made to a C-terminal portion of the drosophila SINA protein (within residues 280-331).

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          tissue culture supernatant

          Contains

          0.1% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2717254

          Product Specific Information

          This is specific for human SIAH 1 and 2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells.

          Does not react with mouse.

          Target Information

          SIAH, the human homologue of the Drosophila seven in absentia (sina) gene, is a tumor suppressor protein that is expressed in intestinal epithelium and activated during apoptosis. Human SIAH proteins are produced as two distinct gene products, SIAH-1, and a slightly larger protein SIAH-2, which share a highly conserved C-terminal sequence and differ in their N-terminal regions. SIAH-1 is a 32 kDa protein that contains an N-terminal RING-finger domain, which is required for proteolysis, and a cystein-rich C-terminal domain, which regulates oligomerization and SIAH binding to target proteins. As a tumor suppressor, SIAH-1 binds DCC (deleted in colorectal cancer) and regulates DCC degradation via the ubiquitin-proteasome pathway. SIAH-1 also binds a Bcl-2 related protein, Bag-1, thereby inhibiting cell growth. The majority of SIAH-1 is localized to the nucleus, however a small percentage is detected in the cytoplasm. This nuclear localization suggests that SIAH proteins may interact with other nuclear matrix proteins and DNA.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: anon-fast-evolving-2H7; CG9949-PA; CG9949-PB; CG9949-PC; EC 6.3.2; FLJ08065; HL08111p; hSIAH1; mSiah2; RING-type E3 ubiquitin transferase SIAH1; RING-type E3 ubiquitin transferase SIAH2; seven in absenti; seven in absentia 1A; seven in absentia 2; seven in absentia homolog 1; seven in absentia homolog 2; seven-in-absentia; siah-1; siah-1a; siah-2; sina-PA; sina-PB; sina-PC; unnamed protein product

          View more View less

          Gene Aliases: anon-EST:fe2H7; BURHAS; CG9949; Dmel\CG9949; Dmel_CG9949; hSiah2; id:ibd2635; sia; SIAH; SIAH1A; SINA; Sinh2; zgc:56026

          View more View less

          UniProt ID: (Human) Q8IUQ4, (Human) O43255, (Rat) Q920M9, (Rat) Q8R4T2, (Mouse) Q06986, (Zebrafish) Q7ZVG6, (Fruit fly) P21461

          View more View less

          Entrez Gene ID: (Pig) 100520614, (Human) 6477, (Human) 6478, (Pig) 100622531, (Rat) 140941, (Rat) 140593, (Mouse) 20439, (Zebrafish) 80955, (Fruit fly) 39884

          View more View less

          Function(s)
          ubiquitin-protein transferase activity protein binding zinc ion binding ubiquitin conjugating enzyme binding identical protein binding ubiquitin protein ligase activity transcription corepressor activity protein homodimerization activity disordered domain specific binding transferase activity metal ion binding ubiquitin-protein ligase protein modifying enzyme
          Process(es)
          ubiquitin-dependent protein catabolic process apoptotic process nervous system development axon guidance anatomical structure morphogenesis protein ubiquitination protein catabolic process protein destabilization positive regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process neuron apoptotic process canonical Wnt signaling pathway amyloid fibril formation regulation of apoptotic signaling pathway positive regulation of intrinsic apoptotic signaling pathway small GTPase mediated signal transduction regulation of protein ubiquitination regulation of circadian rhythm negative regulation of apoptotic process negative regulation of transcription, DNA-templated negative regulation of canonical Wnt signaling pathway negative regulation of extrinsic apoptotic signaling pathway male meiosis I spermatogenesis post-embryonic development regulation of multicellular organism growth multicellular organism development sensory organ development R7 cell fate commitment proteasomal protein catabolic process positive regulation of proteasomal ubiquitin-dependent protein catabolic process enteroendocrine cell differentiation sevenless signaling pathway regulation of R7 cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch