Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • P2X7 Antibodies

          Invitrogen

          P2X7 Monoclonal Antibody (Hano43)

          View all (30) P2X7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite P2X7 Monoclonal Antibody (Hano43)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          P2X7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          P2X7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          P2X7 Antibody (MA5-28184) in WB

          Western Blot was performed using Anti-P2X7 Monoclonal Antibody (Hano43) (Product # MA5-28184) and a 49 kDa and 75 kDa band corresponding to P2X purinoceptor 7 was observed across rat tissues. Tissue extracts (60 µg lysate) of Rat Skeletal Muscle (Lane 1), Rat Kidney (Lane 2), Rat Lung (Lane 3), Rat Heart (Lane 4), Rat Prostate (Lane 5) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP0321BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # LC2002) by iBlot® 2 Dry Blotting System ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          P2X7 Antibody in Western Blot (WB)

          Product Details

          MA5-28184

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Rat / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          Hano43

          Immunogen

          A P2X7-expression construct and a final boost with P2X7-transfected HEK cells

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          250 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2745159

          Product Specific Information

          Rat anti Mouse P2X7 antibody, clone Hano43 recognizes the P2X purinoceptor 7, also known as P2X7. The P2X7 ATP receptor has a distinctive long C-terminal tail with multiple potential protein and lipid interaction motifs and is highly polymorphic.

          Target Information

          The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and is responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Activation of this nuclear receptor by ATP in the cytoplasm may be a mechanism by which cellular activity can be coupled to changes in gene expression. Multiple alternatively spliced variants have been identified, most of which fit nonsense-mediated decay criteria.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATP receptor; MGC20089; P2X purinoceptor 7; p2X receptor 7; P2X7; P2X7 purinoceptor; P2X7 receptor; P2Z; P2Z receptor; Purinergic receptor

          View more View less

          Gene Aliases: AI467586; P2rx7; P2X(7); P2x7; P2X7R

          View more View less

          UniProt ID: (Mouse) Q9Z1M0

          View more View less

          Entrez Gene ID: (Mouse) 18439

          View more View less

          Function(s)
          lipopolysaccharide binding purinergic nucleotide receptor activity receptor activity extracellular ATP-gated cation channel activity ion channel activity protein binding ATP binding channel activity ligand-gated ion channel
          Process(es)
          activation of MAPK activity cell morphogenesis phagolysosome assembly positive regulation of T cell mediated cytotoxicity positive regulation of protein phosphorylation protein phosphorylation membrane protein ectodomain proteolysis phospholipid transfer to membrane transport ion transport cation transport calcium ion transport cell volume homeostasis membrane budding inflammatory response mitochondrion organization plasma membrane organization response to mechanical stimulus response to bacterium response to organic substance response to zinc ion gene expression positive regulation of calcium ion transport into cytosol positive regulation of gene expression programmed cell death positive regulation of glutamate secretion positive regulation of gamma-aminobutyric acid secretion response to organic cyclic compound synaptic vesicle exocytosis protein processing phospholipid scrambling sensory perception of pain cytolysis positive regulation of bone mineralization cellular response to extracellular stimulus bleb assembly positive regulation of prostaglandin secretion response to lipopolysaccharide positive regulation of interleukin-1 beta production positive regulation of interleukin-6 production collagen metabolic process response to ATP response to fluid shear stress positive regulation of ion transmembrane transport purinergic nucleotide receptor signaling pathway T cell proliferation response to drug T cell homeostasis positive regulation of apoptotic process positive regulation of catalytic activity NAD transport negative regulation of MAPK cascade positive regulation of MAPK cascade multicellular organismal protein catabolic process phospholipid translocation positive regulation of ossification negative regulation of bone resorption negative regulation of cell volume positive regulation of glycolytic process ceramide biosynthetic process pore complex assembly skeletal system morphogenesis homeostasis of number of cells within a tissue positive regulation of protein secretion positive regulation of cytokine secretion positive regulation of interleukin-1 alpha secretion positive regulation of interleukin-1 beta secretion defense response to Gram-positive bacterium release of sequestered calcium ion into cytosol protein oligomerization response to calcium ion response to electrical stimulus membrane depolarization positive regulation of mitochondrial depolarization positive regulation of lymphocyte apoptotic process cellular response to dsRNA cellular response to organic cyclic compound reactive oxygen species metabolic process extrinsic apoptotic signaling pathway cation transmembrane transport positive regulation of bleb assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline