Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD62E/CD62P Antibodies

          Invitrogen

          CD62E/CD62P Monoclonal Antibody (1.2B6), PE

          View all (7) CD62E/CD62P antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD62E/CD62P Monoclonal Antibody (1.2B6), PE

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CD62E/CD62P Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD62E/CD62P Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD62E/CD62P Antibody (MA5-28649) in Flow

          Flow cytometry analysis of CD62E/CD62P in PMA and ionomycin stimulated human endothelial cell line HUV-EC-C. Cells were stained with CD62E/CD62P Monoclonal Antibody (1.2B6), PE (Product # MA5-28649). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD62E/CD62P Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CD62E/CD62P Monoclonal Antibody (1.2B6), PE

          Product Details

          MA5-28649

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          10 µL/10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Pig

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1.2B6

          Immunogen

          TNF stimulated human endothelial cells
          3D Epitope / Immunogen

          Conjugate

          PE PE PE
          • APC 
          • Biotin 
          • FITC
          • PE-Cyanine5
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          PBS, pH 7.5, with proprietary stabilizer

          Contains

          <0.1% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2745608

          Target Information

          CD62E represents the 97 kDa type I transmembrane glycoprotein E-selectin, also known as endothelial-leukocyte adhesion molecule-1 (ELAM-1). It is expressed on cytokine-stimulated endothelium, weakly on unactivated endothelium, and is absent on leukocytes. Like other selectins, CD62E is an adhesion molecule that contributes to the initial attachment and rolling of leukocytes on endothelial surfaces, a prerequisite for leukocyte extravasation into tissues.CD62P is a 140 kDa membrane glycoprotein formerly known as platelet activation-dependent granule membrane protein (PADGEM) or GMP-140. It is expressed on platelets, megakaryocytes, and epithelium. It is thought to mediate the adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses and of activated platelets to neutrophils and monocytes in hemostasis. The monoclonal antibody 1.2B6 recognizes a common epitope shared by CD62E and CD62P.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD62 antigen-like family member E; CD62 antigen-like family member P; CD62P; CD62 homolog; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule 1; GMP-140; granule membrane protein 140; granule membrane protein 140kDa; granule membrane protein-140 (GMP-140) precursor; granulocyte membrane protein; LECAM-2; LECAM-3; LECAM2; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 3; platelet activation dependent granule-external membrane protein; platelet activation-dependent granule to external membrane protein; platelet alpha-granule membrane protein; RP1-117P20.2; selectin E (endothelial adhesion molecule 1); selectin P (granule membrane protein 140kDa antigen CD62) isoform SELP_1; selectin P (granule membrane protein 140kDa antigen CD62) isoform SELP_3; selectin P (granule membrane protein 140kDa, antigen CD62); unnamed protein product

          View more View less

          Gene Aliases: CD62; CD62E; CD62P; ELAM; ELAM1; ESEL; GMP140; GRMP; LECAM2; LECAM3; PADGEM; PSEL; selectin-e

          View more View less

          UniProt ID: (Human) P16581, (Pig) P98110, (Human) P16109

          View more View less

          Entrez Gene ID: (Human) 6401, (Pig) 397508, (Pig) 397150, (Human) 6403

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding sialic acid binding phospholipase binding oligosaccharide binding carbohydrate binding metal ion binding lipopolysaccharide binding integrin binding calcium ion binding heparin binding signaling receptor activity fucose binding glycosphingolipid binding calcium-dependent protein binding
          Process(es)
          positive regulation of receptor internalization leukocyte migration involved in inflammatory response inflammatory response cell adhesion heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules leukocyte cell-cell adhesion phospholipase C-activating G-protein coupled receptor signaling pathway calcium-mediated signaling actin filament-based process response to lipopolysaccharide response to cytokine response to tumor necrosis factor regulation of inflammatory response leukocyte tethering or rolling response to interleukin-1 positive regulation of leukocyte tethering or rolling positive regulation of platelet activation calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules regulation of integrin activation defense response to Gram-negative bacterium positive regulation of protein kinase B signaling cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-7n2k2:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0