Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Pan Ras Antibodies

          Invitrogen

          Pan Ras Recombinant Rabbit Monoclonal Antibody (JF10-11)

          View all (17) Pan Ras antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Pan Ras Recombinant Rabbit Monoclonal Antibody (JF10-11)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          Pan Ras Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Pan Ras Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Pan Ras Antibody (MA5-32561) in ICC/IF

          Immunocytochemical analysis of Pan Ras in PC-12 cells using a Pan Ras Monoclonal antibody (Product # MA5-32561) as seen in red. The nuclear counter stain is DAPI (blue). Cells were fixed in paraformaldehyde, permeabilised with 0.25% Triton X100/PBS. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Pan Ras Antibody in Immunocytochemistry (ICC/IF)
          Pan Ras Antibody in Immunocytochemistry (ICC/IF)
          Pan Ras Antibody in Immunocytochemistry (ICC/IF)
          Pan Ras Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Pan Ras Recombinant Rabbit Monoclonal Antibody (JF10-11)

          Product Details

          MA5-32561

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -

          Flow Cytometry (Flow)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          JF10-11

          Immunogen

          Synthetic peptide within Human Ras aa 21-64.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 40% Glycerol, 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2809838

          Product Specific Information

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

          Target Information

          This gene belongs to the Ras oncogene family, whose members are related to the transforming genes of mammalian sarcoma retroviruses. The products encoded by these genes function in signal transduction pathways. These proteins can bind GTP and GDP, and they have intrinsic GTPase activity. This protein undergoes a continuous cycle of de- and re-palmitoylation, which regulates its rapid exchange between the plasma membrane and the Golgi apparatus. Mutations in this gene cause Costello syndrome, a disease characterized by increased growth at the prenatal stage, growth deficiency at the postnatal stage, predisposition to tumor formation, mental retardation, skin and musculoskeletal abnormalities, distinctive facial appearance and cardiovascular abnormalities. Defects in this gene are implicated in a variety of cancers, including bladder cancer, follicular thyroid cancer, and oral squamous cell carcinoma. Multiple transcript variants, which encode different isoforms, have been identified for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: c-Ha-ras p21 protein; c-Ha-ras transgene; c-has/bas p21 protein; c-K-ras; c-K-ras2 protein; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; c-ras-Ki-2 protein; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; cellular transforming proto-oncogene; GTP- and GDP-binding peptide B; GTPase Hras1; H-ras 1 protein; H-Ras-1; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene; Harvey rat sarcoma virus oncogene 1; HRAS; K RAS; K-Ras 2; K-ras p21 protein; ki-Ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); Kirsten rat sarcoma viral proto-oncogene; KRAS-ENOPH fusion; Kras-Enoph1 fusion protein; N-ras oncogene p21; N-ras protein part 4; Neuroblastoma RAS; neuroblastoma ras oncogene; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; Nras; oncogene KRAS2; p19 H-RasIDX protein; p21 protein; p21ras; PR310 c-K-ras oncogene; PR371 c-K-ras oncogene; proto-oncogene GTPase; Ras family small GTP binding protein H-Ras; ras-like protein; RASK; RASN; small GTP binding protein; transformation gene: oncogene HAMSV; transforming protein N-Ras; transforming protein P21; unnamed protein product; v-Ha-ras Harvey rat sarcoma viral oncogene homolog; v-Ha-ras Harvey rat sarcoma viral oncogene homolog a; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog; v-ras neuroblastoma RAS viral oncogene homolog; xRAS1

          View more View less

          Gene Aliases: 'C-K-RAS; ALPS4; C-BAS/HAS; C-H-RAS; c-Ha-ras; C-HA-RAS1; C-K-RAS; c-Ki-ras; c-Ki-ras2; c-rasHa; CFC2; CMNS; CTLO; etID18709.8; fa04e08; fc14b12; fc23g10; fj89d12; fj99a10; H-ras; H-RASIDX; Ha-ras; HAMSV; Harvey-ras; hras; Hras-1; HRAS1; K-Ras; K-Ras 2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; Kras-2; KRAS1; KRAS2; N-ras; NCMS; NRAS1; NS; NS3; NS6; OES; p21; p21B; p21ras; RALD; RALD1; RALD2; ras; RASH1; RASK2; sb:eu826; wu:fa04e08; wu:fc14b12; wu:fc23g10; wu:fj89d12; wu:fj99a10; xRAS2; zgc:100761; zgc:110250; zgc:85725; zras-b1

          View more View less

          UniProt ID: (Human) P01112, (Mouse) Q61411, (Rat) P20171, (Human) P01116, (Mouse) P32883, (Rat) P08644, (Human) P01111, (Rat) Q04970

          View more View less

          Entrez Gene ID: (Human) 3265, (Mouse) 15461, (Rat) 293621, (Human) 3845, (Mouse) 16653, (Rat) 24525, (Human) 4893, (Mouse) 18176, (Rat) 24605, (Zebrafish) 550286, (Zebrafish) 445289, (Zebrafish) 30380

          View more View less

          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding GDP binding protein anchor phospholipase C activator activity macromolecular complex binding GMP binding LRR domain binding identical protein binding nucleotide binding small GTPase
          Process(es)
          MAPK cascade regulation of transcription from RNA polymerase II promoter chemotaxis signal transduction cell surface receptor signaling pathway Ras protein signal transduction positive regulation of cell proliferation negative regulation of cell proliferation insulin receptor signaling pathway organ morphogenesis negative regulation of gene expression Schwann cell development positive regulation of cell migration regulation of actin cytoskeleton organization regulation of cell proliferation myelination positive regulation of MAPK cascade positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade positive regulation of Ras protein signal transduction positive regulation of epithelial cell proliferation regulation of cell cycle adipose tissue development positive regulation of ERK1 and ERK2 cascade cellular response to gamma radiation positive regulation of wound healing positive regulation of protein targeting to membrane cellular senescence regulation of neurotransmitter receptor localization to postsynaptic specialization membrane positive regulation of ruffle assembly positive regulation of miRNA metabolic process endocytosis small GTPase mediated signal transduction positive regulation of gene expression positive regulation of interferon-gamma production ERBB2-ERBB3 signaling pathway T-helper 1 type immune response defense response to protozoan negative regulation of neuron apoptotic process insulin-like growth factor receptor signaling pathway regulation of long-term neuronal synaptic plasticity T cell receptor signaling pathway oncogene-induced cell senescence intrinsic apoptotic signaling pathway liver development response to isolation stress positive regulation of DNA replication I-kappaB kinase/NF-kappaB signaling female pregnancy response to gravity cytokine-mediated signaling pathway positive regulation of TOR signaling negative regulation of programmed cell death response to glucocorticoid response to mineralocorticoid myoblast proliferation cardiac muscle cell proliferation positive regulation of cellular senescence visual learning gene expression forebrain astrocyte development actin cytoskeleton organization regulation of synaptic transmission, GABAergic positive regulation of Rac protein signal transduction positive regulation of MAP kinase activity negative regulation of cell differentiation homeostasis of number of cells within a tissue positive regulation of nitric-oxide synthase activity striated muscle cell differentiation epithelial tube branching involved in lung morphogenesis positive regulation of endothelial cell proliferation regulation of protein phosphorylation regulation of gene expression positive regulation of angiogenesis positive regulation of fibroblast proliferation negative regulation of skeletal muscle tissue development myoblast differentiation heart development hemopoiesis embryonic cranial skeleton morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nbmw6:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline