Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PGC1 alpha/beta Antibodies

          Invitrogen

          PGC1 alpha/beta Recombinant Rabbit Monoclonal Antibody (JF09-71)

          Advanced Verification
          View all (4) PGC1 alpha/beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PGC1 alpha/beta Recombinant Rabbit Monoclonal Antibody (JF09-71)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          PGC1 alpha/beta Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PGC1 alpha/beta Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PGC1 alpha/beta Antibody (MA5-32563) in WB

          Western Blot was performed using Anti-PGC1 alpha/beta Recombinant Rabbit Monoclonal Antibody (JF09-71) (Product # MA5-32563) and a 90 kDa band corresponding to Peroxisome proliferator-activated receptor gamma coactivator 1-alpha was observed across tissues. Tissue extracts (40 µg lysate) of Mouse Heart (Lane 1), Mouse Kidney (Lane 2), Mouse Thymus (Lane 3), Mouse Placenta (Lane 4), Rat Heart (Lane 5) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP0321BOX). Resolved proteins were then transferred onto a nitrocell... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PGC1 alpha/beta Antibody in Western Blot (WB)
          PGC1 alpha/beta Antibody in Western Blot (WB)
          PGC1 alpha/beta Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PGC1 alpha/beta Recombinant Rabbit Monoclonal Antibody (JF09-71)

          Product Details

          MA5-32563

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          JF09-71

          Immunogen

          Synthetic peptide within human pgc1 alpha aa 755-798 and aa 269-323.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 40% Glycerol, 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2809840

          Product Specific Information

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

          Target Information

          PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ERR ligand 1; L-PGC-1alpha; ligand effect modulator-6; PERC; PERC-s; lacks one estrogen receptor interaction domain; peroxisome proliferative activated receptor gamma coactivator 1; peroxisome proliferative activated receptor, gamma, coactivator 1 alpha; peroxisome proliferative activated receptor, gamma, coactivator 1 beta; peroxisome proliferator-activated receptor gamma coactivator 1beta-2a; peroxisome proliferator-activated receptor gamma, coactivator 1 beta; PGC-1; pgc-1 alpha; pgc-1 beta; PGC-1-alpha; PGC-1-beta; PGC-1-related estrogen receptor alpha coactivator; PGC-1alpha; PGC1beta-1a; PGC1beta-1b; PGC1beta-2a; PGC1beta-2b; PPAR gamma coactivator 1alpha variant form-1; PPAR gamma coactivator variant form; PPAR gamma coactivator-1beta protein; PPAR-gamma coactivator 1-alpha; PPAR-gamma coactivator 1-beta; PPARAGCIalpha; PPARAGCIbeta; PPARgamma coactivator 1 beta; PPARgamma coactivator 1alpha; PPARGC-1-alpha; PPARGC-1-beta; Ppargc1a-vf1; transcriptional coactivator; transcriptional coactivator; PGC-1; unnamed protein product

          View more View less

          Gene Aliases: 4631412G21Rik; A830037N07Rik; ERRL1; Gm11133; LEM6; LRPGC1; PERC; PGC-1; PGC-1(alpha); PGC-1(beta); PGC-1alpha; PGC-1beta; PGC-1beta/ERRL1; PGC-1v; PGC1; PGC1A; PGC1B; PGC1beta; Pgco1; PGCvf; PGCvf-1; PGCvf1; PPARGC-1-alpha; PPARGC-1-beta; PPARGC1

          View more View less

          UniProt ID: (Human) Q9UBK2, (Human) Q86YN6, (Mouse) O70343, (Rat) Q9QYK2, (Mouse) Q8VHJ7, (Rat) Q811R2

          View more View less

          Entrez Gene ID: (Human) 10891, (Human) 133522, (Mouse) 19017, (Rat) 83516, (Mouse) 170826, (Rat) 291567

          View more View less

          Function(s)
          nucleic acid binding DNA binding transcription cofactor activity transcription coactivator activity RNA binding protein binding ligand-dependent nuclear receptor binding chromatin DNA binding ubiquitin protein ligase binding sequence-specific DNA binding RNA polymerase II sequence-specific DNA binding transcription factor binding lncRNA binding DNA-binding transcription factor binding estrogen receptor binding AF-2 domain binding chromatin binding peroxisome proliferator activated receptor binding alpha-tubulin binding promoter-specific chromatin binding
          Process(es)
          negative regulation of protein phosphorylation response to dietary excess gluconeogenesis regulation of transcription, DNA-templated mitochondrion organization positive regulation of gene expression response to activity response to muscle activity negative regulation of smooth muscle cell migration respiratory electron transport chain developmental process circadian regulation of gene expression cellular response to oxidative stress regulation of circadian rhythm negative regulation of neuron apoptotic process positive regulation of gluconeogenesis negative regulation of glycolytic process positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of fatty acid oxidation negative regulation of smooth muscle cell proliferation multicellular organismal homeostasis adipose tissue development cellular response to follicle-stimulating hormone stimulus cellular response to transforming growth factor beta stimulus negative regulation of mitochondrial fission energy homeostasis cellular response to thyroid hormone stimulus positive regulation of cold-induced thermogenesis positive regulation of glomerular visceral epithelial cell apoptotic process positive regulation of vascular smooth muscle cell proliferation positive regulation of progesterone biosynthetic process negative regulation of receptor activity regulation of N-methyl-D-aspartate selective glutamate receptor activity intracellular estrogen receptor signaling pathway positive regulation of osteoclast differentiation response to nutrient response to glucocorticoid response to cAMP temperature homeostasis cellular glucose homeostasis transcription initiation from RNA polymerase II promoter mRNA processing digestion RNA splicing fatty acid oxidation response to starvation cellular respiration protein stabilization brown fat cell differentiation protein-containing complex assembly response to reactive oxygen species mitophagy response to hypoxia response to ischemia galactose metabolic process response to xenobiotic stimulus response to fructose skeletal muscle atrophy response to electrical stimulus involved in regulation of muscle adaptation cerebellum development forebrain development response to nutrient levels cellular response to potassium ion response to leucine response to morphine response to electrical stimulus cellular response to lipopolysaccharide cellular response to nitrite cellular response to caffeine cellular response to fructose stimulus cellular response to glucose stimulus cellular response to interleukin-6 cellular response to tumor necrosis factor cellular response to estradiol stimulus cellular response to fatty acid cellular response to hypoxia response to epinephrine response to norepinephrine response to thyroid hormone response to dehydroepiandrosterone cellular response to ionomycin cellular response to resveratrol response to methionine adaptive thermogenesis response to hypobaric hypoxia
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0