Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NR4A1 (Nur77) Antibodies

          Invitrogen

          NUR77 Recombinant Rabbit Monoclonal Antibody (JM59-11)

          2 Published Figures
          3 References
          View all (34) NR4A1 (Nur77) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NUR77 Recombinant Rabbit Monoclonal Antibody (JM59-11)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          • Published Figures (2)
          NUR77 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NUR77 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NUR77 Antibody (MA5-32647) in ICC/IF

          Immunocytochemical analysis of NUR77 in NIH-3T3 cells using a NUR77 Monoclonal antibody (Product # MA5-32647) as seen in green. The nuclear counter stain is DAPI (blue). Cells were fixed in paraformaldehyde, permeabilised with 0.25% Triton X100/PBS. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NUR77 Antibody in Immunocytochemistry (ICC/IF)
          NUR77 Antibody in Immunocytochemistry (ICC/IF)
          NUR77 Antibody in Immunocytochemistry (ICC/IF)
          NUR77 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NUR77 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NUR77 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NUR77 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NUR77 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NUR77 Antibody in Western Blot (WB)
          NUR77 Antibody in Flow Cytometry (Flow)
          NUR77 Antibody in Flow Cytometry (Flow)
          NUR77 Antibody in Western Blot (WB)
          NUR77 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NUR77 Recombinant Rabbit Monoclonal Antibody (JM59-11)

          Product Details

          MA5-32647

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:5,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:5000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:1000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          JM59-11

          Immunogen

          Synthetic peptide within Human NUR77 aa 10-49.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.09 mg/mL

          Amount

          109 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 40% Glycerol, 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2809924

          Product Specific Information

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

          Target Information

          NR4A1 is a member of the steroid-thyroid hormone-retinoid receptor superfamily. NGFI-B-alpha is expressed in brain, adrenal, thyroid, liver, testis, ovary, thymus, muscle, lung and prostate. Its expression is induced in response to various stress stimuli and growth factors. The encoded protein acts as a nuclear transcription factor involved in mediating apoptotic signaling in thymocytes and tumor cells, as well as having a role in signaling in the hypothalamic-pituitary axis. Translocation of the protein from the nucleus to mitochondria induces apoptosis. Three transcript variants encoding two distinct isoforms have been identified for this NR4A1. Variant (3), also known as TRC beta, contains multiple differences compared to variant 1. The resulting isoform b contains a shorter and distinct C-terminus compared to isoform a. RXR has been shown to be a partner for NR4A1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: early response protein NAK1; growth factor-inducible nuclear protein N10; hormone receptor; immediate early gene transcription factor NGFI-B; MGC9485; nerve growth factor induced protein I-B; NR4A1; nuclear hormone receptor NUR/77; nuclear protein N10; Orphan nuclear receptor HMR; orphan nuclear receptor TR3; Orphan receptor tr3; ST-59; steroid receptor TR3; testicular receptor 3; TR3 orphan receptor; unnamed protein product

          View more View less

          Gene Aliases: Gfrp; GFRP1; Hbr-1; Hbr1; HMR; N10; NAK-1; NGFI-B; NGFIB; NP10; NUR77; NUR77-1; NUR77-2; TIS1; TR3

          View more View less

          UniProt ID: (Human) P22736, (Rat) P22829, (Mouse) P12813

          View more View less

          Entrez Gene ID: (Human) 3164, (Rat) 79240, (Mouse) 15370

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding lipopolysaccharide binding DNA binding double-stranded DNA binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding zinc ion binding glucocorticoid receptor binding identical protein binding sequence-specific DNA binding protein heterodimerization activity sequence-specific double-stranded DNA binding transcription factor binding
          Process(es)
          positive regulation of endothelial cell proliferation cell migration involved in sprouting angiogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter signal transduction intracellular receptor signaling pathway detection of lipopolysaccharide endothelial cell chemotaxis cellular response to vascular endothelial growth factor stimulus cellular response to fibroblast growth factor stimulus fat cell differentiation negative regulation of cell cycle positive regulation of transcription from RNA polymerase II promoter regulation of type B pancreatic cell proliferation cellular response to corticotropin-releasing hormone stimulus non-canonical inflammasome complex assembly response to amphetamine protein import into nucleus apoptotic process neurotransmitter secretion involved in regulation of skeletal muscle contraction response to lipopolysaccharide skeletal muscle cell differentiation macrophage activation positive regulation of apoptotic process positive regulation of transcription, DNA-templated response to electrical stimulus regulation of apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline