Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PRMT1 Antibodies

          Invitrogen

          PRMT1 Recombinant Rabbit Monoclonal Antibody (JA11-17)

          View all (22) PRMT1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PRMT1 Recombinant Rabbit Monoclonal Antibody (JA11-17)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          PRMT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PRMT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PRMT1 Antibody (MA5-32666) in ICC/IF

          Immunocytochemical analysis of PRMT1 in SW480 cells using a PRMT1 Monoclonal antibody (Product # MA5-32666) as seen in green. The nuclear counter stain is DAPI (blue). Cells were fixed in paraformaldehyde, permeabilised with 0.25% Triton X100/PBS. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PRMT1 Antibody in Immunocytochemistry (ICC/IF)
          PRMT1 Antibody in Immunocytochemistry (ICC/IF)
          PRMT1 Antibody in Immunocytochemistry (ICC/IF)
          PRMT1 Antibody in Immunocytochemistry (ICC/IF)
          PRMT1 Antibody in Immunocytochemistry (ICC/IF)
          PRMT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PRMT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PRMT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PRMT1 Recombinant Rabbit Monoclonal Antibody (JA11-17)

          Product Details

          MA5-32666

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          JA11-17

          Immunogen

          Synthetic peptide within Human PRMT1 aa 12-61.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 40% Glycerol, 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2809943

          Product Specific Information

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

          Target Information

          PRMT1 is a member of the protein arginine N-methyltransferase (PRMT) family. Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Increased expression of PRMT1 may play a role in various types of cancer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: arginine N-methyltransferase 1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; highly conserved protein 1; histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; HRMT1L2; interferon receptor 1-bound protein 4; Mrmt1; protein arginine N-methyltransferase 1 isoform 10; protein arginine N-methyltransferase 1 isoform 11; protein arginine N-methyltransferase 1 isoform 12; protein arginine N-methyltransferase 1 isoform 13; protein arginine N-methyltransferase 1 isoform 14; protein arginine N-methyltransferase 1 isoform 15; protein arginine N-methyltransferase 1 isoform 16; protein arginine N-methyltransferase 1 isoform 17; protein arginine N-methyltransferase 1 isoform 18; protein arginine N-methyltransferase 1 isoform 19; protein arginine N-methyltransferase 1 isoform 20; protein arginine N-methyltransferase 1 isoform 21; protein arginine N-methyltransferase 1 isoform 22; protein arginine N-methyltransferase 1 isoform 23; protein arginine N-methyltransferase 1 isoform 24; protein arginine N-methyltransferase 1 isoform 25; protein arginine N-methyltransferase 1 isoform 26; protein arginine N-methyltransferase 1 isoform 27; protein arginine N-methyltransferase 1 isoform 28; protein arginine N-methyltransferase 1 isoform 29; protein arginine N-methyltransferase 1 isoform 30; protein arginine N-methyltransferase 1 isoform 31; protein arginine N-methyltransferase 1 isoform 32; protein arginine N-methyltransferase 1 isoform 33; protein arginine N-methyltransferase 1 isoform 34; protein arginine N-methyltransferase 1 isoform 35; protein arginine N-methyltransferase 1 isoform 36; protein arginine N-methyltransferase 1 isoform 37; protein arginine N-methyltransferase 1 isoform 38; protein arginine N-methyltransferase 1 isoform 39; protein arginine N-methyltransferase 1 isoform 4; protein arginine N-methyltransferase 1 isoform 40; protein arginine N-methyltransferase 1 isoform 41; protein arginine N-methyltransferase 1 isoform 42; protein arginine N-methyltransferase 1 isoform 43; protein arginine N-methyltransferase 1 isoform 44; protein arginine N-methyltransferase 1 isoform 45; protein arginine N-methyltransferase 1 isoform 46; protein arginine N-methyltransferase 1 isoform 47; protein arginine N-methyltransferase 1 isoform 48; protein arginine N-methyltransferase 1 isoform 5; protein arginine N-methyltransferase 1 isoform 6; protein arginine N-methyltransferase 1 isoform 7; protein arginine N-methyltransferase 1 isoform 8; protein arginine N-methyltransferase 1 isoform 9; unnamed protein product

          View more View less

          Gene Aliases: 6720434D09Rik; ANM1; HCP1; HRMT1L2; IR1B4; Mrmt1

          View more View less

          UniProt ID: (Human) Q99873, (Mouse) Q9JIF0, (Rat) Q63009

          View more View less

          Entrez Gene ID: (Human) 3276, (Mouse) 15469, (Rat) 60421

          View more View less

          Function(s)
          RNA binding protein binding methyltransferase activity N-methyltransferase activity protein methyltransferase activity methyl-CpG binding arginine N-methyltransferase activity protein-arginine N-methyltransferase activity enzyme binding protein-arginine omega-N monomethyltransferase activity protein-arginine omega-N asymmetric methyltransferase activity histone methyltransferase activity identical protein binding histone methyltransferase activity (H4-R3 specific) mitogen-activated protein kinase p38 binding GATOR1 complex binding histone H4 methyltransferase activity S-adenosyl-L-methionine binding snoRNP binding S-adenosylmethionine-dependent methyltransferase activity
          Process(es)
          double-strand break repair via homologous recombination in utero embryonic development chromatin remodeling regulation of transcription, DNA-templated protein methylation cellular response to DNA damage stimulus positive regulation of cell proliferation RNA splicing negative regulation of translation peptidyl-arginine methylation peptidyl-arginine methylation, to asymmetrical-dimethyl arginine BMP signaling pathway regulation of BMP signaling pathway negative regulation of BMP signaling pathway neuron projection development cellular response to nutrient levels cellular response to amino acid starvation peptidyl-arginine omega-N-methylation TORC1 signaling negative regulation of apoptotic process positive regulation of erythrocyte differentiation regulation of megakaryocyte differentiation negative regulation of megakaryocyte differentiation positive regulation of translation negative regulation of translational initiation negative regulation of JNK cascade cardiac muscle tissue development protein homooligomerization cellular response to methionine protein localization to lysosome membraneless organelle assembly negative regulation of TORC1 signaling positive regulation of TORC1 signaling positive regulation of double-strand break repair via homologous recombination negative regulation of double-strand break repair via homologous recombination cell surface receptor signaling pathway viral protein processing positive regulation of hemoglobin biosynthetic process positive regulation of p38MAPK cascade peptidyl-arginine modification S-adenosylmethionine metabolic process liver regeneration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline