Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Nuclear Pore Complex Antibodies

          Invitrogen

          Nuclear Pore Complex Monoclonal Antibody (39C7)

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Nuclear Pore Complex Monoclonal Antibody (39C7)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          Nuclear Pore Complex Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Nuclear Pore Complex Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Nuclear Pore Complex Antibody (MA5-33097) in ICC/IF

          Immunocytochemistry analysis of Nuclear Pore Complex in HeLa cells, stained with Nuclear Pore Complex monoclonal antibody (Product # MA5-33097) using a dilution of 1:100 (red) and rabbit Anti-Vimentin antibody using a dilution of 1:10,000 (green), the blue is DAPI staining nuclear DNA. The Anti-Nuclear Pore Complex antibody reveals strong granular labeling of the nuclei corresponding to the protein, while t... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Nuclear Pore Complex Antibody in Immunocytochemistry (ICC/IF)
          Nuclear Pore Complex Antibody in Immunocytochemistry (ICC/IF)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA5-33097

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          39C7

          Immunogen

          Yeast nuclear preparations

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          25 µg

          Storage buffer

          tissue culture supernatant

          Contains

          10mM sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2802647

          Target Information

          The nuclear pore complex (NPC) serves as the sole bidirectional gateway of macromolecules in and out of the nucleus. Each NPC consists of ~1,000 protein subunits, termed nucleoporins, and totals to a molecular mass of ~110 Mda. NPCs maintain the integrity of the nuclear compartment by preventing macromolecules from freely diffusing in or out of the nucleus. (Annu Rev Biochem, 2019. PMID 30883195)

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 107 kDa nucleoporin; 133 kDa nucleoporin; 153 kDa nucleoporin; 155 kDa nucleoporin; 160 kDa nucleoporin; 205 kDa nucleoporin; 214 kDa nucleoporin; 35 kDa nucleoporin; 50 kDa nucleoporin; 62 kDa nucleoporin; 85 kDa nucleoporin; 88 kDa nuclear pore complex protein; 88 kDa nucleoporin; 93 kDa nucleoporin; 98 kDa nucleoporin; alternatively spliced; CAN protein, putative oncogene; CBP-interacting protein 4; gene trap locus 1-13 protein; gene trap locus-13; GLFG-repeat containing nucleoporin; karyoporin; KIAA0023; KIAA0095; KIAA0225; mitotic phosphoprotein 44; nuclear envelope pore membrane protein POM 210; nuclear pore associated protein; nuclear pore associated protein 60 kDa; nuclear pore complex 1; nuclear pore complex protein 88; nuclear pore complex protein hnup153; nuclear pore complex protein Nup53; nuclear pore complex protein Nup98; nuclear pore glycoprotein 62; nuclear pore membrane protein 210; nuclear pore protein gp210; nuclear pore-associated protein 60L; nucleoporin; nucleoporin 107kDa; nucleoporin 133kD; nucleoporin 133kDa; nucleoporin 153kD; nucleoporin 153kDa; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin 160kD; nucleoporin 160kDa; nucleoporin 205kDa; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 214kDa; nucleoporin 35kDa; nucleoporin 50 kDa; nucleoporin 50kD; nucleoporin 50kDa; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin 85kDa; nucleoporin 88kDa; nucleoporin 93kDa; nucleoporin 96; nucleoporin 98kD; nucleoporin 98kDa; nucleoporin Nup107; nucleoporin Nup133; nucleoporin Nup153; nucleoporin Nup155; nucleoporin Nup160; nucleoporin Nup205; nucleoporin Nup210; nucleoporin Nup214; nucleoporin Nup35; nucleoporin Nup50; nucleoporin NUP53; nucleoporin Nup62; nucleoporin Nup75; nucleoporin Nup84; nucleoporin NUP85; nucleoporin Nup88; nucleoporin Nup93; nucleoporin Nup98; nucleoporin p62 homolog; nucleoprotein 50; Nucleoprotein 50kD; Nup155p; NUP50; NUP62; Nup98-Nup96; NUP98/PHF23 fusion 2 protein; p105; P140; pericentrin-1; pore membrane protein of 210 kDa; predicted protein of HQ1146; preimplantation protein 2; unnamed protein product

          View more View less

          Gene Aliases: 160kDa; 1700030K07Rik; 2310006I24Rik; 2410008G02Rik; 2810011M03Rik; 35kDa; 3830404O05Rik; 4732457F17; 5330402E05Rik; 9830001L10; ADIR2; ATFB15; B130015D15Rik; C7orf14; CAIN; CAN; D2H9S46E; D7Ertd649e; D930027M19Rik; FADS4; FROUNT; GAMOS7; GAMOS8; gp190; GP210; Gtl-13; Gtl1-13; Gtl13; hNUP133; HNUP153; IBSN; IIAE9; mermaid; mKIAA0197; mKIAA0225; mKIAA0791; MP-44; MP44; N153; N155; NIC96; NO44; NP44; Np62; NPAP60; NPAP60L; NPHS11; NPHS13; NPHS17; NPHS18; NPHS19; NUCZINK; NUP196; NUP53; Nup75; NUP84; NUP96; Nup98-96; Nupc1; ODG6; ODG6; GAMOS7; p62; Pcnt1; POM210; Prei2; RGD1304977; Rtp60; SNDI

          View more View less

          UniProt ID: (Human) P57740, (Human) Q8WUM0, (Human) P49790, (Human) O75694, (Human) Q12769, (Human) Q92621, (Human) Q8TEM1, (Human) P35658, (Human) Q8NFH5, (Human) Q9UKX7, (Human) P37198, (Human) Q9BW27, (Human) Q99567, (Human) Q8N1F7, (Human) P52948, (Mouse) Q8BH74, (Mouse) Q8R0G9, (Mouse) E9Q3G8, (Rat) P37199, (Mouse) Q99P88, (Mouse) Q9Z0W3, (Rat) P11654, (Mouse) Q9QY81, (Mouse) Q80U93, (Rat) Q68FY1, (Mouse) Q8R4R6, (Rat) O08587, (Mouse) Q9JIH2, (Rat) P17955, (Mouse) Q63850, (Rat) Q4QQS8, (Mouse) Q8R480, (Mouse) Q8CEC0, (Rat) O08658, (Mouse) Q8BJ71, (Rat) Q66HC5, (Rat) P49793, (Mouse) Q6PFD9

          View more View less

          Entrez Gene ID: (Human) 57122, (Human) 55746, (Human) 9972, (Human) 9631, (Human) 23279, (Human) 23165, (Human) 23225, (Human) 8021, (Human) 129401, (Human) 10762, (Human) 23636, (Human) 79902, (Human) 4927, (Human) 9688, (Human) 4928, (Rat) 116555, (Mouse) 103468, (Mouse) 234865, (Rat) 292085, (Mouse) 218210, (Rat) 25281, (Rat) 117021, (Mouse) 170762, (Mouse) 59015, (Rat) 311182, (Mouse) 70699, (Rat) 362335, (Rat) 58958, (Mouse) 54563, (Rat) 296634, (Mouse) 227720, (Rat) 295692, (Mouse) 69482, (Rat) 25497, (Mouse) 18141, (Rat) 65274, (Mouse) 18226, (Rat) 287830, (Mouse) 445007, (Mouse) 19069, (Rat) 113929, (Mouse) 71805, (Rat) 291874, (Rat) 81738, (Mouse) 269966

          View more View less

          Function(s)
          protein binding structural constituent of nuclear pore nuclear localization sequence binding identical protein binding protein anchor molecular condensate scaffold activity nuclear export signal receptor activity nucleic acid binding phospholipid binding chromatin binding receptor signaling complex scaffold activity Hsp70 protein binding SH2 domain binding ubiquitin binding PTB domain binding Hsp90 protein binding transcription coactivator activity RNA binding mRNA binding peptide binding promoter-specific chromatin binding double-stranded DNA binding zinc ion binding small GTPase binding kinesin binding macromolecular complex binding thyroid hormone receptor binding CCR2 chemokine receptor binding
          Process(es)
          nucleocytoplasmic transport protein-containing complex assembly posttranscriptional tethering of RNA polymerase II gene DNA at nuclear periphery mRNA export from nucleus protein import into nucleus biological_process female gonad development nuclear pore complex assembly nephron development nuclear envelope organization poly(A)+ mRNA export from nucleus positive regulation of SMAD protein import into nucleus neural tube formation nuclear pore organization ribosomal large subunit export from nucleus ribosomal small subunit export from nucleus chemotaxis lamellipodium assembly positive regulation of transcription, DNA-templated macrophage chemotaxis transcription-dependent tethering of RNA polymerase II gene DNA at nuclear periphery neural tube development neurogenesis paraxial mesoderm development animal organ development system development somite development RNA export from nucleus protein targeting to nuclear inner membrane protein localization to nucleus production of miRNAs involved in gene silencing by miRNA atrial cardiac muscle cell action potential mitotic metaphase plate congression centrosome cycle mitotic centrosome separation cell surface receptor signaling pathway negative regulation of cell proliferation regulation of signal transduction negative regulation of epidermal growth factor receptor signaling pathway negative regulation of apoptotic process negative regulation of programmed cell death positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of epidermal growth factor receptor signaling pathway positive regulation of mitotic nuclear division regulation of Ras protein signal transduction negative regulation of Ras protein signal transduction positive regulation of centriole replication regulation of mitotic spindle organization centriole assembly positive regulation of mitotic cytokinetic process positive regulation of protein localization to centrosome NLS-bearing protein import into nucleus cellular response to leukemia inhibitory factor negative regulation of RNA export from nucleus amyloid fibril formation spermatogenesis regulation of protein import into nucleus negative regulation of MAP kinase activity cellular senescence protein export from nucleus intracellular protein transport regulation of nucleocytoplasmic transport regulation of cell cycle cytokine-mediated signaling pathway telomere tethering at nuclear periphery positive regulation of mRNA splicing, via spliceosome cell migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch