Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD11a/CD18 Antibodies

          Invitrogen

          CD11a/CD18 Monoclonal Antibody (24)

          View all (4) CD11a/CD18 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD11a/CD18 Monoclonal Antibody (24)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          MA5-33356

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Flow Cytometry (Flow)

          20 µg/mL
          -

          Immunoprecipitation (IP)

          Assay-Dependent
          -

          Functional Assay (Functional)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          24

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          100 µg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS with 0.1% BSA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2815494

          Product Specific Information

          Product is supplied at 1 mL (100 µL/mL). This antibody inhibits monocyte-dependent, antigen specific T cell proliferation and IL-2-activated natural killer cell assays. The antibody does not interfere with mitogen-stimulated T cell proliferation. Furthermore it prevents deadhesion of receptor/ligand pairs, possible locking leukocyte integrins in an active conformation.

          Application Notes: Flow cytometry - This antibody stains the extracellular domain of CD11/CD18. Functional Assay -This antibody acts as an inhibitor. The antibody was functionally tested by inhibition of antigen-specific T cell proliferation and LAK cell cytotoxy, which are LEA-1-dependent, and f-Met-Leu-Phe-stimulated chemotaxis, which is a CR3-dependent process. Positive control: PMN, monocytes, lymphocytes.

          Target Information

          CD11/CD18 (LFA-1) are beta 2-type integrins. The leukocyte integrins are a family of heterodimeric receptors that mediate divalent cation-dependent cellular adhesion reactions. T cells use integrins in essentially all of their functions. Integrins become active following signalling through other membrane receptors, which cause both affinity alteration and an increase in integrin clustering. The monoclonal antibody 24 recognizes a structural feature and is strictly dependent upon the presence of Mg2+. The epitope is located within, or in close proximity to, the three conserved cation binding domains and therefore a measure of Mg2+ bound to the leukocyte integrins and thus reflects functionally active molecules. The epitope can be induced on polymorphonuclear leukocytes and on monocytes. Glu173 and Glu175 of the beta(2) I domain are identified as critical for antibody 24 binding. The monoclonal antibody 24 inhibits monocyte-dependent, antigen specific T cell proliferation and IL-2- activated natural killer cell assays. The antibody does not interfere with mitogen-stimulated T cell proliferation. Furthermore the monoclonal antibody 24 prevents deadhesion of receptor/ligand pairs, possible locking leukocyte integrins in an active conformation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; CD11 antigen-like family member A; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; human neutrophil antigen 5; integrin beta chain, beta 2; integrin gene promoter; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte adhesion protein beta-subunit precursor; leukocyte cell adhesion molecule CD18; leukocyte function-associated molecule 1 alpha chain; leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C; LFA-1 alpha; LFA-1A; lymphocyte function-associated antigen 1, alpha polypeptide; putative; unnamed protein product

          View more View less

          Gene Aliases: CD11A; CD18; EV6; HNA-5; LAD; LCAMB; LFA-1; LFA1A; MAC-1; MF17; MFI7

          View more View less

          UniProt ID: (Human) P20701, (Human) P05107

          View more View less

          Entrez Gene ID: (Human) 3683, (Human) 3689

          View more View less

          Function(s)
          obsolete cell adhesion receptor activity protein binding ICAM-3 receptor activity signaling receptor activity macromolecular complex binding cell adhesion molecule binding beta-amyloid binding complement component C3b binding integrin binding protein kinase binding heat shock protein binding cargo receptor activity
          Process(es)
          T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell inflammatory response cell adhesion heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules neuron cell-cell adhesion leukocyte cell-cell adhesion cell-matrix adhesion signal transduction integrin-mediated signaling pathway cell adhesion mediated by integrin cell-cell adhesion mediated by integrin memory T cell extravasation T cell activation receptor clustering cell-cell adhesion microglial cell activation leukocyte migration involved in inflammatory response receptor-mediated endocytosis phagocytosis phagocytosis, engulfment apoptotic process I-kappaB kinase/NF-kappaB signaling cell-cell signaling regulation of cell shape neutrophil chemotaxis receptor internalization positive regulation of superoxide anion generation heterotypic cell-cell adhesion endodermal cell differentiation positive regulation of neutrophil degranulation endothelial cell migration cellular extravasation positive regulation of nitric oxide biosynthetic process positive regulation of angiogenesis negative regulation of dopamine metabolic process anatomical structure formation involved in morphogenesis regulation of peptidyl-tyrosine phosphorylation leukocyte adhesion to vascular endothelial cell cellular response to low-density lipoprotein particle stimulus positive regulation of protein targeting to membrane beta-amyloid clearance positive regulation of leukocyte adhesion to vascular endothelial cell neutrophil migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0