Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HSP90 Antibodies

          Invitrogen

          HSP90 Recombinant Rabbit Monoclonal Antibody (5D7Y6)

          View all (46) HSP90 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite HSP90 Recombinant Rabbit Monoclonal Antibody (5D7Y6)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          HSP90 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HSP90 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HSP90 Antibody (MA5-35624) in WB

          Western blot analysis of HSP90 in extracts of various cell lines. Samples were incubated with HSP90 Monoclonal antibody (Product # MA5-35624) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 10s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HSP90 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          HSP90 Recombinant Rabbit Monoclonal Antibody (5D7Y6)

          Product Details

          MA5-35624

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:20,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          5D7Y6

          Immunogen

          Recombinant Protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.1 mg/mL

          Amount

          110 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2849524

          Product Specific Information

          Immunogen sequence: LKEFEGKTLV SVTKEGLELP EDEEEKKKQE EKKTKFENLC KIMKDILEKK VEKVVVSNRL VTSPCCIVTS TYGWTANMER IMKAQALRDN STMGYMAAKK HLEINPDHSI IETLRQKAEA DKNDKSVKDL VILLYETALL SSGFSLEDPQ THANRIYRMI KLGLGIDEDD PTADDTSAAV TEEMPPLEGD DDTSRMEEVD

          Target Information

          HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 and mitochondria (Chen et al., 2005 ).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 84 kD heat shock protein; 90kDa heat shock protein; ATHS83; AtHsp90-1; ATHSP90.1; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; F6N7.13; F6N7_13; heat shock 84 kDa; heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein 1, beta; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 1, beta; HEAT SHOCK PROTEIN 81-1; HEAT SHOCK PROTEIN 83; heat shock protein 86; heat shock protein 90 alpha (cytosolic), class B member 1; heat shock protein 90 kDa; heat shock protein 90, alpha (cytosolic), class A member 1; HEAT SHOCK PROTEIN 90-1; heat shock protein 90.1; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein 90kDa alpha family class B member 1; heat shock protein family C member 1; heat shock protein family C member 3; heat shock protein, 1; heat shock protein, 84 kDa 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; heat-shock protein hsp84; heat-shock protein hsp86; HSP; HSP 84; HSP 86; HSP81-1; HSP81.1; HSP83; HSP84; HSP86; HSP90-beta; lipopolysaccharide-associated protein 2; LPS-associated protein 2; renal carcinoma antigen NY-REN-38; retinal degeneration slow protein; TSTA; tumor-specific transplantation 84 kDa antigen; tumor-specific transplantation 86 kDa antigen; unnamed protein product

          View more View less

          Gene Aliases: 86kDa; 89kDa; 90kDa; D6S182; EL52; HEL-S-65p; Hsp103; hsp4; HSP84; Hsp84-1; HSP86; Hsp86-1; Hsp89; HSP89A; Hsp90; HSP90-BETA; HSP90A; HSP90B; HSP90N; HSPC1; HSPC2; HSPCA; HSPCAL1; HSPCAL4; HSPCB; HSPN; LAP-2; LAP2

          View more View less

          UniProt ID: (Human) P07900, (Human) P08238, (Mouse) P07901, (Rat) P82995, (Mouse) P11499, (Rat) P34058

          View more View less

          Entrez Gene ID: (Human) 3320, (Human) 3326, (Mouse) 15519, (Rat) 299331, (Mouse) 15516, (Rat) 301252

          View more View less

          Function(s)
          UTP binding CTP binding RNA binding mRNA binding protein binding ATP binding GTP binding ATPase activity sulfonylurea receptor binding protein phosphatase binding MHC class II protein complex binding nitric-oxide synthase regulator activity TPR domain binding ubiquitin protein ligase binding dATP binding AP-3 adaptor complex binding identical protein binding protein homodimerization activity histone deacetylase binding protein binding involved in protein folding ion channel binding tau protein binding GTPase binding Rho GDP-dissociation inhibitor binding DNA polymerase binding scaffold protein binding disordered domain specific binding ATP-dependent protein folding chaperone enzyme-substrate adaptor activity protein tyrosine kinase binding double-stranded RNA binding protein kinase regulator activity kinase binding protein kinase binding heat shock protein binding peptide binding ATP-dependent protein binding cadherin binding protein dimerization activity protein phosphatase activator activity receptor ligand inhibitor activity histone methyltransferase binding protein carrier chaperone heterocyclic compound binding
          Process(es)
          placenta development protein folding virion attachment to host cell positive regulation of transforming growth factor beta receptor signaling pathway regulation of protein ubiquitination negative regulation of proteasomal ubiquitin-dependent protein catabolic process regulation of protein localization cellular response to heat positive regulation of protein import into nucleus negative regulation of apoptotic process negative regulation of neuron apoptotic process positive regulation of nitric oxide biosynthetic process positive regulation of cell differentiation positive regulation of cell size protein stabilization chaperone-mediated protein complex assembly regulation of cell cycle cellular response to interleukin-4 positive regulation of protein serine/threonine kinase activity fibril organization negative regulation of proteasomal protein catabolic process negative regulation of complement-dependent cytotoxicity telomerase holoenzyme complex assembly positive regulation of protein localization to cell surface response to unfolded protein telomere maintenance via telomerase neuron migration response to dietary excess activation of innate immune response positive regulation of defense response to virus by host skeletal muscle contraction nitric oxide biosynthetic process response to heat response to cold response to xenobiotic stimulus response to salt stress positive regulation of lamellipodium assembly cardiac muscle cell apoptotic process negative regulation of striated muscle cell apoptotic process protein import into mitochondrial matrix positive regulation of protein polymerization response to follicle-stimulating hormone positive regulation of interferon-beta production response to isolation stress cellular response to platelet-derived growth factor stimulus protein refolding response to cocaine regulation of apoptotic process response to estrogen positive regulation of cytotoxic T cell differentiation positive regulation of protein catabolic process response to antibiotic protein maturation positive regulation of cardiac muscle contraction response to caloric restriction cellular response to hydrogen peroxide cellular response to lipopolysaccharide cellular response to mechanical stimulus cellular response to hypoxia cellular response to virus regulation of postsynaptic membrane neurotransmitter receptor levels neurofibrillary tangle assembly regulation of protein complex stability chaperone-mediated autophagy mitochondrial transport positive regulation of telomere maintenance via telomerase regulation of protein complex assembly protein unfolding nitric oxide metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0