Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RUNX1/RUNX2/RUNX3 Antibodies

          Invitrogen

          RUNX1/RUNX2/RUNX3 Recombinant Rabbit Monoclonal Antibody (6G5U2)

          View all (7) RUNX1/RUNX2/RUNX3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RUNX1/RUNX2/RUNX3 Recombinant Rabbit Monoclonal Antibody (6G5U2)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RUNX1/RUNX2/RUNX3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RUNX1/RUNX2/RUNX3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RUNX1/RUNX2/RUNX3 Antibody (MA5-35649) in WB

          Western blot analysis of RUNX1/RUNX2/RUNX3 in extracts of various cell lines. Samples were incubated with RUNX1/RUNX2/RUNX3 Monoclonal antibody (Product # MA5-35649) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 90s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RUNX1/RUNX2/RUNX3 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          RUNX1/RUNX2/RUNX3 Recombinant Rabbit Monoclonal Antibody (6G5U2)

          Product Details

          MA5-35649

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          6G5U2

          Immunogen

          A synthetic peptide corresponding to a sequence within amino acids 354-453 of human RUNX1/2/3 (NP_0010018901)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.28 mg/mL

          Amount

          28 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2849549

          Product Specific Information

          Immunogen sequence: LPPPYPGSSQ AQGGPFQASS PSYHLYYGAS AGSYQFSMVG GERSPPRILP PCTNASTGSA LLNPSLPNQS DVVEAEGSHS NSPTNMAPSA RLEEAVWRPY

          Target Information

          Core binding factor (CBF) is a heterodimeric transcription factor that binds to the core element of many enhancers and promoters. The protein encoded by this gene represents the alpha subunit of CBF and is thought to be involved in the development of normal hematopoiesis. Chromosomal translocations involving this gene are welldocumented and have been associated with several types of leukemia. Three transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: acute myeloid leukemia 1 protein; acute myeloid leukemia 2 protein; acute myeloid leukemia 3 protein; acute myeloid leukemia gene 2; AKV core binding factor; aml1; AML1-ETO fusion; AML1-ETO fusion protein; AML1-EVI-1 fusion protein; CBF-alpha-3; CBFA2; core binding factor alpha 1; core binding factor alpha 2; core binding factor alpha 3; core-binding factor subunit alpha-2; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 1; core-binding factor, runt domain, alpha subunit 2; core-binding factor, runt domain, alpha subunit 3; mutant RUNX1; nuclear protein; oncogene AML-1; oncogene AML-2; oncogene AML-3; osteoblast-specific transcription factor 2; PEA2 alpha C; PEA2-alpha A; PEA2-alpha B; PEA2-alpha C; PEBP2 alpha A; PEBP2 alpha C; PEBP2-alpha A; PEBP2-alpha B; PEBP2-alpha C; polyomavirus enhancer-binding protein 2 alpha A subunit; polyomavirus enhancer-binding protein 2 alpha B subunit; polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 1; runt domain, alpha subunit 2; runt domain, alpha subunit 3; runt related transcription factor 1; runt related transcription factor 2; runt related transcription factor 3; RUNX 2; RUNX-2; RUNX1 with skipped exon 5; similar to Homo sapiens AML1c; acute myeloid leukemia 1; SL3-3 enhancer factor 1 alpha A subunit; SL3-3 enhancer factor 1 alpha B subunit; SL3-3 enhancer factor 1 alpha C subunit; SL3/AKV core-binding factor alpha A subunit; SL3/AKV core-binding factor alpha B subunit; SL3/AKV core-binding factor alpha C subunit; transcription factor; transcription factor AML2; transcription factor AML2/CBFA3; transcription factor Cbfa1/Osf2; unnamed protein product

          View more View less

          Gene Aliases: AML1; AML1-EVI-1; AML2; AML3; AMLCR1; Cbf; CBF-alpha-1; CBF-alpha-2; CBF2alpha; Cbfa-1; CBFA1; CBFA2; CBFA3; CCD; CCD1; CLCD; EVI-1; LS3; OSF-2; OSF2; PEA2aA; Pebp2a1; Pebp2a2; Pebp2a3; PEBP2aA; PEBP2aB; PEBP2aC; PEBP2alpha; Pebpa2a; Pebpa2b; Rx3

          View more View less

          UniProt ID: (Human) Q01196, (Human) Q13950, (Human) Q13761, (Mouse) Q08775, (Mouse) Q64131

          View more View less

          Entrez Gene ID: (Human) 861, (Human) 860, (Human) 864, (Mouse) 12394, (Mouse) 12393, (Mouse) 12399

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription corepressor binding transcription coactivator binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding calcium ion binding protein binding ATP binding protein homodimerization activity protein heterodimerization activity DNA-binding transcription factor binding sequence-specific double-stranded DNA binding bacterial-type RNA polymerase transcriptional activator activity, sequence-specific DNA binding promoter-specific chromatin binding core promoter proximal region sequence-specific DNA binding chromatin binding protein domain specific binding chromatin DNA binding histone deacetylase binding bHLH transcription factor binding general transcription initiation factor binding SMAD binding
          Process(es)
          skeletal system development ossification osteoblast differentiation endochondral ossification osteoblast fate commitment chondrocyte differentiation chondrocyte development osteoblast development regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transmembrane receptor protein serine/threonine kinase signaling pathway cell proliferation positive regulation of cell proliferation gene expression regulation of gene expression positive regulation of Schwann cell proliferation positive regulation of gene expression hemopoiesis neuron differentiation T cell differentiation regulation of ossification BMP signaling pathway positive regulation of chondrocyte differentiation embryonic forelimb morphogenesis ligamentous ossification regulation of fibroblast growth factor receptor signaling pathway odontogenesis of dentin-containing tooth regulation of odontogenesis of dentin-containing tooth regulation of cell differentiation regulation of osteoblast differentiation positive regulation of osteoblast differentiation positive regulation of ossification negative regulation of smoothened signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cell development cell maturation embryonic cranial skeleton morphogenesis skeletal system morphogenesis stem cell differentiation positive regulation of multicellular organismal process SMAD protein signal transduction response to sodium phosphate regulation of multicellular organismal development negative regulation of transcription from RNA polymerase II promoter myeloid leukocyte differentiation regulation of plasminogen activation myeloid cell differentiation negative regulation of granulocyte differentiation positive regulation of granulocyte differentiation positive regulation of interleukin-2 production positive regulation of collagen biosynthetic process negative regulation of CD4-positive, alpha-beta T cell differentiation positive regulation of CD8-positive, alpha-beta T cell differentiation positive regulation of angiogenesis peripheral nervous system neuron development positive regulation of developmental process regulation of cardiac muscle cell proliferation cardiac muscle tissue regeneration hematopoietic stem cell proliferation positive regulation of extracellular matrix organization regulation of connective tissue replacement protein phosphorylation negative regulation of cell cycle negative regulation of epithelial cell proliferation response to transforming growth factor beta in utero embryonic development liver development myeloid progenitor cell differentiation regulation of T cell anergy central nervous system development negative regulation of cell proliferation regulation of signal transduction hair follicle morphogenesis response to retinoic acid positive regulation of interferon-gamma production embryonic hemopoiesis behavioral response to pain neuron fate commitment neuron development definitive hemopoiesis regulation of hair follicle cell proliferation cellular response to transforming growth factor beta stimulus positive regulation of cell maturation positive regulation of progesterone secretion axon guidance neuron projection development positive regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nbmw6:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline