Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Histone Macro-H2A.1 Antibodies

          Invitrogen

          Histone Macro-H2A.1 Recombinant Rabbit Monoclonal Antibody (4D2O4)

          View all (32) Histone Macro-H2A.1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Histone Macro-H2A.1 Recombinant Rabbit Monoclonal Antibody (4D2O4)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          Histone Macro-H2A.1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Histone Macro-H2A.1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Histone Macro-H2A.1 Antibody (MA5-35752) in ICC/IF

          Immunofluorescence analysis of Histone Macro-H2A.1 in U-2 OS cells. Samples were incubated with Histone Macro-H2A.1 Monoclonal antibody (Product # MA5-35752) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Histone Macro-H2A.1 Antibody in Immunocytochemistry (ICC/IF)
          Histone Macro-H2A.1 Antibody in Immunocytochemistry (ICC/IF)
          Histone Macro-H2A.1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Histone Macro-H2A.1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Histone Macro-H2A.1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Histone Macro-H2A.1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Histone Macro-H2A.1 Recombinant Rabbit Monoclonal Antibody (4D2O4)

          Product Details

          MA5-35752

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          4D2O4

          Immunogen

          A synthetic peptide corresponding to a sequence within amino acids 200-300 of human macroH2A1 (O75367)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.95 mg/mL

          Amount

          95 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2849652

          Product Specific Information

          Immunogen sequence: IHSEISNLAG FEVEAIINPT NADIDLKDDL GNTLEKKGGK EFVEAVLELR KKNGPLEVAG AAVSAGHGLP AKFVIHCNSP VWGADKCEEL LEKTVKNCLA L

          Target Information

          Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: core histone; H2A histone family member Y; H2A histone family, member Y; H2A.y; H2A/y; histone H2A.y; histone macroH2A1; histone macroH2A1.1; histone macroH2A1.2; MACROH2A1.2; medulloblastoma antigen MU-MB-50.205; unnamed protein product

          View more View less

          Gene Aliases: H2A.y; H2A/y; H2AF12M; H2AFY; MACROH2A1.1; macroH2A1.2; mH2A1

          View more View less

          UniProt ID: (Human) O75367, (Rat) Q02874, (Mouse) Q9QZQ8

          View more View less

          Entrez Gene ID: (Human) 9555, (Rat) 29384, (Mouse) 26914

          View more View less

          Function(s)
          rDNA binding transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding DNA binding chromatin binding protein binding double-stranded methylated DNA binding enzyme binding protein kinase binding protein serine/threonine kinase inhibitor activity structural constituent of chromatin chromatin DNA binding nucleosomal DNA binding protein heterodimerization activity ADP-D-ribose binding ADP-D-ribose modification-dependent protein binding poly-ADP-D-ribose modification-dependent protein binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of oxidative phosphorylation DNA repair nucleosome assembly biological_process dosage compensation by inactivation of X chromosome regulation of lipid metabolic process heterochromatin assembly positive regulation of maintenance of mitotic sister chromatid cohesion regulation of gene expression, epigenetic positive regulation of keratinocyte differentiation negative regulation of gene expression, epigenetic positive regulation of gene expression, epigenetic regulation of chromatin silencing at rDNA establishment of protein localization to chromatin negative regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter regulation of NAD metabolic process negative regulation of cell cycle G2/M phase transition regulation of response to oxidative stress negative regulation of response to oxidative stress positive regulation of response to oxidative stress positive regulation of endodermal cell differentiation negative regulation of protein localization to chromosome, telomeric region protein poly-ADP-ribosylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline