Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PAK4/6/7 Antibodies

          Invitrogen

          Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Recombinant Rabbit Monoclonal Antibody (4E1H7)

          View all (13) PAK4/6/7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Recombinant Rabbit Monoclonal Antibody (4E1H7)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Antibody (MA5-35881) in WB

          Western blot analysis of Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) in extracts of various cell lines. Samples were incubated with Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Monoclonal antibody (Product # MA5-35881) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) secondary antibody at a dilution of 1:10,000. Both NIH/3T3 cells and C6 cells were treated by PMA/TPA (200 nM) at 37°C for 3... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Recombinant Rabbit Monoclonal Antibody (4E1H7)

          Product Details

          MA5-35881

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          4E1H7

          Immunogen

          A synthetic phosphorylated peptide around S474 of human PAK4PAK4 (O96013)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          200 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2849779

          Product Specific Information

          Immunogen sequence: RKSLV

          Target Information

          PAK4 is highly expressed in prostate, testis and colon. PAK4 interacts tightly with GTP-bound but not GDP-bound CDC42 and weakly with RAC. PAK4 phosphorylates and autophosphorylates and also activates the JNK pathway. PAK5, a member of the PAK family of protein kinases contains a CDC42/Rac1 interactive binding (CRIB) motif at the N-terminus and a Ste20-like kinase domain at the C-terminus. PAK5 preferentially binds to CDC42 in the presence of GTP and the CRIB motif is essential for this interaction. PAK6 protein cotranslocates into the nucleus with androgen receptor, which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development, in response to androgen. PAK6 was initially identified as an androgen receptor in a yeast two hybrid screen and was found to be highly expressed in testis and prostate tissues. Later experiments have shown it to be activated by MAP kinase 6 and p38 MAP kinase, suggesting that PAK6 may play a role in the cellular response to stress-related signals.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: p21 (CDKN1A)-activated kinase 4; p21 (CDKN1A)-activated kinase 6; p21 (CDKN1A)-activated kinase 7; p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 6; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 4; p21(CDKN1A)-activated kinase 6; p21(CDKN1A)-activated kinase 7; p21-activated kinase 4; p21-activated kinase 5; p21-activated kinase 6; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK 4/5/6; PAK 4/6/7; PAK-4; PAK-6; PAK1-related kinase; PAK4; PAK4/5/6; protein kinase PAK5; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; serine/threonine-protein kinase; serine/threonine-protein kinase PAK 7; serine/threonine-protein kinase PAK7; unnamed protein product

          View more View less

          Gene Aliases: 2900083L08Rik; 4732456M09; 5730488L07Rik; mKIAA1142; PAK-5; PAK-7; PAK7

          View more View less

          UniProt ID: (Human) O96013, (Human) Q9P286, (Human) Q9NQU5, (Mouse) Q8BTW9, (Rat) D4A280, (Mouse) Q8C015, (Mouse) Q3ULB5

          View more View less

          Entrez Gene ID: (Human) 10298, (Human) 57144, (Human) 56924, (Mouse) 70584, (Rat) 292756, (Rat) 311450, (Mouse) 241656, (Mouse) 214230, (Rat) 296078

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity protein binding ATP binding protein binding, bridging cadherin binding involved in cell-cell adhesion protein serine kinase activity cadherin binding nucleotide binding
          Process(es)
          apoptotic process cytoskeleton organization learning memory locomotory behavior cellular response to starvation intracellular signal transduction regulation of MAPK cascade negative regulation of extrinsic apoptotic signaling pathway negative regulation of triglyceride catabolic process positive regulation of angiogenesis protein stabilization dendritic spine development positive regulation of focal adhesion disassembly negative regulation of endothelial cell apoptotic process neuron projection arborization neuron projection extension regulation of transcription, DNA-templated regulation of cell growth signal transduction cell migration cell-cell adhesion cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline