Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • VANGL2 Antibodies

          Invitrogen

          Phospho-VANGL2 (Ser79, Ser82, Ser84) Recombinant Rabbit Monoclonal Antibody (6O6T6)

          1 Reference
          View all (11) VANGL2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-VANGL2 (Ser79, Ser82, Ser84) Recombinant Rabbit Monoclonal Antibody (6O6T6)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Phospho-VANGL2 (Ser79, Ser82, Ser84) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-VANGL2 (Ser79, Ser82, Ser84) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-VANGL2 (Ser79, Ser82, Ser84) Antibody (MA5-38242) in WB

          Western blot analysis of Phospho-VANGL2 (Ser79, Ser82, Ser84) in extracts of various cell lines. Samples were incubated with Phospho-VANGL2 (Ser79, Ser82, Ser84) Monoclonal antibody (Product # MA5-38242) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) secondary antibody at a dilution of 1:10,000. HeLa and C6 cells were treated by wnt (100 ng/mL) for 30 minutes. Lysates/proteins: 25 µ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-VANGL2 (Ser79, Ser82, Ser84) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-VANGL2 (Ser79, Ser82, Ser84) Recombinant Rabbit Monoclonal Antibody (6O6T6)

          Product Details

          MA5-38242

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 1 publication 1 publication

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          6O6T6

          Immunogen

          Synthetic peptide.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2898158

          Product Specific Information

          Positive Samples: HeLa

          Immunogen sequence: GTSEHSISHD

          Target Information

          Van Gogh is an alternate name for the Drosophila strabismus protein. Van Gogh like-2 (VANGL2) is a 521 amino acidprotein in vertebrates similar to strabismus 1 of Drosophila and a component of the frizzled-disheveled tissue polarity pathway. It interacts through it's C-terminal region with the N-terminal half of DVL1, DVL2 and DVL3 and is a potent tumor suppressor. It ispredominantly involved in the control of early morphogenesis and patterning of both axial midline structures and the development of neural plate. It plays a crucial role in the regulation of planar cell polarity, particularly in the orientation of stereociliary bundles in the cochlea. VANGL2 is required for polarization and movement of myocardializing cells and seems to act via RHOA signaling to regulate this process. It is expressed in a wide range of tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: loop tail associated protein; loop-tail protein 1 homolog; loop-tail-associated protein; MGC119403; MGC119404; strabismus 1; unnamed protein product; van Gogh-like protein 2; vang; vang-like 2 (van gogh, Drosophila)

          View more View less

          Gene Aliases: LPP1; LTAP; STB1; STBM; STBM1

          View more View less

          UniProt ID: (Human) Q9ULK5, (Rat) P84889

          View more View less

          Entrez Gene ID: (Human) 57216, (Rat) 289229

          View more View less

          Function(s)
          protein binding
          Process(es)
          establishment of planar polarity neural tube closure heart looping cell migration involved in kidney development dopaminergic neuron axon guidance serotonergic neuron axon guidance apical protein localization Wnt signaling pathway, planar cell polarity pathway non-motile cilium assembly regulation of synapse pruning hair follicle development membranous septum morphogenesis muscular septum morphogenesis Rho protein signal transduction biological_process anterior/posterior pattern specification heparan sulfate proteoglycan biosynthetic process convergent extension involved in neural plate elongation regulation of Wnt signaling pathway glomerulus development regulation of actin cytoskeleton organization somatic stem cell population maintenance non-canonical Wnt signaling pathway post-anal tail morphogenesis wound healing positive regulation of JUN kinase activity establishment or maintenance of epithelial cell apical/basal polarity somatic stem cell division establishment of body hair planar orientation digestive tract morphogenesis convergent extension involved in axis elongation convergent extension involved in organogenesis inner ear receptor cell development inner ear receptor stereocilium organization orthogonal dichotomous subdivision of terminal units involved in lung branching morphogenesis planar dichotomous subdivision of terminal units involved in lung branching morphogenesis lateral sprouting involved in lung morphogenesis kidney morphogenesis cochlea development cochlea morphogenesis regulation of establishment of planar polarity establishment of planar polarity involved in neural tube closure
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0