Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Fibronectin Antibodies

          Invitrogen

          Fibronectin Recombinant Rabbit Monoclonal Antibody (5I8T1)

          Advanced Verification
          View all (105) Fibronectin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Fibronectin Recombinant Rabbit Monoclonal Antibody (5I8T1)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Advanced Verification (1)
          Fibronectin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Fibronectin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Fibronectin Antibody (MA5-42661) in ICC/IF

          Confocal imaging (Immunocytochemistry) of Fibronectin in HeLa cells. Samples were incubated with Fibronectin Monoclonal antibody (Product # MA5-42661) using a dilution of 1:50 (Red). The cells were counterstained with alpha Tubulin Mouse mAb (dilution 1:400) (Green). DAPI was used for nuclear staining (blue). Objective: 100x. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Fibronectin Antibody in Immunocytochemistry (ICC/IF)
          Fibronectin Antibody in Western Blot (WB)
          Fibronectin Antibody in Western Blot (WB)
          Fibronectin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Fibronectin Recombinant Rabbit Monoclonal Antibody (5I8T1)

          Product Details

          MA5-42661

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:6,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          5I8T1

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.5 mg/mL

          Amount

          150 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2911802

          Product Specific Information

          Positive test controls include: HepG2. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space.

          Immunogen sequence: MLRGPGPGLL LLAVQCLGTA VPSTGASKSK RQAQQMVQPQ SPVAVSQSKP GCYDNGKHYQ INQQWERTYL GNALVCTCYG GSRGFNCESK PEAEETCFDK YTGNTYRVGD TYERPKDSMI WDCTCIGAGR GRISCTIANR CHEGGQSYKI GDTWRRPHET GGYMLECVCL GNGKGEWTCK PIAEKCFDHA AGTSYVVGET WEKPYQGWMM VDCTCLGEGS GRITCTSRNR CNDQDTRTSY RIGDTWSKKD NRGNLLQCIC TGNGRGEWKC ERHTSVQTTS SGSGPFTDVR AAVYQPQPHP QPPPYGHCVT DSGVVYSVGM QWLKTQGNKQ MLCTCLGNGV SCQETAVTQT YGGNSNGEPC VLPFTYNGRT FYSCTTEGRQ DGHLWCSTTS NYEQDQKYSF CTDHTVLVQT RGGNSNGALC HFPFLYNNHN YTDCTSEGRR DNMKWCGTTQ NYDADQKFGF CPMAAHEEIC TTNEGVMYRI GDQWDKQHDM GHMMRCTCVG NGRGEWTCIA YSQLRDQCIV DDITYNVNDT FHKRHEEGHM LNCTCFGQGR GRWKCDPVDQ CQDSETGTFY QIGDSWEKYV HGVRYQCYCY GRGIGEWHCQ PLQTYPSSSG PVEVFITETP SQPNSHPIQW NAPQPSHISK YILRWRPKNS VGRWKEATIP GHLNSYTIKG LKPGVVYEGQ LISIQQYGHQ EVTRFDFTTT STSTPVTSNT VTGETTPFSP LVATSESVTE ITASSFVVSW VSASDTVSGF RVEYELSEEG DEPQYLDLPS TATSVNIPDL LPGRKYIVNV YQISEDGEQS LILSTSQTTA PDAPPDTTVD QVDDTSIVVR WSRPQAPITG YRIVYSPSVE GSSTELNLPE TANSVTLSDL QPGVQYNITI YAVEENQEST PVVIQQETTG TPRSDTVPSP RDLQFVEVTD VKVTIMWTPP ESAVTGYRVD VIPVNLPGEH GQRLPISRNT FAEVTGLSPG VTYYFKVFAV SHGRESKPLT AQQTTKLDAP TNLQFVNETD STVLVRWTPP RAQITGYRLT VGLTRRGQPR QYNVGPSVSK YPLRNLQPAS EYTVSLVAIK GNQESPKATG VFTTLQPGSS IPPYNTEVTE TTIVITWTPA PRIGFKLGVR PSQGGEAPRE VTSDSGSIVV SGLTPGVEYV YTIQVLRDGQ ERDAPIVNKV VTPLSPPTNL HLEANPDTGV LTVSWERSTT PDITGYRITT TPTNGQQGNS LEEVVHADQS SCTFDNLSPG LEYNVSVYTV KDDKESVPIS DTIIPEVPQL TDLSFVDITD SSIGLRWTPL NSSTIIGYRI TVVAAGEGIP IFEDFVDSSV GYYTVTGLEP GIDYDISVIT LINGGESAPT TLTQQTAVPP PTDLRFTNIG PDTMRVTWAP PPSIDLTNFL VRYSPVKNEE DVAELSISPS DNAVVLTNLL PGTEYVVSVS SVYEQHESTP LRGRQKTGLD SPTGIDFSDI TANSFTVHWI APRATITGYR IRHHPEHFSG RPREDRVPHS RNSITLTNLT PGTEYVVSIV ALNGREESPL LIGQQSTVSD VPRDLEVVAA TPTSLLISWD APAVTVRYYR ITYGETGGNS PVQEFTVPGS KSTATISGLK PGVDYTITVY AVTGRGDSPA SSKPISINYR TEIDKPSQMQ VTDVQDNSIS VKWLPSSSPV TGYRVTTTPK NGPGPTKTKT AGPDQTEMTI EGLQPTVEYV VSVYAQNPSG ESQPLVQTAV TNIDRPKGLA FTDVDVDSIK IAWESPQGQV SRYRVTYSSP EDGIHELFPA PDGEEDTAEL QGLRPGSEYT VSVVALHDDM ESQPLIGTQS TAIPAPTDLK FTQVTPTSLS AQWTPPNVQL TGYRVRVTPK EKTGPMKEIN LAPDSSSVVV SGLMVATKYE VSVYALKDTL TSRPAQGVVT TLENVSPPRR ARVTDATETT ITISWRTKTE TITGFQVDAV PANGQTPIQR TIKPDVRSYT ITGLQPGTDY KIYLYTLNDN ARSSPVVIDA STAIDAPSNL RFLATTPNSL LVSWQPPRAR ITGYIIKYEK PGSPPREVVP RPRPGVTEAT ITGLEPGTEY TIYVIALKNN QKSEPLIGRK KTDELPQLVT LPHPNLHGPE ILDVPSTVQK TPFVTHPGYD TGNGIQLPGT SGQQPSVGQQ MIFEEHGFRR TTPPTTATPI RHRPRPYPPN VGEEIQIGHI PREDVDYHLY PHGPGLNPNA STGQEALSQT TISWAPFQDT SEYIISCHPV GTDEEPLQFR VPGTSTSATL TGLTRGATYN VIVEALKDQQ RHKVREEVVT VGNSVNEGLN QPTDDSCFDP YTVSHYAVGD EWERMSESGF KLLCQCLGFG SGHFRCDSSR WCHDNGVNYK IGEKWDRQGE NGQMMSCTCL GNGKGEFKCD PHEATCYDDG KTYHVGEQWQ KEYLGAICSC TCFGGQRGWR CDNCRRPGGE PSPEGTTGQS YNQYSQRYHQ RTNTNVNCPI ECFMPLDVQA DREDSRE

          Target Information

          Fibronectin is a disulfide-bonded dimer with a molecular weight range of 230-250 kDa. In the extracellular matrix of several connective tissues and vessels, fibronectin is present as an insoluble protein that is extensively cross-linked by interchain disulfide bonds forming high molecular mass polymers. Fibronectin is most abundant during embryonic development and tissue remodeling. Fibronectin is also present at high concentrations as a soluble plasma protein. Fibronectin is organized as a linear series of repeating modules which form domains for interaction with fibronectin itself, other matrix components (e.g. collagen and heparin) and receptors on cells (e.g. integrins). Fibronectin is present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. Fibronectin has been implicated in carcinoma development in lung cancer. Further, Fibronectin expression is increased especially in non-small cell lung carcinoma. The adhesion of lung carcinoma cells to Fibronectin enhances tumorgenecity and confers resistance to apoptosis induced by standard chemotherapeutic agents. The gene encoding Fibronectin has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some Fibronectin variants has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cold insoluble globulin (CIG); cold-insoluble globulin; DKFZp686F10164; DKFZp686H0342; DKFZp686I1370; DKFZp686O13149; epididymis secretory sperm binding protein; extracellular matrix protein; ferritin L subunit; ferritin L-chain; fibronectin; lnc-ABCA12-8; Migration stimulating factor (MSF); migration-stimulating factor; unnamed protein product

          View more View less

          Gene Aliases: CIG; E330027I09; ED-B; FINC; FN; Fn-1; FNZ; GFND; GFND2; LETS; MSF; SMDCF

          View more View less

          UniProt ID: (Human) P02751, (Mouse) P11276

          View more View less

          Entrez Gene ID: (Human) 2335, (Mouse) 14268

          View more View less

          Function(s)
          protease binding receptor binding integrin binding extracellular matrix structural constituent protein binding collagen binding heparin binding peptidase activator activity identical protein binding proteoglycan binding receptor agonist activity mercury ion binding
          Process(es)
          neural crest cell migration regulation of protein phosphorylation cell-substrate junction assembly cell adhesion cell-matrix adhesion integrin-mediated signaling pathway nervous system development heart development positive regulation of cell proliferation response to wounding positive regulation of gene expression response to muscle activity integrin activation substrate adhesion-dependent cell spreading endodermal cell differentiation endothelial cell migration positive regulation of fibroblast proliferation enteric nervous system development interaction with symbiont positive regulation of protein kinase B signaling regulation of ERK1 and ERK2 cascade negative regulation of transforming growth factor beta production blood coagulation, fibrin clot formation negative regulation of monocyte activation neural crest cell migration involved in autonomic nervous system development positive regulation of substrate-dependent cell migration, cell attachment to substrate calcium-independent cell-matrix adhesion glial cell migration response to activity extracellular matrix organization positive regulation of cell migration negative regulation of collagen biosynthetic process wound healing negative regulation of apoptotic process positive regulation of axon extension positive regulation of chemotaxis cellular response to mercury ion cellular response to interleukin-1
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline