Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Aurora A/B/C Antibodies

          Invitrogen

          Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Recombinant Rabbit Monoclonal Antibody (RM454)

          View all (8) Aurora A/B/C antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Recombinant Rabbit Monoclonal Antibody (RM454)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Antibody (MA5-44574) in IHC (P)

          Immunohistochemistry analysis of formalin fixed and paraffin embedded human tonsil tissue usingPhospho-Aurora A/B/C (Thr288, Thr232, Thr198) Recombinant Monoclonal antibody (Product # MA5-44574) at a dilution of 1:50. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Recombinant Rabbit Monoclonal Antibody (RM454)

          Product Details

          MA5-44574

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          RM454

          Immunogen

          A phospho-peptide corresponding to Phospho-Aurora A (Thr288)/Aurora B (Thr232)/Aurora C (Thr198). The immunogen range is a synthetic peptide within amino acids 242-342 of human Aurora A in which Thr288 is phosphorylated.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          10 µg/mL

          Amount

          1 µg

          Purification

          Protein A

          Storage buffer

          PBS with 1% BSA, 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2926704

          Target Information

          Serine/threonine-protein kinase component of the chromosomal passenger complex (CPC), a complex that acts as a key regulator of mitosis. The CPC complex has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. Involved in the bipolar attachment of spindle microtubules to kinetochores and is a key regulator for the onset of cytokinesis during mitosis. Required for central/midzone spindle assembly and cleavage furrow formation. Key component of the cytokinesis checkpoint, a process required to delay abscission to prevent both premature resolution of intercellular chromosome bridges and accumulation of DNA damage: phosphorylates CHMP4C, leading to retain abscission-competent VPS4 (VPS4A and/or VPS4B) at the midbody ring until abscission checkpoint signaling is terminated at late cytokinesis (PubMed:22422861, PubMed:24814515). AURKB phosphorylates the CPC complex subunits BIRC5/survivin, CDCA8/borealin and INCENP. Phosphorylation of INCENP leads to increased AURKB activity. Other known AURKB substrates involved in centromeric functions and mitosis are CENPA, DES/desmin, GPAF, KIF2C, NSUN2, RACGAP1, SEPT1, VIM/vimentin, GSG2/Haspin, and histone H3. A positive feedback loop involving GSG2 and AURKB contributes to localization of CPC to centromeres. Phosphorylation of VIM controls vimentin filament segregation in cytokinetic process, whereas histone H3 is phosphorylated at 'Ser-10' and 'Ser-28' during mitosis (H3S10ph and H3S28ph, respectively). A positive feedback between GSG2 and AURKB contributes to CPC localization. AURKB is also required for kinetochore localization of BUB1 and SGOL1. Phosphorylation of p53/TP53 negatively regulates its transcriptional activity. Key regulator of active promoters in resting B- and T-lymphocytes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ARK-3; aurora 2; aurora 3; aurora kinase B-Sv1; aurora kinase B-Sv2; aurora- and Ipl1-like midbody-associated protein 1; aurora- and Ipl1-like midbody-associated protein 1 homolog; aurora-1; aurora-B; aurora-related kinase 2; aurora-related kinase 3; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; aurora/IPL1-related kinase 2; aurora/IPL1-related kinase 3; aurora/IPL1/EG2 protein 2; breast tumor-amplified kinase; epididymis secretory protein Li 90; protein kinase; chromosomal passenger protein; protein phosphatase 1, regulatory subunit 47; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; serine/threonine kinase 13 (aurora/IPL1-like); serine/threonine protein kinase 15; serine/threonine-protein kinase 12; serine/threonine-protein kinase 13; serine/threonine-protein kinase 5; serine/threonine-protein kinase 6; serine/threonine-protein kinase aurora-A; serine/threonine-protein kinase aurora-B; serine/threonine-protein kinase aurora-C; unnamed protein product

          View more View less

          Gene Aliases: AIE2; AIK; AIK2; AIK3; AIM-1; AIM1; ARK-2; ARK1; ARK2; ARK3; AURA; AurB; AurC; aurkb-sv1; aurkb-sv2; aurora-C; BTAK; HEL-S-90; IPL1; PPP1R47; PPP1R48; SPGF5; STK-1; STK12; STK13; STK15; STK5; STK6; STK7

          View more View less

          UniProt ID: (Human) O14965, (Human) Q96GD4, (Human) Q9UQB9

          View more View less

          Entrez Gene ID: (Human) 6790, (Human) 9212, (Human) 6795

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity protein serine/threonine/tyrosine kinase activity protein binding ATP binding protein kinase binding protein heterodimerization activity protein serine kinase activity molecular function activator activity kinase binding histone H1-4S27 kinase activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter mitotic cell cycle mitotic cytokinesis negative regulation of B cell apoptotic process chromatin remodeling spindle organization mitotic spindle organization positive regulation of microtubule depolymerization positive regulation of telomere maintenance regulation of cytokinesis negative regulation of cytokinesis positive regulation of cytokinesis regulation of microtubule-based process protein localization to kinetochore cellular response to UV cleavage furrow formation post-translational protein modification cell cycle G2/M phase transition mitotic cytokinesis checkpoint mitotic spindle midzone assembly regulation of chromosome segregation midbody abscission positive regulation of mitotic sister chromatid segregation positive regulation of cell cycle process positive regulation of mitotic cell cycle spindle assembly checkpoint mitotic spindle assembly negative regulation of protein localization to chromatin repair of mitotic kinetochore microtubule attachment defect negative regulation of cGAS/STING signaling pathway regulation of signal transduction by p53 class mediator positive regulation of mitotic sister chromatid separation positive regulation of attachment of mitotic spindle microtubules to kinetochore positive regulation of mitotic cytokinesis positive regulation of lateral attachment of mitotic spindle microtubules to kinetochore mitotic sister chromatid biorientation G2/M transition of mitotic cell cycle meiotic spindle organization microtubule cytoskeleton organization protein phosphorylation centrosome cycle mitotic centrosome separation response to wounding regulation of G2/M transition of mitotic cell cycle negative regulation of gene expression peptidyl-serine phosphorylation regulation of protein stability negative regulation of protein binding positive regulation of mitotic nuclear division positive regulation of mitotic cell cycle regulation of centrosome cycle protein autophosphorylation meiotic cell cycle cilium disassembly positive regulation of mitochondrial fission liver regeneration attachment of spindle microtubules to kinetochore regulation of cell cycle process spindle assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0