Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • mGluR1/mGluR5 Antibodies

          Invitrogen

          mGluR1/mGluR5 Monoclonal Antibody (N75/33), PerCP

          View all (22) mGluR1/mGluR5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite mGluR1/mGluR5 Monoclonal Antibody (N75/33), PerCP

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          mGluR1/mGluR5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          mGluR1/mGluR5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          mGluR1/mGluR5 Antibody (MA5-45552) in ICC/IF

          Immunocytochemistry/Immunofluorescence analysis using human neuroblastoma cells. Fixation involved 4% Formaldehyde for 15 min at RT. Samples were incubated with mGluR1/5 monoclonal antibody (Product # MA5-45552) at 1:100 for 60 min at RT, followed by Goat Anti-Mouse ATTO 488 at 1:200 for 60 min at RT. Counterstain used was Phalloidin Texas Red F-Actin stain; DAPI (blue) nuclear stain at 1:1,000, 1:5,000 for... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          mGluR1/mGluR5 Antibody in Immunocytochemistry (ICC/IF)
          mGluR1/mGluR5 Antibody in Immunocytochemistry (ICC/IF)
          mGluR1/mGluR5 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          mGluR1/mGluR5 Monoclonal Antibody (N75/33), PerCP

          Product Details

          MA5-45552

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N75/33

          Immunogen

          Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b
          3D Epitope / Immunogen

          Conjugate

          PerCP PerCP PerCP
          • APC 
          • FITC
          • PE 
          • Request custom conjugation

          Excitation/Emission Max

          482/675 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          95.64mM phosphate/2.48mM MES, pH 7.4, with 0.5M EDTA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2932006

          Product Specific Information

          1 µg/mL of MA5-45552 was sufficient for detection of mGluR1/5 glutamate receptor in 20 µg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 130kDa (both mGluR1 and mGluR5).

          This antibody was formerly sold as clone S75-33.

          Target Information

          may play a role in synaptic transmission; may interact with mGluR1, another group I metabotropic glutamate receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: G protein coupled receptor family C group 1 member A; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member E; glutamate receptor; glutamate receptor, metabotropic 1; glutamate receptor, metabotropic 5; GPCR family C; metabotropic glutamate receptor (mGluR5); metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5 homolog; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; mGluR1; mGluR5; mGluR5f; mGluR5g; mGluR5h; mGluR5i; mGlurR1a; protein phosphatase 1, regulatory subunit 85; protein phosphatase 1, regulatory subunit 86; unnamed protein product; variant b of mGluR5

          View more View less

          Gene Aliases: 4930455H15Rik; 6430542K11Rik; Glu5R; Gm10828; GPRC1A; GPRC1E; MGLU1; mGlu5; MGLUR1; MGLUR5; mGluR5b; nmf373; PPP1R85; PPP1R86; rcw; SCA44; SCAR13; wobl

          View more View less

          UniProt ID: (Human) Q13255, (Human) P41594, (Rat) P23385, (Mouse) P97772, (Rat) P31424, (Mouse) Q3UVX5

          View more View less

          Entrez Gene ID: (Human) 2911, (Human) 2915, (Rat) 24414, (Mouse) 14816, (Rat) 24418, (Mouse) 108071

          View more View less

          Function(s)
          adenylate cyclase inhibiting G-protein coupled glutamate receptor activity G-protein coupled receptor activity protein binding glutamate receptor activity G protein-coupled neurotransmitter receptor activity G protein-coupled receptor activity involved in regulation of postsynaptic membrane potential protein tyrosine kinase activator activity A2A adenosine receptor binding identical protein binding protein tyrosine kinase binding PLC activating G-protein coupled glutamate receptor activity fibronectin binding collagen binding estrogen receptor binding laminin binding protein sequestering activity calmodulin binding PDZ domain binding
          Process(es)
          G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled glutamate receptor signaling pathway G-protein coupled glutamate receptor signaling pathway chemical synaptic transmission locomotory behavior sensory perception of pain regulation of membrane potential positive regulation of MAPK cascade regulation of sensory perception of pain regulation of synaptic transmission, glutamatergic regulation of postsynaptic membrane potential synaptic signaling via neuropeptide temperature homeostasis response to amphetamine desensitization of G-protein coupled receptor protein signaling pathway regulation of transcription, DNA-templated positive regulation of cytosolic calcium ion concentration learning or memory associative learning positive regulation of dopamine secretion operant conditioning protein targeting to nuclear inner membrane response to morphine response to ethanol response to antibiotic positive regulation of long-term neuronal synaptic plasticity synapse organization positive regulation of calcium-mediated signaling cognition response to corticosterone negative regulation of dendritic spine morphogenesis sensory perception of hot stimulus negative regulation of excitatory postsynaptic potential positive regulation of cellular response to hypoxia regulation of long term synaptic depression positive regulation of intracellular signal transduction positive regulation of sensory perception of pain cellular response to beta-amyloid conditioned place preference positive regulation of neural precursor cell proliferation protein phosphorylation learning phosphatidylinositol-mediated signaling regulation of long-term neuronal synaptic plasticity modulation of synaptic transmission regulation of glutamate secretion calcium-mediated signaling positive regulation of cell migration negative regulation of viral entry into host cell receptor-mediated virion attachment to host cell membrane depolarization synaptic transmission, GABAergic cellular response to electrical stimulus L-glutamate import across plasma membrane negative regulation of protein localization to plasma membrane
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nbmw6:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline