Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Actin Antibodies

          Invitrogen

          Actin Monoclonal Antibody (5J11)

          View all (273) Actin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Actin Monoclonal Antibody (5J11)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Actin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Actin Antibody (MA5-47355) in ICC/IF

          Immunofluorescent analysis of Actin in HeLa cells. Samples were stained with Actin monoclonal antibody (Product # MA5-47454) in green. Costained with chicken pAb against vimentin in red. The actin antibody labels the submembranous actin-rich cytoskeleton, stress fibers and bundles of actin associated with cell adhesion sites. The vimentin antibody stains a different cytoskeletal network, the intermediate or 10 nm filaments. The blue is DAPI staining of nuclear DNA. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Actin Antibody in Immunocytochemistry (ICC/IF)
          Actin Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Actin Monoclonal Antibody (5J11)

          Product Details

          MA5-47355

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Bovine, Horse, Human, Mouse, Pig, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          5J11

          Immunogen

          Actin preparation from bovine brain
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          500 µg

          Purification

          Protein G

          Storage buffer

          PBS with 50% glycerol

          Contains

          5mM sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942337

          Target Information

          Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: A-X actin; A-X actin-like protein; ACTA1; ACTA2; ACTB; ACTC; ACTG2; actin; actin alpha 1; actin alpha cardiac; Actin is a protein with a high degree of evolutionary conservation that undergoes polymerization to generate filaments, which subsequently assemble into interconnected networks within the cytoplasm of cells.; actin, alpha 2, smooth muscle, aorta; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; actin, beta, cytoplasmic; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin-like protein; alfa sma; alpha actin 1; alpha actin 2; alpha smooth muscle actin; alpha-actin; alpha-actin cardiac; alpha-actin-1; alpha-actin-2; alpha-actin-3; alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; aSMA; aSmooth Muscle Actin; Atcb; B-actin; beta actin; beta cytoskeletal actin; beta-actin; beta-actin FE-3; cardiac alpha actin; cardiac muscle alpha actin 1; cell growth-inhibiting gene 46 protein; cytoplasmic beta-actin; cytoplasmic protein; cytoskeletal beta actin; cytoskeletal gamma-actin; epididymis luminal protein 176; gamma-2-actin; gamma-actin; gamma-enteric smooth muscle actin; GIG46; I(2)-actin; melanoma X-actin; nemaline myopathy type 3; pan actin; PS1TP5-binding protein 1; skeletal alpha actin; skeletal muscle alpha-actin; skeletal muscle alpha-actin 1; smooth muscle alpha-actin; smooth muscle gamma-actin; unnamed protein product; vascular smooth muscle alpha-actin

          View more View less

          Gene Aliases: 0610041G09Rik; a-SMA; ACT; ACT-4; Act4; ACTA; Acta-2; ACTA3; ACTC; Actc-1; ACTE; ACTG; ACTGE; actin; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; Actx; alphaSMA; ASD5; ASMA; beta-actin; BKRNS; BNS; BRWS1; CFTD; CFTD1; CFTDM; CMD1R; CMH11; CMYO2A; CMYO2B; CMYO2C; CMYP2A; CMYP2B; CMYP2C; CSMH; DDS1; DFNA20; DFNA26; E430023M04Rik; E51; HEL-176; LVNC4; MMIHS5; MPFD; NEM1; NEM2; NEM3; PS1TP5BP1; SHPM; SMAalpha; SMalphaA; SMDYS; SMGA; THC8; VSCM; VSCM1

          View more View less

          UniProt ID: (Human) P68133, (Bovine) P68138, (Pig) P68137, (Bovine) P62739, (Human) P62736, (Horse) P60708, (Bovine) P60712, (Pig) Q6QAQ1, (Human) P60709, (Bovine) Q3ZC07, (Human) P68032, (Human) P63261, (Bovine) P63258, (Bovine) Q5E9B5, (Human) P63267, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737, (Rat) P60711, (Mouse) P60710, (Rat) P68035, (Mouse) P68033, (Mouse) P63260, (Rat) P63259, (Rat) P63269, (Mouse) P63268

          View more View less

          Entrez Gene ID: (Horse) 100065454, (Human) 58, (Bovine) 281592, (Pig) 100154254, (Pig) 733615, (Bovine) 515610, (Horse) 100062175, (Human) 59, (Horse) 100033878, (Bovine) 280979, (Pig) 414396, (Human) 60, (Bovine) 533219, (Horse) 100057678, (Human) 70, (Pig) 100152267, (Human) 71, (Bovine) 404122, (Pig) 397653, (Horse) 100050139, (Bovine) 281595, (Pig) 100520667, (Human) 72, (Horse) 100057561, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Rat) 81822, (Mouse) 11461, (Rat) 29275, (Mouse) 11464, (Mouse) 11465, (Rat) 287876, (Rat) 25365, (Mouse) 11468

          View more View less

          Function(s)
          structural constituent of cytoskeleton protein binding ATP binding ATPase activity myosin binding ADP binding protein kinase binding kinesin binding Tat protein binding identical protein binding nitric-oxide synthase binding nitric-oxide synthase regulator activity nucleosomal DNA binding tau protein binding structural constituent of postsynaptic actin cytoskeleton transporter regulator activity microfilament motor activity profilin binding ubiquitin protein ligase binding actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          muscle contraction positive regulation of gene expression skeletal muscle thin filament assembly skeletal muscle fiber development mesenchyme migration regulation of blood pressure response to virus vascular smooth muscle contraction glomerular mesangial cell development cytoskeleton organization positive regulation of hepatic stellate cell migration positive regulation of hepatic stellate cell contraction positive regulation of ERK1 and ERK2 cascade cellular response to transforming growth factor beta stimulus juxtaglomerular apparatus development positive regulation of hepatic stellate cell activation morphogenesis of a polarized epithelium establishment or maintenance of cell polarity regulation of transmembrane transporter activity negative regulation of protein binding adherens junction assembly apical protein localization synaptic vesicle endocytosis regulation of norepinephrine uptake protein localization to adherens junction cellular response to cytochalasin B postsynaptic actin cytoskeleton organization regulation of protein localization to plasma membrane chromatin remodeling regulation of transcription from RNA polymerase II promoter microtubule nucleation axonogenesis positive regulation of cell proliferation substantia nigra development regulation of mitotic metaphase/anaphase transition maintenance of blood-brain barrier regulation of apoptotic process positive regulation of T cell differentiation negative regulation of cell differentiation positive regulation of cell differentiation positive regulation of myoblast differentiation positive regulation of transcription, DNA-templated cell motility positive regulation of norepinephrine uptake regulation of cell cycle regulation of G0 to G1 transition platelet aggregation regulation of transepithelial transport regulation of synaptic vesicle endocytosis positive regulation of stem cell population maintenance positive regulation of double-strand break repair via homologous recombination regulation of G1/S transition of mitotic cell cycle regulation of double-strand break repair positive regulation of double-strand break repair regulation of nucleotide-excision repair actin filament organization actin filament-based movement actomyosin structure organization actin-myosin filament sliding cardiac myofibril assembly cardiac muscle tissue morphogenesis heart contraction angiogenesis positive regulation of cell migration regulation of stress fiber assembly regulation of focal adhesion assembly positive regulation of wound healing tight junction assembly protein localization to bicellular tight junction response to ethanol cellular response to interleukin-6 cellular response to acetaldehyde response to mechanical stimulus cellular response to potassium ion skeletal muscle fiber adaptation response to steroid hormone circadian rhythm dynein-driven meiotic oscillatory nuclear movement response to immobilization stress retina development in camera-type eye cellular response to electrical stimulus response to xenobiotic stimulus negative regulation of apoptotic process cardiac muscle contraction cytoplasmic actin-based contraction involved in cell motility sarcomere organization cellular response to interferon-gamma response to calcium ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-m8c75:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0