Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RAB3a Antibodies
          Validated with Transparency. Independently tested through the YCharOS Open Science Initiative.

          Invitrogen

          RAB3A Recombinant Rabbit Monoclonal Antibody (PSH01-08)

          Advanced Verification
          View all (28) RAB3a antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RAB3A Recombinant Rabbit Monoclonal Antibody (PSH01-08)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (20)
          • Advanced Verification (8)
          RAB3A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RAB3A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 28

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAB3A Antibody (MA5-49607) in ICC/IF

          Immunofluorescence of RAB3A was performed using SK-N-FI WT and RAB3A KO cells that were labeled with a green or a far-red fluorescent dye, respectively. Post-labeling, WT and KO cells were mixed and plated to a 1:1 ratio in a 96-well plate with an optically clear flat bottom as a mosaic and incubated for 24 hrs. Cells were fixed in 4% PFA (in PBS) for 15 min at RT; cells were permeabilized with 0.1% Triton X-100 for 10 min at RT and blocked with PBS containing 5% BSA, 5% goat serum, and 0.01% Triton X-100 for 30 min at RT. Cells were stain... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAB3A Antibody in Immunocytochemistry (ICC/IF)
          RAB3A Antibody in Immunocytochemistry (ICC/IF)
          RAB3A Antibody in Immunocytochemistry (ICC/IF)
          RAB3A Antibody in Immunocytochemistry (ICC/IF)
          RAB3A Antibody in Immunocytochemistry (ICC/IF)
          RAB3A Antibody in Immunocytochemistry (ICC/IF)
          RAB3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB3A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB3A Antibody in Western Blot (WB)
          RAB3A Antibody in Western Blot (WB)
          RAB3A Antibody in Western Blot (WB)
          RAB3A Antibody in Western Blot (WB)
          RAB3A Antibody in Western Blot (WB)
          RAB3A Antibody in Western Blot (WB)
          RAB3A Antibody in Western Blot (WB)
          RAB3A Antibody in Flow Cytometry (Flow)
          RAB3A Antibody in Flow Cytometry (Flow)
          RAB3A Antibody
          RAB3A Antibody
          RAB3A Antibody
          RAB3A Antibody
          RAB3A Antibody
          RAB3A Antibody
          RAB3A Antibody
          RAB3A Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          RAB3A Recombinant Rabbit Monoclonal Antibody (PSH01-08)

          Product Details

          MA5-49607

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Flow Cytometry (Flow)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          Expi293

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PSH01-08

          Immunogen

          Synthetic peptide within human RAB3a aa 121-170.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 0.1% BSA, 40% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3094127

          Product Specific Information

          Sequence Similarities: 100% Mouse/Rat.

          Tissue Specificity: Specifically expressed in brain.

          Positive Control: Human brain tissue lysate, mouse brain tissue lysate, rat brain tissue lysate, human cerebellum tissue, mouse cerebellum tissue, rat cerebellum tissue, SH-SY5Y, N2A, PC-12.

          Subcellular Location: Cytoplasm, cytosol, Lysosome, Cytoplasmic vesicle, secretory vesicle, Cell projection, axon, Cell membrane, Presynapse, Postsynapse.

          Predicted band size: 25 kDa

          Target Information

          Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: C18A3.6; RAS-associated protein RAB3A; Ras-related small GTP binding protein 3A; unnamed protein product

          View more View less

          Gene Aliases: RAB3; SCA52

          View more View less

          UniProt ID: (Human) P20336, (Rat) P63012, (Mouse) P63011

          View more View less

          Entrez Gene ID: (Human) 5864, (Rat) 25531, (Mouse) 19339

          View more View less

          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding protein binding, bridging GTP-dependent protein binding myosin V binding ATPase activator activity GDP-dissociation inhibitor binding ATPase binding
          Process(es)
          plasma membrane repair respiratory system process exocytosis mitochondrion organization neuromuscular synaptic transmission axonogenesis post-embryonic development regulation of synaptic vesicle priming regulation of dopamine secretion synaptic vesicle exocytosis synaptic vesicle maturation calcium ion regulated exocytosis regulation of exocytosis insulin secretion lung development regulation of synaptic vesicle fusion to presynaptic membrane lysosome localization Rab protein signal transduction synaptic vesicle recycling regulated exocytosis regulation of short-term neuronal synaptic plasticity synaptic vesicle transport maintenance of presynaptic active zone structure sensory perception of touch response to electrical stimulus acrosomal vesicle exocytosis evoked neurotransmitter secretion synaptic vesicle clustering regulation of presynaptic dense core granule exocytosis positive regulation of regulated secretory pathway regulation of plasma membrane repair regulation of synaptic vesicle exocytosis constitutive secretory pathway positive regulation of exocytosis regulation of long-term synaptic potentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline