Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GNAI1/GNAI2/GNAI3 Antibodies

          Invitrogen

          GNAI1/GNAI2/GNAI3 Recombinant Rabbit Monoclonal Antibody (HL2093)

          View all (2) GNAI1/GNAI2/GNAI3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite GNAI1/GNAI2/GNAI3 Recombinant Rabbit Monoclonal Antibody (HL2093)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GNAI1/GNAI2/GNAI3 Antibody (MA5-50379) in WB

          Western Blot analysis of GNAI1/GNAI2/GNAI3 using GNAI1/GNAI2/GNAI3 Recombinant Monoclonal Antibody (Product # MA5-50379) in A431 (30 μg) and HeLa (30 μg) whole cell extracts at a dilution of 1:1,000. Extracts were separated by 10% SDS-PAGE. An HRP-conjugated anti-rabbit IgG antibody was used to detect the primary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA5-50379

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1000-10000
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          HL2093

          Immunogen

          Recombinant fragment of human GNAI3.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3094417

          Target Information

          GNAI1/GNAI2/GNAI3 are genes that encode for the G protein alpha inhibiting activity subunit. These proteins play a crucial role in signal transduction pathways, particularly in the inhibition of adenylyl cyclase. They are involved in various physiological processes, including neurotransmitter release, hormone secretion, and regulation of smooth muscle contraction. Mutations in these genes have been associated with various diseases, including hypoparathyroidism, pituitary adenoma, and cardiovascular disorders.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: adenylate cyclase-inhibiting G alpha protein; G alpha inhibitory type 2 (a); G alpha inhibitory type 2 (b); G protein; G protein alpha subunit; g(i) alpha-3; Galpha i1a; Galpha i3; Gi1 protein alpha subunit; Gi3 protein alpha subunit; GNAI2; GTP-binding protein; GTP-binding protein (G-alpha-i2); GTP-binding protein alpha subunit; GTP-binding regulatory protein Gi alpha-2 chain; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein (G protein), alpha inhibiting 2; guanine nucleotide binding protein (G protein), alpha inhibiting 3; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3; guanine nucleotide binding protein, alpha inhibiting 1; guanine nucleotide binding protein, alpha inhibiting 2; guanine nucleotide binding protein, alpha inhibiting 3; Guanine nucleotide binding, protein, alpha inhibiting polypeptide 3; guanine nucleotide-binding protein (G) alpha subunit; Guanine nucleotide-binding protein alpha subunit; guanine nucleotide-binding protein alpha-i subunit; guanine nucleotide-binding protein G(i) subunit alpha; guanine nucleotide-binding protein G(i), alpha-1 subunit; guanine nucleotide-binding protein G(i), alpha-2 subunit; guanine nucleotide-binding protein G(k) subunit alpha; guanine nucleotide-binding regulatory protein alpha-inhibitory subunit; heterotrimeric guanine nucleotide-binding protein 1A; heterotrimeric guanine nucleotide-binding protein 1B; heterotrimeric guanine nucleotide-binding protein 1C; HG1B/HG1C/HG1A; inhibitory G protein of adenylate cyclase, alpha chain; sGi2; unnamed protein product

          View more View less

          Gene Aliases: 87U6; ARCND1; ARCODS; BPGTPB; Galphai2; Galphai3; Gi; Gia; Gialpha1; GIP; GIP3A; Gnai-1; Gnai-2; Gnai-3; GNAI2B; H_LUCA15.1; H_LUCA16.1; HG1A; HG1B; HG1C; NEDHISB; zgc:63957

          View more View less

          UniProt ID: (Human) P63096, (Human) P04899, (Human) P08754, (Mouse) B2RSH2, (Rat) P10824, (Rat) P04897, (Mouse) P08752, (Mouse) Q9DC51, (Rat) P08753

          View more View less

          Entrez Gene ID: (Human) 2770, (Human) 2771, (Human) 2773, (Mouse) 14677, (Rat) 25686, (Rat) 81664, (Mouse) 14678, (Mouse) 14679, (Rat) 25643, (Zebrafish) 393946, (Zebrafish) 100006888

          View more View less

          Function(s)
          magnesium ion binding G-protein coupled receptor binding GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding adenylate cyclase inhibitor activity guanyl nucleotide binding GDP binding G-protein beta/gamma-subunit complex binding D2 dopamine receptor binding G-protein coupled serotonin receptor binding adenylate cyclase regulator activity GTPase activating protein binding protein domain specific binding nucleotide binding metal ion binding heterotrimeric G-protein
          Process(es)
          establishment of mitotic spindle orientation signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting dopamine receptor signaling pathway adenylate cyclase-inhibiting serotonin receptor signaling pathway neuropeptide signaling pathway regulation of G-protein coupled receptor protein signaling pathway response to prostaglandin E mast cell degranulation response to peptide hormone positive regulation of cholesterol biosynthetic process cell division regulation of mitotic spindle organization chemokine-mediated signaling pathway T cell migration positive regulation of relaxation of smooth muscle regulation of eating behavior cellular response to forskolin positive regulation of protein localization to cell cortex angiogenesis adenosine receptor signaling pathway negative regulation of adenylate cyclase activity gamma-aminobutyric acid signaling pathway response to nutrient positive regulation of cell proliferation positive regulation of cell migration positive regulation of superoxide anion generation positive regulation of urine volume negative regulation of calcium ion-dependent exocytosis modulation of synaptic transmission negative regulation of synaptic transmission regulation of calcium ion transport positive regulation of ERK1 and ERK2 cascade negative regulation of adrenergic receptor signaling pathway positive regulation of vascular smooth muscle cell proliferation positive regulation of neural precursor cell proliferation negative regulation of apoptotic signaling pathway positive regulation of macroautophagy GTP metabolic process neurotransmitter receptor localization to postsynaptic specialization membrane G-protein coupled acetylcholine receptor signaling pathway G-protein coupled glutamate receptor signaling pathway cell proliferation positive regulation of NAD(P)H oxidase activity vesicle fusion negative regulation of secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline