Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • beta Tubulin Antibodies

          Invitrogen

          beta Tubulin Monoclonal Antibody (C3)

          View all (89) beta Tubulin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite beta Tubulin Monoclonal Antibody (C3)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (18)
          beta Tubulin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          beta Tubulin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          beta Tubulin Antibody (MA5-50839) in ICC/IF

          Immunocytochemistry analysis of beta Tubulin in Human Hela cells using beta Tubulin Monoclonal Antibody (C3) (Product # MA5-50839) at a dilution of 30 µg/mL, followed by secondary antibody staining with 2 µg/mL FITC-Linked Caprine Anti-Mouse IgG Polyclonal Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          beta Tubulin Antibody in Immunocytochemistry (ICC/IF)
          beta Tubulin Antibody in Immunocytochemistry (ICC/IF)
          beta Tubulin Antibody in Immunocytochemistry (ICC/IF)
          beta Tubulin Antibody in Immunocytochemistry (ICC/IF)
          beta Tubulin Antibody in Immunocytochemistry (ICC/IF)
          beta Tubulin Antibody in Immunocytochemistry (ICC/IF)
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Tubulin Antibody in Western Blot (WB)
          beta Tubulin Antibody in Western Blot (WB)
          beta Tubulin Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          beta Tubulin Monoclonal Antibody (C3)

          Product Details

          MA5-50839

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-2 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          5-20 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          5-40 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Chicken, Guinea pig, Goat, Human, Sheep, Zebrafish

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          C3

          Immunogen

          Recombinant Human TUBb protein, Val170-Val419 (Accession #P07437)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          200 µg

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.05% Proclin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3094428

          Target Information

          Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Beta-III tubulin is primarily expressed in neurons and may be involved in neurogenesis, axon guidance and maintenance. Mutations in the beta tubulin gene are the cause of congenital fibrosis of the extraocular muscles type 3. Beta-III tubulin was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues. Glutamate residues at the C-terminus of beta III tubulin can be glutamylated. The precise function of such modifications is unclear. Tubulin is phosphorylated on Ser-172 by CDK1 during cell cycle progression. Ser-172 phosphorylation inhibits tubulin incorporation into microtubules.Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55 kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton. Because beta-tubulin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: b-Tubulin; beta 5-tubulin; beta Ib tubulin; beta tubulin 1; beta-4 tubulin; beta-4-tubulin; beta-5 tubulin; beta-tubulin; Beta-tubulin class-I; beta-tubulin class-II; beta-tubulin class-III; beta-tubulin class-V; beta-tubulin class-VI; c(beta)7 tubulin; class I beta-tubulin; class II beta-tubulin isotype; class IIa beta-tubulin; class IIb beta-tubulin; class III beta-tubulin; class IVa beta-tubulin; class V beta-tubulin; dJ40E16.7; dystonia 4, torsion (autosomal dominant); epididymis secretory sperm binding protein; HSA10p15 beta-tubulin 4Q; HSA18p11 beta-tubulin 4Q pseudogene; M40 antibody; MGC8685; microtubule protein; similar to beta tubulin; TUBB; tubulin beta; tubulin beta 4'; tubulin beta 8 pseudogene 12; tubulin beta class V; tubulin beta MGC4083; Tubulin beta-1 chain; tubulin beta-2B chain; tubulin beta-4 chain; tubulin beta-5 chain; Tubulin beta-8 chain-like protein LOC260334; tubulin beta-III; tubulin, beta 1; tubulin, beta 2A; tubulin, beta 2A class IIa; tubulin, beta 2B; tubulin, beta 3; tubulin, beta 4; tubulin, beta 6; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta polypeptide paralog; tubulin, beta, 3; tubulin, beta, 4; tubulin, beta, 5; unnamed protein product; XTP3TPATP1; zbetaI

          View more View less

          Gene Aliases: bA506K6.1; bA631M21.2; beta-4; beta-5; CDCBM; CDCBM1; CDCBM5; CDCBM6; CDCBM7; CFEOM3; CFEOM3A; CSCSC1; DYT4; fb40f09; FEOM3; FPVEPD; HsT1601; M40; MACTHC1; OK/SW-cl.56; OOMD; OOMD2; OZEMA2; PMGYSA; si:ch211-122i22.4; si:ch211-9a18.2; TUBB; TUBB-5; TUBB1; TUBB2; TUBB4; TUBB5; TUBB8P12; wu:fb40f09; zgc:112515; zgc:123194

          View more View less

          UniProt ID: (Chicken) P09244, (Human) P07437, (Bovine) Q2KJD0, (Chicken) P09207, (Human) Q9H4B7, (Human) Q13885, (Human) Q9BVA1, (Bovine) Q6B856, (Chicken) P32882, (Human) Q13509, (Bovine) Q2T9S0, (Bovine) Q3ZBU7, (Human) P04350, (Chicken) P09653, (Human) Q9BUF5, (Bovine) Q2HJ81, (Human) Q3ZCM7, (Human) A6NNZ2

          View more View less

          Entrez Gene ID: (Goat) 102168953, (Chicken) 396254, (Human) 203068, (Dog) 474830, (Bovine) 615087, (Sheep) 100303606, (Chicken) 396427, (Sheep) 101115144, (Bovine) 541271, (Goat) 102188330, (Human) 81027, (Dog) 485950, (Human) 7280, (Chicken) 768337, (Goat) 108633269, (Human) 347733, (Dog) 478702, (Bovine) 281555, (Chicken) 420883, (Chicken) 431043, (Dog) 102154194, (Human) 10381, (Bovine) 768070, (Dog) 100855815, (Bovine) 540236, (Sheep) 105609865, (Human) 10382, (Goat) 102178062, (Sheep) 101106224, (Chicken) 421037, (Human) 84617, (Bovine) 534206, (Dog) 480213, (Goat) 102178409, (Human) 347688, (Human) 260334, (Guinea pig) 100718713, (Guinea pig) 100713236, (Guinea pig) 100731652, (Guinea pig) 100720651, (Guinea pig) 100733481, (Zebrafish) 558552, (Zebrafish) 641421, (Zebrafish) 386701, (Zebrafish) 550458

          View more View less

          Function(s)
          GTPase activity structural constituent of cytoskeleton GTP binding ubiquitin protein ligase binding MHC class I protein binding structural molecule activity protein binding protein domain specific binding GTPase activating protein binding macromolecular complex binding protein heterodimerization activity calcium ion binding peptide binding netrin receptor binding nucleotide binding tubulin
          Process(es)
          microtubule cytoskeleton organization mitotic cell cycle microtubule-based process regulation of synapse organization spindle assembly cytoskeleton organization cytoskeleton-dependent intracellular transport natural killer cell mediated cytotoxicity cell division odontoblast differentiation platelet formation thyroid gland development microtubule polymerization thyroid hormone transport platelet aggregation neuron migration cerebral cortex development positive regulation of axon guidance negative regulation of microtubule polymerization axon guidance neuron differentiation netrin-activated signaling pathway dorsal root ganglion development oocyte maturation spindle assembly involved in female meiosis fin regeneration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0