Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TNF alpha Antibodies

          Invitrogen

          TNF alpha (Humicade Biosimilar) Recombinant Mouse Monoclonal Antibody (CDP 571 (Humicade))

          View all (255) TNF alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TNF alpha (Humicade Biosimilar) Recombinant Mouse Monoclonal Antibody (CDP 571 (Humicade))

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          MA5-51587

          Applications
          Tested Dilution
          Publications

          ELISA (ELISA)

          Assay-dependent
          -

          Neutralization (Neu)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          CDP 571 (Humicade)

          Immunogen

          Human TNF-alpha.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          200 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.02% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3250288

          Product Specific Information

          The antibody binds to human TNF-alpha with a Kd of 0.087 nM.

          MA5-51587 is a chimeric antibody. The variable regions from one species have been grafted to the backbone of the other species.

          Target Information

          TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: APC1 protein; cell apoptosis-related protein; DADB-70P7.1; TNF alpha; TNF superfamily; TNF superfamily member 2; TNF α; TNF, macrophage-derived; TNF, monocyte-derived; TNF-a; TNFα; tumor necrosis factor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; tumor necrosis factor-alpha; tumor necrotic factor alpha; unnamed protein product

          View more View less

          Gene Aliases: DIF; IMD127; TNF-alpha; TNFA; TNFSF2; TNLG1F

          View more View less

          UniProt ID: (Human) P01375

          View more View less

          Entrez Gene ID: (Human) 7124

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding protease binding cytokine activity tumor necrosis factor receptor binding protein binding identical protein binding receptor agonist activity histone H3K9ac reader activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter response to hypoxia microglial cell activation positive regulation of cytokine production negative regulation of endothelial cell proliferation response to amphetamine negative regulation of L-glutamate transport macrophage activation involved in immune response chronic inflammatory response to antigenic stimulus positive regulation of immunoglobulin production regulation of production of molecular mediator of immune response negative regulation of cytokine production involved in immune response skeletal muscle contraction negative regulation of systemic arterial blood pressure chromatin organization inflammatory response immune response JAK-STAT cascade circadian rhythm negative regulation of cell proliferation extrinsic apoptotic signaling pathway via death domain receptors intrinsic apoptotic signaling pathway in response to DNA damage response to xenobiotic stimulus response to mechanical stimulus response to virus response to bacterium response to salt stress response to fructose negative regulation of heart rate vascular endothelial growth factor production positive regulation of gene expression negative regulation of gene expression positive regulation of macrophage derived foam cell differentiation negative regulation of lipid storage response to activity calcium-mediated signaling positive regulation of protein complex assembly positive regulation of defense response positive regulation of fever generation negative regulation of myelination lipopolysaccharide-mediated signaling pathway response to nutrient levels response to lipopolysaccharide negative regulation of interleukin-6 production positive regulation of chemokine production positive regulation of fractalkine production positive regulation of interleukin-1 beta production positive regulation of interleukin-18 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production tumor necrosis factor-mediated signaling pathway positive regulation of heterotypic cell-cell adhesion toll-like receptor 3 signaling pathway response to isolation stress response to macrophage colony-stimulating factor regulation of cell proliferation vasodilation positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of programmed cell death regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of protein complex disassembly positive regulation of MAPK cascade positive regulation of neuron apoptotic process negative regulation of blood vessel endothelial cell migration negative regulation of viral genome replication innate immune response positive regulation of nitric oxide biosynthetic process response to ethanol regulation of fat cell differentiation negative regulation of fat cell differentiation negative regulation of osteoblast differentiation positive regulation of osteoclast differentiation positive regulation of protein catabolic process positive regulation of action potential positive regulation of cell adhesion positive regulation of mitotic nuclear division negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated negative regulation of mitotic cell cycle positive regulation of transcription from RNA polymerase II promoter negative regulation of glucose import positive regulation of JNK cascade response to antibiotic astrocyte activation embryonic digestive tract development positive regulation of smooth muscle cell proliferation regulation of epithelial cell proliferation positive regulation of inflammatory response negative regulation of neurogenesis regulation of developmental process regulation of insulin secretion modulation of synaptic transmission positive regulation of synaptic transmission regulation of synapse organization cognition leukocyte tethering or rolling detection of mechanical stimulus involved in sensory perception of pain negative regulation of lipid catabolic process positive regulation of membrane protein ectodomain proteolysis regulation of secretion positive regulation of protein transport regulation of multicellular organismal process response to glucocorticoid positive regulation of hair follicle development positive regulation of protein kinase B signaling regulation of synaptic transmission, glutamatergic positive regulation of glial cell proliferation positive regulation of vitamin D biosynthetic process negative regulation of vascular wound healing negative regulation of branching involved in lung morphogenesis positive regulation of ERK1 and ERK2 cascade positive regulation of calcineurin-NFAT signaling cascade cellular response to molecule of bacterial origin cellular response to lipopolysaccharide cellular response to retinoic acid cellular response to nicotine cellular response to interferon-gamma cellular response to ionizing radiation positive regulation of mononuclear cell migration positive regulation of podosome assembly endothelial cell apoptotic process protein localization to plasma membrane negative regulation of oxidative phosphorylation inflammatory response to wounding extrinsic apoptotic signaling pathway cellular response to toxic substance liver regeneration necroptotic signaling pathway negative regulation of bile acid secretion antiviral innate immune response response to Gram-negative bacterium positive regulation of neuroinflammatory response positive regulation of interleukin-33 production positive regulation of cytokine production involved in inflammatory response negative regulation of beta-amyloid clearance positive regulation of NIK/NF-kappaB signaling positive regulation of synoviocyte proliferation positive regulation of beta-amyloid formation response to L-glutamate positive regulation of neutrophil activation negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of protein localization to plasma membrane regulation of establishment of endothelial barrier negative regulation of bicellular tight junction assembly positive regulation of cellular protein localization response to resveratrol cellular response to beta-amyloid positive regulation of vascular smooth muscle cell proliferation response to hydrogen sulfide positive regulation of leukocyte adhesion to vascular endothelial cell positive regulation of leukocyte adhesion to arterial endothelial cell response to quercetin response to 3,3',5-triiodo-L-thyronine response to gold nanoparticle positive regulation of protein localization to cell surface positive regulation of blood microparticle formation positive regulation of chemokine (C-X-C motif) ligand 2 production positive regulation of hepatocyte proliferation regulation of endothelial cell apoptotic process regulation of reactive oxygen species metabolic process positive regulation of DNA biosynthetic process negative regulation of apoptotic signaling pathway positive regulation of extrinsic apoptotic signaling pathway negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0