Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cannabinoid Receptor 1 Antibodies

          Invitrogen

          Cannabinoid Receptor 1 Recombinant Rabbit Monoclonal Antibody (PSH02-43)

          Advanced Verification
          View all (31) Cannabinoid Receptor 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cannabinoid Receptor 1 Recombinant Rabbit Monoclonal Antibody (PSH02-43)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          • Advanced Verification (3)
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cannabinoid Receptor 1 Antibody (MA5-52083) in IHC (P)

          Immunohistochemical analysis of Cannabinoid Receptor 1 was performed using formalin-fixed paraffin-embedded mouse brain and liver tissue sections. To expose the target protein, heat-induced epitope retrieval was performed on de-paraffinized sections using eBioscience™ IHC Antigen Retrieval Solution - High pH (10X) (Product # 00-4956-58) diluted to 1X solution in water in a decloaking chamber at 110 degree C... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cannabinoid Receptor 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cannabinoid Receptor 1 Antibody in Western Blot (WB)
          Cannabinoid Receptor 1 Antibody in Western Blot (WB)
          Cannabinoid Receptor 1 Antibody
          Cannabinoid Receptor 1 Antibody
          Cannabinoid Receptor 1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Cannabinoid Receptor 1 Recombinant Rabbit Monoclonal Antibody (PSH02-43)

          Product Details

          MA5-52083

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PSH02-43

          Immunogen

          Recombinant protein within human Cannabinoid receptor 1 aa 1-472.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 0.1% BSA, 40% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3099397

          Product Specific Information

          Tissue Specificity: Human brain tissue lysate, mouse brain tissue lysate, mouse cerebellum tissue lysate, rat brain tissue lysate, rat cerebellum tissue lysate, mouse brain tissue, mouse hippocampus tissue, rat brain tissue, rat hippocampus tissue.

          Positive Control: Human brain tissue lysate, mouse brain tissue lysate, mouse cerebellum tissue lysate, rat brain tissue lysate, rat cerebellum tissue lysate, mouse brain tissue, mouse hippocampus tissue, rat brain tissue, rat hippocampus tissue.

          Subcellular Location: Cell membrane, Membrane raft, Mitochondrion outer membrane, Cell projection, axon, Presynapse.

          Predicted band size: 53 kDa.

          Target Information

          This gene encodes one of two cannabinoid receptors. The cannabinoids, principally delta-9-tetrahydrocannabinol and synthetic analogs, are psychoactive ingredients of marijuana. The cannabinoid receptors are members of the guanine-nucleotide-binding protein coupled receptor family, which inhibit adenylate cyclase activity in a dose-dependent, stereoselective and pertussis toxin-sensitive manner. The two receptors have been found to be involved in the cannabinoid-induced CNS effects experienced by users of marijuana. Multiple transcript variants encoding two different protein isoforms have been described for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: brain-type cannabinoid receptor; Cannabinoid CB1 receptor; cannabinoid receptor 1 (brain); CB1 receptor; central cannabinoid receptor; HGNC:2159; N-terminal variant; OTTHUMP00000016839; RP1-23D17.1; striatal cannabinoid receptor type 1 protein; unnamed protein product

          View more View less

          Gene Aliases: CANN6; CB-R; CB1; CB1A; CB1B; CB1K5; CB1R; CNR; SKR6R

          View more View less

          UniProt ID: (Human) P21554, (Mouse) P47746, (Rat) P20272

          View more View less

          Entrez Gene ID: (Human) 1268, (Mouse) 12801, (Rat) 25248

          View more View less

          Function(s)
          G-protein coupled receptor activity cannabinoid receptor activity protein binding identical protein binding
          Process(es)
          G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway cannabinoid signaling pathway retrograde trans-synaptic signaling by endocannabinoid regulation of presynaptic cytosolic calcium ion concentration positive regulation of acute inflammatory response to antigenic stimulus axonal fasciculation learning or memory memory positive regulation of neuron projection development negative regulation of serotonin secretion regulation of lipid metabolic process positive regulation of fever generation negative regulation of fatty acid beta-oxidation regulation of synaptic transmission, GABAergic response to lipopolysaccharide negative regulation of mast cell activation negative regulation of dopamine secretion response to nicotine response to cocaine glucose homeostasis positive regulation of apoptotic process negative regulation of ion transport response to ethanol negative regulation of action potential negative regulation of blood pressure positive regulation of blood pressure regulation of insulin secretion negative regulation of nitric-oxide synthase activity regulation of synaptic transmission, glutamatergic regulation of feeding behavior regulation of penile erection trans-synaptic signaling by endocannabinoid regulation of long term synaptic depression spermatogenesis response to nutrient maternal process involved in female pregnancy
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nbmw6:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline