Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CDK4/CDK6 Antibodies

          Invitrogen

          CDK4/CDK6 Recombinant Rabbit Monoclonal Antibody (HL2323)

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CDK4/CDK6 Recombinant Rabbit Monoclonal Antibody (HL2323)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          CDK4/CDK6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CDK4/CDK6 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CDK4/CDK6 Antibody (MA5-56640) in WB

          Western Blot analysis of CDK4/CDK6 using CDK4/CDK6 Recombinant Monoclonal Antibody (Product # MA5-56640). Jurkat, Raji, and NCI-H929 whole cell extracts (30 μg) were separated by 10% SDS-PAGE, and the membrane was blotted with CDK4/CDK6 antibody diluted at 1:1,000. A HRP-conjugated anti-rabbit IgG antibody was used to detect the primary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CDK4/CDK6 Antibody in Western Blot (WB)
          CDK4/CDK6 Antibody in Western Blot (WB)
          CDK4/CDK6 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA5-56640

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          HL2323

          Immunogen

          Recombinant fragment of human CDK4 + CDK6.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          RRID

          AB_3679666

          Target Information

          CDK4 (Cyclin-Dependent Kinase 4) and CDK6 (Cyclin-Dependent Kinase 6) are crucial regulators of the cell cycle, specifically involved in the transition from the G1 phase to the S phase. These kinases form complexes with D-type cyclins (Cyclin D1, D2, and D3), which are necessary for their activation. Once activated, CDK4 and CDK6 phosphorylate the retinoblastoma protein (RB1), leading to the release of E2F transcription factors that drive the expression of genes required for DNA synthesis and cell cycle progression. Dysregulation of CDK4 and CDK6 activity is a common feature in various cancers, as it can lead to uncontrolled cell proliferation. Consequently, CDK4 and CDK6 have become important targets in cancer therapy, with inhibitors such as palbociclib, ribociclib, and abemaciclib being developed and approved for the treatment of certain types of cancer, particularly hormone receptor-positive breast cancer. These inhibitors work by blocking the kinase activity of CDK4 and CDK6, thereby halting cell cycle progression and reducing tumor growth.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CDK4; cell division protein kinase 4; cell division protein kinase 6; CR2 protein kinase; orf; p34/cdk4; PSK-J3; serine/threonine kinase; serine/threonine-protein kinase PLSTIRE; unnamed protein product

          View more View less

          Gene Aliases: CMM3; Crk2; Crk3; MCPH12; MCPH31; PLSTIRE; PSK-J3

          View more View less

          UniProt ID: (Human) P11802, (Human) Q00534, (Rat) P35426, (Mouse) P30285, (Mouse) Q64261

          View more View less

          Entrez Gene ID: (Human) 1019, (Human) 1021, (Rat) 94201, (Mouse) 12567, (Mouse) 12571, (Rat) 114483

          View more View less

          Function(s)
          protein kinase activity cyclin-dependent protein serine/threonine kinase activity protein binding ATP binding kinase activity cyclin-dependent protein serine/threonine kinase regulator activity cyclin binding protein serine kinase activity FBXO family protein binding macromolecular complex binding nucleotide binding
          Process(es)
          G1/S transition of mitotic cell cycle G2/M transition of mitotic cell cycle positive regulation of cell-matrix adhesion type B pancreatic cell development cellular response to DNA damage stimulus signal transduction negative regulation of cell proliferation response to virus regulation of gene expression astrocyte development dentate gyrus development lateral ventricle development hemopoiesis gliogenesis regulation of cell proliferation cell dedifferentiation negative regulation of cell differentiation negative regulation of myeloid cell differentiation regulation of erythrocyte differentiation negative regulation of monocyte differentiation negative regulation of osteoblast differentiation negative regulation of cell cycle positive regulation of fibroblast proliferation generation of neurons negative regulation of epithelial cell proliferation regulation of cell cycle regulation of cell motility negative regulation of cellular senescence negative regulation of transcription from RNA polymerase II promoter hematopoietic progenitor cell differentiation Notch signaling pathway positive regulation of gene expression T cell differentiation in thymus hematopoietic stem cell differentiation regulation of hematopoietic stem cell differentiation positive regulation of cell proliferation response to xenobiotic stimulus positive regulation of G2/M transition of mitotic cell cycle regulation of transcription initiation from RNA polymerase II promoter lens development in camera-type eye protein phosphorylation circadian rhythm response to toxic substance organ regeneration response to testosterone positive regulation of apoptotic process positive regulation of translation positive regulation of cell size positive regulation of epithelial cell proliferation response to hyperoxia cellular response to lipopolysaccharide cellular response to interleukin-4 cellular response to polyamine macromolecule cellular response to phorbol 13-acetate 12-myristate cellular response to ionomycin
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline