Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD22 Antibodies

          Invitrogen

          CD22 Recombinant Rabbit Monoclonal Antibody (3F2M10)

          View all (149) CD22 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD22 Recombinant Rabbit Monoclonal Antibody (3F2M10)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          CD22 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CD22 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD22 Antibody (MA5-58067) in WB

          Western blot analysis of CD22 in Raji, Ramos, and Jurkat (negative) (25 µg/lane). Samples were incubated overnight at 4°C with CD22 Recombinant Monoclonal Antibody (Product # MA5-58068) using a dilution of 1:3, 000. Followed by a HRP-conjugated Goat anti-Rabbit IgG (H+L) Secondary Antibody at 1:10,000 dilution. Blocking buffer: 3% nonfat dry milk in TBST; Negative control (NC): Jurkat; Exposure: 60 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD22 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CD22 Recombinant Rabbit Monoclonal Antibody (3F2M10)

          Product Details

          MA5-58067

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,500-1:3,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          3F2M10

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.57 mg/mL

          Amount

          31.4 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 0.05% BSA, 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          RRID

          AB_3681089

          Product Specific Information

          Immunogen sequence: DSSKWVFEHP ETLYAWEGA DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP

          Target Information

          CD22, also known as BL-CAM, is a type I transmembrane glycoprotein composed of two polypeptide chains, CD22alpha and CD22beta, with molecular weights of 130 and 140 kDa, respectively. These chains are produced by alternative splicing of the CD22 gene. CD22 is prominently expressed on mature B cells and B cell lymphomas, including hairy cell leukemia, diffuse large B-cell lymphoma, and nodular lymphocyte predominance Hodgkin's lymphoma, but is negative in classical Hodgkin's lymphoma. The extracellular portion of CD22 contains seven Ig-like domains that preferentially bind alpha2,6-linked sialic acid moieties found on epithelial, endothelial, B, and T cells. This binding can be masked by cis interactions with sialic acids on the same cell surface. CD22 expression is limited to late stages of B-cell differentiation, making it useful for phenotyping mature leukemias. Intracellularly, CD22 features six tyrosine residues within immunotyrosine-based inhibitory motifs (ITIM) and activation-like motifs. These residues are phosphorylated upon B-cell receptor engagement, allowing CD22 to regulate B-cell receptor signaling. CD22 participates in positive regulation through interactions with Src family tyrosine kinases and acts as an inhibitory receptor by recruiting cytoplasmic phosphatases via SH2 domains, which block signal transduction through dephosphorylation of signaling molecules. CD22's role in both positive and negative regulation of B-cell signaling, along with its specific expression pattern, makes it a valuable marker for antibody customers interested in B-cell-related research and diagnostics.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); BL-CAM; CD22 antigen; FLJ22814; Lectin 2; Leu-14; MGC130020; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); T-cell surface antigen Leu-14; unnamed protein product

          View more View less

          Gene Aliases: SIGLEC-2; SIGLEC2

          View more View less

          UniProt ID: (Human) P20273

          View more View less

          Entrez Gene ID: (Human) 933

          View more View less

          Function(s)
          IgM binding receptor binding protein binding protein phosphatase binding sialic acid binding CD4 receptor binding
          Process(es)
          negative regulation of immunoglobulin production negative regulation of immune system process cell adhesion negative regulation of signal transduction regulation of endocytosis regulation of B cell proliferation B cell activation regulation of immune response negative regulation of calcium-mediated signaling negative regulation of B cell receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0