Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NF2 Antibodies

          Invitrogen

          NF2 Recombinant Rabbit Monoclonal Antibody (4Q7P7)

          View all (31) NF2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NF2 Recombinant Rabbit Monoclonal Antibody (4Q7P7)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          NF2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NF2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NF2 Antibody (MA5-58077) in WB

          Western blot analysis of NF2 in MCF7, NIH/3T3, mouse brain, and C6 (25 µg/lane). Samples were incubated overnight at 4°C with NF2 Recombinant Monoclonal Antibody (Product # MA5-58078) using a dilution of 1:3, 000. Followed by a HRP-conjugated Goat anti-Rabbit IgG (H+L) Secondary Antibody at 1:10,000 dilution. Blocking buffer: 3% nonfat dry milk in TBST; Exposure: 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NF2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NF2 Recombinant Rabbit Monoclonal Antibody (4Q7P7)

          Product Details

          MA5-58077

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:3,000-1:12,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          4Q7P7

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 347-595 of human NF2/Merlin (NP_000259.1).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.98 mg/mL

          Amount

          39.6 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 0.05% BSA, 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          RRID

          AB_3681099

          Product Specific Information

          Immunogen sequence: EEAERTRDEL ERRLLQMKEE ATMANEALMR SEETADLLAE KAQITEEEAK LLAQKAAEAE QEMQRIKATA IRTEEEKRLM EQKVLEAEVL ALKMAEESER RAKEADQLKQ DLQEAREAER RAKQKLLEIA TKPTYPPMNP IPAPLPPDIP SFNLIGDSLS FDFKDTDMKR LSMEIEKEKV EYMEKSKHLQ EQLNELKTEI EALKLKERET ALDILHNENS DRGGSSKHNT IKKLTLQSAK SRVAFFEEL

          Target Information

          NF2 encodes a protein, Merlin, that is similar to some members of the ERM family of proteins that are thought to link cytoskeletal components with proteins in the cell membrane. Merlin has been shown to interact with cell-surface proteins, proteins involved in cytoskeletal dynamics and proteins involved in regulating ion transport. This gene is expressed at high levels during embryonic development; in adults, significant expression is found in Schwann cells, meningeal cells, lens and nerve. Mutations in this gene are associated with neurofibromatosis type II which is characterized by nervous system and skin tumors and ocular abnormalities. Two predominant isoforms and a number of minor isoforms are produced by alternatively spliced transcripts.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bilateral acoustic neurofibromatosis; MGC123124; moesin-ezrin-radixin like; moesin-ezrin-radixin-like protein; moesin-ezrin-radizin-like protein; neurofibromatosis 2; neurofibromatosis type 2; neurofibromin 2 (bilateral acoustic neuroma); neurofibromin-2; NF2; OTTHUMP00000199607; schwannomerlin; schwannomin; tumor suppressor; tumor suppressor; bilateral acoustic neuroma; Schwannomin/Merlin/Schwannomerlin; unnamed protein product

          View more View less

          Gene Aliases: ACN; BANF; merlin; merlin-1; SCH; SWNV

          View more View less

          UniProt ID: (Human) P35240, (Mouse) P46662, (Rat) Q63648

          View more View less

          Entrez Gene ID: (Human) 4771, (Mouse) 18016, (Rat) 25744

          View more View less

          Function(s)
          actin binding integrin binding protein binding cytoskeletal protein binding signaling adaptor activity beta-catenin binding protein domain specific binding
          Process(es)
          mesoderm formation negative regulation of protein kinase activity ectoderm development brain development negative regulation of cell proliferation regulation of cell shape Schwann cell proliferation regulation of gliogenesis hippocampus development negative regulation of cell-cell adhesion actin cytoskeleton organization negative regulation of cell growth regulation of protein stability hippo signaling regulation of hippo signaling regulation of cell proliferation odontogenesis of dentin-containing tooth regulation of apoptotic process negative regulation of MAPK cascade cell-cell junction organization positive regulation of cell differentiation negative regulation of JAK-STAT cascade regulation of neurogenesis positive regulation of stress fiber assembly regulation of cell cycle negative regulation of cell growth involved in contact inhibition lens fiber cell differentiation regulation of stem cell proliferation regulation of protein localization to nucleus regulation of organelle assembly positive regulation of protein localization to early endosome regulation of neural precursor cell proliferation positive regulation of early endosome to late endosome transport negative regulation of cell-matrix adhesion cell differentiation negative regulation of cell migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0